Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26941713-26942209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL5
Entrez Gene ID
56896
Gene Name
dihydropyrimidinase like 5
Gene Aliases
CRAM, CRMP-5, CRMP5, Ulip6
UniGene
Hs.299315
Gene Chromosome Location
Chr.2: 26848101-26950351 on Build GRCh38
Transcripts
NM_001253723.1 NM_001253724.1 NM_020134.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3580462 c.1105G>A p.D369N Chr.2: 26941965-26941965 on Build GRCh38
5922156 c.1104G>A p.M368I Chr.2: 26941964-26941964 on Build GRCh38
4616857 c.1124C>T p.A375V Chr.2: 26941984-26941984 on Build GRCh38
4589750 c.1229_1230insA p.T412fs*17 Chr.2: 26942089-26942090 on Build GRCh38
1741730 c.1203T>A p.D401E Chr.2: 26942063-26942063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200568891 Chr.2: 26941999-26941999 on Build GRCh38
rs760317762 Chr.2: 26942098-26942098 on Build GRCh38
rs1029233058 Chr.2: 26941814-26941814 on Build GRCh38
rs921000313 Chr.2: 26942118-26942118 on Build GRCh38
rs138627302 Chr.2: 26942019-26942019 on Build GRCh38
rs372069126 Chr.2: 26942030-26942030 on Build GRCh38
rs924271830 Chr.2: 26942069-26942069 on Build GRCh38
rs974672689 Chr.2: 26942006-26942006 on Build GRCh38
rs3820823 Chr.2: 26941938-26941938 on Build GRCh38
rs755806141 Chr.2: 26942049-26942049 on Build GRCh38
rs763171947 Chr.2: 26941816-26941816 on Build GRCh38
rs41288787 Chr.2: 26941986-26941986 on Build GRCh38
rs766604944 Chr.2: 26942113-26942113 on Build GRCh38
rs185472268 Chr.2: 26942040-26942040 on Build GRCh38
rs113982137 Chr.2: 26941913-26941913 on Build GRCh38
rs761859712 Chr.2: 26941989-26941989 on Build GRCh38
rs769293152 Chr.2: 26941983-26941983 on Build GRCh38
rs765314115 Chr.2: 26942025-26942025 on Build GRCh38
rs973459018 Chr.2: 26941985-26941985 on Build GRCh38
rs569396889 Chr.2: 26942182-26942182 on Build GRCh38
rs755567069 Chr.2: 26942131-26942131 on Build GRCh38
rs770862625 Chr.2: 26941992-26941992 on Build GRCh38
rs775365699 Chr.2: 26942082-26942082 on Build GRCh38
rs773832704 Chr.2: 26942055-26942055 on Build GRCh38
rs747513449 Chr.2: 26941975-26941975 on Build GRCh38
rs201235120 Chr.2: 26941974-26941974 on Build GRCh38
rs767298253 Chr.2: 26941909-26941909 on Build GRCh38
rs773363999 Chr.2: 26942109-26942109 on Build GRCh38
rs1053895964 Chr.2: 26941801-26941801 on Build GRCh38
rs906703604 Chr.2: 26941818-26941818 on Build GRCh38
rs758953762 Chr.2: 26941973-26941973 on Build GRCh38
rs187940086 Chr.2: 26942108-26942108 on Build GRCh38
rs1009315797 Chr.2: 26941911-26941911 on Build GRCh38
rs748412363 Chr.2: 26942048-26942048 on Build GRCh38
rs750517458 Chr.2: 26942028-26942028 on Build GRCh38
rs767351583 Chr.2: 26942001-26942001 on Build GRCh38
rs752159624 Chr.2: 26942037-26942037 on Build GRCh38
rs202116000 Chr.2: 26942054-26942054 on Build GRCh38
rs890710699 Chr.2: 26941849-26941849 on Build GRCh38
rs766178174 Chr.2: 26941899-26941899 on Build GRCh38
rs745834449 Chr.2: 26942066-26942066 on Build GRCh38
rs771945767 Chr.2: 26942080-26942080 on Build GRCh38
rs765339264 Chr.2: 26942134-26942135 on Build GRCh38
rs781671833 Chr.2: 26941982-26941982 on Build GRCh38
rs954523709 Chr.2: 26942027-26942027 on Build GRCh38
rs1045288755 Chr.2: 26942171-26942171 on Build GRCh38
rs759458186 Chr.2: 26942000-26942000 on Build GRCh38
rs947746217 Chr.2: 26942166-26942166 on Build GRCh38
rs893953175 Chr.2: 26941804-26941804 on Build GRCh38
rs755509744 Chr.2: 26942038-26942038 on Build GRCh38
rs1003706577 Chr.2: 26941854-26941854 on Build GRCh38
rs1041383053 Chr.2: 26941741-26941741 on Build GRCh38
rs768079923 Chr.2: 26942133-26942133 on Build GRCh38
rs991314942 Chr.2: 26942026-26942026 on Build GRCh38
rs752445689 Chr.2: 26942009-26942009 on Build GRCh38
rs751651395 Chr.2: 26942116-26942116 on Build GRCh38
rs935729950 Chr.2: 26941752-26941752 on Build GRCh38
rs1003631277 Chr.2: 26942183-26942183 on Build GRCh38
rs758472887 Chr.2: 26942029-26942029 on Build GRCh38
rs753772127 Chr.2: 26942170-26942170 on Build GRCh38
rs753111239 Chr.2: 26942144-26942144 on Build GRCh38
rs764257438 Chr.2: 26941908-26941908 on Build GRCh38
rs769199565 Chr.2: 26941977-26941977 on Build GRCh38
rs566417952 Chr.2: 26941837-26941837 on Build GRCh38
rs751373377 Chr.2: 26941901-26941901 on Build GRCh38
rs77970849 Chr.2: 26941987-26941987 on Build GRCh38
rs547995668 Chr.2: 26941774-26941774 on Build GRCh38
rs757388162 Chr.2: 26941910-26941910 on Build GRCh38
rs996284200 Chr.2: 26941810-26941810 on Build GRCh38

View Full Product Details