Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27055597-27056107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_001035507.2 NM_021831.5 NR_104246.1 XM_005264477.2 XM_011533011.2 XM_011533012.2 XM_011533013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5396776 c.1010A>G p.H337R Chr.2: 27055783-27055783 on Build GRCh38
5647097 c.1260C>G p.I420M Chr.2: 27056033-27056033 on Build GRCh38
5474469 c.920G>A p.R307H Chr.2: 27055693-27055693 on Build GRCh38
7331644 c.967C>A p.H323N Chr.2: 27055740-27055740 on Build GRCh38
5647096 c.1260C>G p.I420M Chr.2: 27056033-27056033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757938018 Chr.2: 27055926-27055926 on Build GRCh38
rs766381819 Chr.2: 27055720-27055720 on Build GRCh38
rs778625808 Chr.2: 27055822-27055822 on Build GRCh38
rs921522314 Chr.2: 27055837-27055837 on Build GRCh38
rs777264819 Chr.2: 27055640-27055640 on Build GRCh38
rs1050928731 Chr.2: 27056066-27056066 on Build GRCh38
rs757989531 Chr.2: 27056034-27056034 on Build GRCh38
rs373726260 Chr.2: 27055987-27055987 on Build GRCh38
rs768511974 Chr.2: 27055931-27055931 on Build GRCh38
rs899744662 Chr.2: 27055670-27055670 on Build GRCh38
rs201152933 Chr.2: 27055888-27055888 on Build GRCh38
rs759643795 Chr.2: 27055997-27055997 on Build GRCh38
rs754682228 Chr.2: 27055941-27055944 on Build GRCh38
rs772072440 Chr.2: 27056002-27056002 on Build GRCh38
rs766193452 Chr.2: 27055744-27055744 on Build GRCh38
rs772125178 Chr.2: 27055679-27055679 on Build GRCh38
rs766582593 Chr.2: 27055879-27055879 on Build GRCh38
rs924806973 Chr.2: 27055694-27055694 on Build GRCh38
rs774432313 Chr.2: 27055657-27055657 on Build GRCh38
rs760360747 Chr.2: 27055692-27055692 on Build GRCh38
rs762247398 Chr.2: 27056014-27056014 on Build GRCh38
rs550872992 Chr.2: 27055733-27055733 on Build GRCh38
rs768297587 Chr.2: 27055859-27055859 on Build GRCh38
rs764659580 Chr.2: 27056005-27056005 on Build GRCh38
rs912485968 Chr.2: 27056074-27056074 on Build GRCh38
rs753109711 Chr.2: 27055751-27055751 on Build GRCh38
rs772180234 Chr.2: 27055829-27055829 on Build GRCh38
rs200469656 Chr.2: 27055889-27055889 on Build GRCh38
rs746031691 Chr.2: 27056062-27056062 on Build GRCh38
rs1039465928 Chr.2: 27055664-27055664 on Build GRCh38
rs932566108 Chr.2: 27056041-27056041 on Build GRCh38
rs557002163 Chr.2: 27055669-27055669 on Build GRCh38
rs529046053 Chr.2: 27055718-27055718 on Build GRCh38
rs778680779 Chr.2: 27055910-27055910 on Build GRCh38
rs781019965 Chr.2: 27055961-27055961 on Build GRCh38
rs35445642 Chr.2: 27055882-27055882 on Build GRCh38
rs374692761 Chr.2: 27055929-27055929 on Build GRCh38
rs780913358 Chr.2: 27055954-27055954 on Build GRCh38
rs140111286 Chr.2: 27056038-27056038 on Build GRCh38
rs990277706 Chr.2: 27055784-27055784 on Build GRCh38
rs966883191 Chr.2: 27055667-27055667 on Build GRCh38
rs745370626 Chr.2: 27055826-27055826 on Build GRCh38
rs748822860 Chr.2: 27055820-27055820 on Build GRCh38
rs750002918 Chr.2: 27055924-27055924 on Build GRCh38
rs779110735 Chr.2: 27056052-27056052 on Build GRCh38
rs373237623 Chr.2: 27056039-27056039 on Build GRCh38
rs140050281 Chr.2: 27055745-27055745 on Build GRCh38
rs149553528 Chr.2: 27055693-27055693 on Build GRCh38
rs773489159 Chr.2: 27055863-27055863 on Build GRCh38
rs187389989 Chr.2: 27056037-27056037 on Build GRCh38
rs201486305 Chr.2: 27055793-27055793 on Build GRCh38
rs750064297 Chr.2: 27056023-27056023 on Build GRCh38
rs754195407 Chr.2: 27056012-27056012 on Build GRCh38
rs535101915 Chr.2: 27055901-27055901 on Build GRCh38
rs745744385 Chr.2: 27055677-27055677 on Build GRCh38
rs754219257 Chr.2: 27055903-27055903 on Build GRCh38
rs912547580 Chr.2: 27055914-27055914 on Build GRCh38
rs774504166 Chr.2: 27055995-27055995 on Build GRCh38
rs775550831 Chr.2: 27055839-27055839 on Build GRCh38
rs1013680987 Chr.2: 27055646-27055646 on Build GRCh38
rs761101955 Chr.2: 27055898-27055898 on Build GRCh38
rs377620315 Chr.2: 27055869-27055869 on Build GRCh38
rs898322571 Chr.2: 27055621-27055621 on Build GRCh38
rs1023076790 Chr.2: 27055841-27055841 on Build GRCh38
rs747733044 Chr.2: 27056032-27056033 on Build GRCh38
rs756929559 Chr.2: 27055907-27055907 on Build GRCh38
rs370334733 Chr.2: 27055955-27055955 on Build GRCh38
rs751058065 Chr.2: 27055644-27055644 on Build GRCh38
rs770304728 Chr.2: 27055648-27055648 on Build GRCh38
rs1025088650 Chr.2: 27056051-27056051 on Build GRCh38
rs909298115 Chr.2: 27055639-27055639 on Build GRCh38
rs1033556945 Chr.2: 27055631-27055633 on Build GRCh38
rs773259169 Chr.2: 27055697-27055697 on Build GRCh38
rs995419012 Chr.2: 27055625-27055625 on Build GRCh38
rs1035947182 Chr.2: 27055917-27055917 on Build GRCh38
rs372287130 Chr.2: 27055641-27055641 on Build GRCh38
rs756801984 Chr.2: 27055821-27055821 on Build GRCh38
rs113809750 Chr.2: 27055928-27055928 on Build GRCh38
rs765004279 Chr.2: 27056022-27056022 on Build GRCh38
rs755544280 Chr.2: 27055734-27055734 on Build GRCh38
rs963822087 Chr.2: 27056020-27056020 on Build GRCh38
rs372015397 Chr.2: 27055659-27055659 on Build GRCh38
rs1011217644 Chr.2: 27055828-27055828 on Build GRCh38
rs942014296 Chr.2: 27055660-27055660 on Build GRCh38
rs751370407 Chr.2: 27055845-27055845 on Build GRCh38
rs780953495 Chr.2: 27055947-27055962 on Build GRCh38
rs368050370 Chr.2: 27056003-27056003 on Build GRCh38
rs143744999 Chr.2: 27055794-27055794 on Build GRCh38
rs757182855 Chr.2: 27055881-27055881 on Build GRCh38

View Full Product Details