Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27277672-27278168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5G
Entrez Gene ID
285126
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Gene Aliases
CSP-gamma
UniGene
Hs.116303
Gene Chromosome Location
Chr.2: 27275421-27281430 on Build GRCh38
Transcripts
NM_001303127.1 NM_001303128.1 NM_173650.2 NR_130126.1 XM_006711995.3 XM_017003872.1 XM_017003873.1 XM_017003874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5785072 c.157G>T p.G53C Chr.2: 27277797-27277797 on Build GRCh38
335622 c.175C>T p.R59W Chr.2: 27277815-27277815 on Build GRCh38
5676160 c.141T>A p.P47P Chr.2: 27277781-27277781 on Build GRCh38
2939721 c.150T>A p.Y50* Chr.2: 27277790-27277790 on Build GRCh38
5659189 c.182A>G p.H61R Chr.2: 27277822-27277822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776496456 Chr.2: 27278008-27278008 on Build GRCh38
rs375265383 Chr.2: 27277842-27277842 on Build GRCh38
rs767221432 Chr.2: 27277707-27277707 on Build GRCh38
rs749481022 Chr.2: 27277954-27277954 on Build GRCh38
rs752457242 Chr.2: 27277797-27277797 on Build GRCh38
rs563876708 Chr.2: 27277786-27277786 on Build GRCh38
rs774670847 Chr.2: 27277782-27277782 on Build GRCh38
rs778017118 Chr.2: 27277951-27277951 on Build GRCh38
rs762487080 Chr.2: 27278027-27278027 on Build GRCh38
rs1018678378 Chr.2: 27278077-27278077 on Build GRCh38
rs374152945 Chr.2: 27278016-27278016 on Build GRCh38
rs753617355 Chr.2: 27277780-27277781 on Build GRCh38
rs749293959 Chr.2: 27277731-27277731 on Build GRCh38
rs745899221 Chr.2: 27277977-27277977 on Build GRCh38
rs766351207 Chr.2: 27277930-27277930 on Build GRCh38
rs539866520 Chr.2: 27277815-27277815 on Build GRCh38
rs368248450 Chr.2: 27277877-27277877 on Build GRCh38
rs987988312 Chr.2: 27277698-27277698 on Build GRCh38
rs1002394264 Chr.2: 27278142-27278142 on Build GRCh38
rs769114019 Chr.2: 27277999-27277999 on Build GRCh38
rs149346046 Chr.2: 27277960-27277960 on Build GRCh38
rs185030275 Chr.2: 27277976-27277976 on Build GRCh38
rs777930417 Chr.2: 27277843-27277843 on Build GRCh38
rs776098513 Chr.2: 27277985-27277985 on Build GRCh38
rs961044239 Chr.2: 27277893-27277893 on Build GRCh38
rs764886826 Chr.2: 27278026-27278026 on Build GRCh38
rs758282251 Chr.2: 27277923-27277923 on Build GRCh38
rs755721541 Chr.2: 27277721-27277721 on Build GRCh38
rs141546081 Chr.2: 27277827-27277827 on Build GRCh38
rs533193936 Chr.2: 27278092-27278092 on Build GRCh38
rs192714047 Chr.2: 27277761-27277761 on Build GRCh38
rs993619798 Chr.2: 27278055-27278055 on Build GRCh38
rs888762270 Chr.2: 27278014-27278014 on Build GRCh38
rs771745813 Chr.2: 27277740-27277740 on Build GRCh38
rs774962567 Chr.2: 27277885-27277885 on Build GRCh38
rs770009682 Chr.2: 27277833-27277833 on Build GRCh38
rs777414817 Chr.2: 27277724-27277724 on Build GRCh38
rs779976641 Chr.2: 27277925-27277925 on Build GRCh38
rs771457236 Chr.2: 27277771-27277771 on Build GRCh38
rs772541173 Chr.2: 27277899-27277899 on Build GRCh38
rs183643555 Chr.2: 27277801-27277801 on Build GRCh38
rs866801938 Chr.2: 27277902-27277902 on Build GRCh38
rs967939611 Chr.2: 27278137-27278137 on Build GRCh38
rs755257029 Chr.2: 27277703-27277703 on Build GRCh38
rs149067267 Chr.2: 27278108-27278108 on Build GRCh38
rs143064063 Chr.2: 27277957-27277957 on Build GRCh38
rs759951998 Chr.2: 27277898-27277898 on Build GRCh38
rs761452920 Chr.2: 27277914-27277914 on Build GRCh38
rs766247645 Chr.2: 27278028-27278028 on Build GRCh38
rs199500100 Chr.2: 27278105-27278105 on Build GRCh38
rs560209092 Chr.2: 27278075-27278075 on Build GRCh38
rs946714489 Chr.2: 27277927-27277927 on Build GRCh38
rs756976376 Chr.2: 27277879-27277883 on Build GRCh38
rs778677815 Chr.2: 27277736-27277736 on Build GRCh38
rs1043542218 Chr.2: 27277712-27277712 on Build GRCh38
rs748364890 Chr.2: 27277760-27277760 on Build GRCh38
rs550880968 Chr.2: 27277912-27277912 on Build GRCh38
rs149024695 Chr.2: 27277809-27277809 on Build GRCh38
rs764653726 Chr.2: 27277920-27277920 on Build GRCh38
rs757433308 Chr.2: 27278140-27278140 on Build GRCh38
rs756003390 Chr.2: 27277879-27277879 on Build GRCh38
rs13414011 Chr.2: 27277792-27277792 on Build GRCh38
rs370045452 Chr.2: 27277819-27277819 on Build GRCh38
rs767294388 Chr.2: 27278060-27278060 on Build GRCh38
rs551535393 Chr.2: 27277929-27277929 on Build GRCh38
rs751475483 Chr.2: 27278031-27278031 on Build GRCh38
rs752268950 Chr.2: 27277708-27277708 on Build GRCh38
rs770624109 Chr.2: 27277911-27277911 on Build GRCh38
rs192512997 Chr.2: 27277910-27277910 on Build GRCh38
rs754807610 Chr.2: 27278043-27278043 on Build GRCh38
rs370298588 Chr.2: 27277735-27277735 on Build GRCh38
rs144758776 Chr.2: 27278015-27278015 on Build GRCh38
rs779701255 Chr.2: 27277826-27277826 on Build GRCh38
rs747429598 Chr.2: 27277987-27277987 on Build GRCh38
rs374436385 Chr.2: 27277992-27277992 on Build GRCh38
rs781520564 Chr.2: 27277747-27277747 on Build GRCh38
rs370889514 Chr.2: 27277822-27277822 on Build GRCh38
rs374461251 Chr.2: 27277882-27277882 on Build GRCh38
rs767741472 Chr.2: 27277794-27277794 on Build GRCh38
rs145782994 Chr.2: 27277876-27277876 on Build GRCh38
rs772458896 Chr.2: 27277984-27277984 on Build GRCh38
rs878867778 Chr.2: 27277770-27277770 on Build GRCh38
rs754717720 Chr.2: 27277933-27277933 on Build GRCh38
rs750432820 Chr.2: 27277922-27277922 on Build GRCh38
rs371768076 Chr.2: 27278064-27278064 on Build GRCh38
rs778962253 Chr.2: 27277817-27277817 on Build GRCh38
rs756781840 Chr.2: 27277816-27277816 on Build GRCh38
rs763214668 Chr.2: 27277706-27277707 on Build GRCh38
rs779123700 Chr.2: 27278143-27278143 on Build GRCh38
rs937385182 Chr.2: 27277766-27277766 on Build GRCh38
rs866747487 Chr.2: 27277779-27277780 on Build GRCh38
rs946625429 Chr.2: 27277725-27277725 on Build GRCh38

View Full Product Details