Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29017195-29017741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179A
Entrez Gene ID
165186
Gene Name
family with sequence similarity 179 member A
Gene Aliases
-
UniGene
Hs.525977
Gene Chromosome Location
Chr.2: 28981298-29052230 on Build GRCh38
Transcripts
NM_001321538.1 NM_001321539.1 NM_199280.2 XM_011532628.2 XM_011532631.1 XM_017003507.1 XM_017003508.1 XM_017003509.1 XM_017003510.1 XM_017003511.1 XM_017003512.1 XM_017003514.1 XM_017003516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437791 c.1157C>A p.S386Y Chr.2: 29017266-29017266 on Build GRCh38
5524943 c.1195+1G>A p.? Chr.2: 29017305-29017305 on Build GRCh38
1284831 c.1174C>T p.Q392* Chr.2: 29017283-29017283 on Build GRCh38
721302 c.1121G>T p.R374M Chr.2: 29017230-29017230 on Build GRCh38
5042903 c.1139C>T p.T380I Chr.2: 29017248-29017248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965430818 Chr.2: 29017412-29017412 on Build GRCh38
rs72788138 Chr.2: 29017600-29017600 on Build GRCh38
rs995810969 Chr.2: 29017696-29017696 on Build GRCh38
rs763883767 Chr.2: 29017259-29017259 on Build GRCh38
rs757946364 Chr.2: 29017272-29017272 on Build GRCh38
rs536681472 Chr.2: 29017663-29017663 on Build GRCh38
rs929687474 Chr.2: 29017547-29017547 on Build GRCh38
rs910822307 Chr.2: 29017246-29017246 on Build GRCh38
rs774471433 Chr.2: 29017223-29017223 on Build GRCh38
rs182571022 Chr.2: 29017557-29017557 on Build GRCh38
rs1046830457 Chr.2: 29017550-29017550 on Build GRCh38
rs1045343544 Chr.2: 29017564-29017564 on Build GRCh38
rs1030394651 Chr.2: 29017447-29017447 on Build GRCh38
rs543517815 Chr.2: 29017653-29017653 on Build GRCh38
rs751163226 Chr.2: 29017282-29017282 on Build GRCh38
rs553034298 Chr.2: 29017662-29017662 on Build GRCh38
rs768749498 Chr.2: 29017337-29017337 on Build GRCh38
rs1033575893 Chr.2: 29017317-29017317 on Build GRCh38
rs537934904 Chr.2: 29017402-29017402 on Build GRCh38
rs1009622763 Chr.2: 29017414-29017414 on Build GRCh38
rs963634090 Chr.2: 29017679-29017679 on Build GRCh38
rs938404105 Chr.2: 29017603-29017603 on Build GRCh38
rs143775385 Chr.2: 29017560-29017560 on Build GRCh38
rs1010028017 Chr.2: 29017273-29017273 on Build GRCh38
rs772222478 Chr.2: 29017227-29017227 on Build GRCh38
rs200034953 Chr.2: 29017345-29017345 on Build GRCh38
rs1007686816 Chr.2: 29017674-29017674 on Build GRCh38
rs973729696 Chr.2: 29017539-29017539 on Build GRCh38
rs151077354 Chr.2: 29017708-29017708 on Build GRCh38
rs13014013 Chr.2: 29017521-29017521 on Build GRCh38
rs114252468 Chr.2: 29017357-29017357 on Build GRCh38
rs1041441895 Chr.2: 29017324-29017324 on Build GRCh38
rs202125633 Chr.2: 29017268-29017268 on Build GRCh38
rs541016389 Chr.2: 29017685-29017685 on Build GRCh38
rs765247449 Chr.2: 29017267-29017267 on Build GRCh38
rs1027639567 Chr.2: 29017705-29017705 on Build GRCh38
rs776619466 Chr.2: 29017341-29017341 on Build GRCh38
rs576870685 Chr.2: 29017677-29017677 on Build GRCh38
rs779592142 Chr.2: 29017275-29017275 on Build GRCh38
rs921234404 Chr.2: 29017540-29017540 on Build GRCh38
rs13409364 Chr.2: 29017583-29017583 on Build GRCh38
rs66482868 Chr.2: 29017716-29017716 on Build GRCh38
rs779360500 Chr.2: 29017321-29017321 on Build GRCh38
rs201148338 Chr.2: 29017265-29017265 on Build GRCh38
rs11899716 Chr.2: 29017406-29017406 on Build GRCh38
rs1002475223 Chr.2: 29017332-29017332 on Build GRCh38
rs371456773 Chr.2: 29017325-29017325 on Build GRCh38
rs201395358 Chr.2: 29017219-29017219 on Build GRCh38
rs756437206 Chr.2: 29017289-29017289 on Build GRCh38
rs148998008 Chr.2: 29017551-29017551 on Build GRCh38
rs780776946 Chr.2: 29017336-29017336 on Build GRCh38
rs1018196671 Chr.2: 29017675-29017675 on Build GRCh38
rs749638434 Chr.2: 29017295-29017295 on Build GRCh38
rs898204581 Chr.2: 29017279-29017279 on Build GRCh38
rs1027154166 Chr.2: 29017369-29017369 on Build GRCh38
rs954860843 Chr.2: 29017456-29017456 on Build GRCh38
rs13013823 Chr.2: 29017453-29017453 on Build GRCh38
rs368163808 Chr.2: 29017598-29017598 on Build GRCh38
rs771422334 Chr.2: 29017221-29017221 on Build GRCh38
rs572324239 Chr.2: 29017407-29017407 on Build GRCh38
rs906978842 Chr.2: 29017601-29017601 on Build GRCh38
rs910801554 Chr.2: 29017485-29017485 on Build GRCh38
rs761673048 Chr.2: 29017342-29017342 on Build GRCh38
rs905144710 Chr.2: 29017392-29017392 on Build GRCh38
rs963967210 Chr.2: 29017512-29017512 on Build GRCh38
rs775580881 Chr.2: 29017331-29017331 on Build GRCh38
rs919605036 Chr.2: 29017543-29017543 on Build GRCh38
rs200945881 Chr.2: 29017226-29017226 on Build GRCh38
rs755871112 Chr.2: 29017475-29017475 on Build GRCh38
rs892446311 Chr.2: 29017611-29017611 on Build GRCh38
rs145856602 Chr.2: 29017502-29017502 on Build GRCh38
rs986736794 Chr.2: 29017473-29017473 on Build GRCh38
rs555242621 Chr.2: 29017416-29017416 on Build GRCh38
rs750332223 Chr.2: 29017353-29017353 on Build GRCh38
rs929719899 Chr.2: 29017310-29017310 on Build GRCh38
rs951626246 Chr.2: 29017377-29017377 on Build GRCh38
rs760401640 Chr.2: 29017251-29017251 on Build GRCh38
rs990471170 Chr.2: 29017568-29017568 on Build GRCh38
rs775733626 Chr.2: 29017240-29017240 on Build GRCh38
rs539972767 Chr.2: 29017239-29017239 on Build GRCh38
rs771475670 Chr.2: 29017314-29017314 on Build GRCh38
rs61750881 Chr.2: 29017266-29017266 on Build GRCh38
rs1009550639 Chr.2: 29017647-29017647 on Build GRCh38
rs386644311 Chr.2: 29017265-29017266 on Build GRCh38
rs949192688 Chr.2: 29017552-29017552 on Build GRCh38
rs772132921 Chr.2: 29017327-29017327 on Build GRCh38

View Full Product Details