Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32401331-32401880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
UniGene
Hs.150107
Gene Chromosome Location
Chr.2: 32356907-32618899 on Build GRCh38
Transcripts
NM_016252.3 XM_005264449.4 XM_005264450.4 XM_005264451.4 XM_005264452.4 XM_005264453.4 XM_005264454.4 XM_005264455.4 XM_006712054.3 XM_006712055.3 XM_006712056.3 XM_011533003.2 XM_011533005.2 XM_017004556.1 XM_017004557.1 XM_017004558.1 XM_017004559.1 XM_017004560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3991271 c.1306C>T p.Q436* Chr.2: 32401511-32401511 on Build GRCh38
4583472 c.1378T>C p.S460P Chr.2: 32401583-32401583 on Build GRCh38
1020203 c.1224G>T p.K408N Chr.2: 32401352-32401352 on Build GRCh38
7103012 c.1319C>T p.S440L Chr.2: 32401524-32401524 on Build GRCh38
3939073 c.1216C>T p.R406* Chr.2: 32401344-32401344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139494254 Chr.2: 32401557-32401557 on Build GRCh38
rs950787769 Chr.2: 32401848-32401848 on Build GRCh38
rs369443191 Chr.2: 32401399-32401399 on Build GRCh38
rs964749971 Chr.2: 32401791-32401791 on Build GRCh38
rs954601032 Chr.2: 32401449-32401449 on Build GRCh38
rs200691665 Chr.2: 32401673-32401673 on Build GRCh38
rs138222460 Chr.2: 32401507-32401507 on Build GRCh38
rs768731818 Chr.2: 32401652-32401652 on Build GRCh38
rs760508325 Chr.2: 32401577-32401577 on Build GRCh38
rs569421445 Chr.2: 32401727-32401727 on Build GRCh38
rs755254977 Chr.2: 32401359-32401359 on Build GRCh38
rs988096981 Chr.2: 32401428-32401428 on Build GRCh38
rs199598287 Chr.2: 32401655-32401655 on Build GRCh38
rs746738525 Chr.2: 32401647-32401647 on Build GRCh38
rs1017412452 Chr.2: 32401751-32401751 on Build GRCh38
rs888307397 Chr.2: 32401842-32401842 on Build GRCh38
rs746057727 Chr.2: 32401617-32401617 on Build GRCh38
rs377237765 Chr.2: 32401410-32401410 on Build GRCh38
rs767759482 Chr.2: 32401569-32401569 on Build GRCh38
rs781072196 Chr.2: 32401388-32401389 on Build GRCh38
rs551433069 Chr.2: 32401755-32401755 on Build GRCh38
rs1001836468 Chr.2: 32401415-32401415 on Build GRCh38
rs756617915 Chr.2: 32401583-32401583 on Build GRCh38
rs779237403 Chr.2: 32401601-32401601 on Build GRCh38
rs910604212 Chr.2: 32401480-32401480 on Build GRCh38
rs115882868 Chr.2: 32401654-32401654 on Build GRCh38
rs1044592030 Chr.2: 32401632-32401632 on Build GRCh38
rs200372255 Chr.2: 32401412-32401412 on Build GRCh38
rs568997793 Chr.2: 32401721-32401721 on Build GRCh38
rs973354758 Chr.2: 32401540-32401540 on Build GRCh38
rs370670186 Chr.2: 32401653-32401653 on Build GRCh38
rs763769012 Chr.2: 32401511-32401511 on Build GRCh38
rs929534924 Chr.2: 32401566-32401566 on Build GRCh38
rs777606336 Chr.2: 32401745-32401745 on Build GRCh38
rs13033581 Chr.2: 32401549-32401549 on Build GRCh38
rs537823403 Chr.2: 32401725-32401725 on Build GRCh38
rs778194980 Chr.2: 32401584-32401584 on Build GRCh38
rs924330671 Chr.2: 32401408-32401408 on Build GRCh38
rs775424222 Chr.2: 32401627-32401627 on Build GRCh38
rs748098528 Chr.2: 32401657-32401658 on Build GRCh38
rs781741200 Chr.2: 32401375-32401375 on Build GRCh38
rs1020907988 Chr.2: 32401718-32401718 on Build GRCh38
rs1021191965 Chr.2: 32401429-32401429 on Build GRCh38
rs533804594 Chr.2: 32401807-32401807 on Build GRCh38
rs765988258 Chr.2: 32401663-32401663 on Build GRCh38
rs987294570 Chr.2: 32401732-32401732 on Build GRCh38
rs895970680 Chr.2: 32401604-32401604 on Build GRCh38
rs1057404409 Chr.2: 32401761-32401761 on Build GRCh38
rs1032023539 Chr.2: 32401416-32401416 on Build GRCh38
rs914332265 Chr.2: 32401371-32401371 on Build GRCh38
rs762506703 Chr.2: 32401661-32401661 on Build GRCh38
rs1013745074 Chr.2: 32401376-32401376 on Build GRCh38
rs920588595 Chr.2: 32401809-32401809 on Build GRCh38
rs201426570 Chr.2: 32401621-32401621 on Build GRCh38
rs755234690 Chr.2: 32401661-32401662 on Build GRCh38
rs987956258 Chr.2: 32401456-32401456 on Build GRCh38
rs370521293 Chr.2: 32401418-32401418 on Build GRCh38
rs745708744 Chr.2: 32401465-32401465 on Build GRCh38
rs1048209606 Chr.2: 32401825-32401825 on Build GRCh38
rs753269227 Chr.2: 32401582-32401582 on Build GRCh38
rs749393381 Chr.2: 32401589-32401589 on Build GRCh38
rs977222665 Chr.2: 32401687-32401687 on Build GRCh38
rs771735619 Chr.2: 32401502-32401502 on Build GRCh38
rs774387913 Chr.2: 32401453-32401453 on Build GRCh38
rs773288158 Chr.2: 32401550-32401550 on Build GRCh38
rs374523128 Chr.2: 32401405-32401405 on Build GRCh38
rs887862067 Chr.2: 32401505-32401505 on Build GRCh38
rs904913646 Chr.2: 32401639-32401639 on Build GRCh38
rs760679474 Chr.2: 32401509-32401509 on Build GRCh38
rs757246195 Chr.2: 32401592-32401592 on Build GRCh38
rs968444164 Chr.2: 32401675-32401675 on Build GRCh38
rs755417574 Chr.2: 32401568-32401568 on Build GRCh38
rs771154958 Chr.2: 32401433-32401433 on Build GRCh38
rs776291129 Chr.2: 32401545-32401545 on Build GRCh38

View Full Product Details