Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36499077-36499575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635495 c.1440G>T p.K480N Chr.2: 36499286-36499286 on Build GRCh38
5953198 c.1373-1G>C p.? Chr.2: 36499218-36499218 on Build GRCh38
5862473 c.1481G>A p.R494Q Chr.2: 36499327-36499327 on Build GRCh38
218723 c.1420A>G p.N474D Chr.2: 36499266-36499266 on Build GRCh38
3743790 c.1474G>T p.G492C Chr.2: 36499320-36499320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913277849 Chr.2: 36499551-36499551 on Build GRCh38
rs755612207 Chr.2: 36499190-36499190 on Build GRCh38
rs898406265 Chr.2: 36499389-36499389 on Build GRCh38
rs939678234 Chr.2: 36499368-36499368 on Build GRCh38
rs35954643 Chr.2: 36499257-36499258 on Build GRCh38
rs773320653 Chr.2: 36499294-36499295 on Build GRCh38
rs747423363 Chr.2: 36499227-36499227 on Build GRCh38
rs963069661 Chr.2: 36499439-36499439 on Build GRCh38
rs1014306321 Chr.2: 36499431-36499431 on Build GRCh38
rs751426719 Chr.2: 36499173-36499173 on Build GRCh38
rs76008222 Chr.2: 36499420-36499420 on Build GRCh38
rs559369885 Chr.2: 36499427-36499427 on Build GRCh38
rs575481242 Chr.2: 36499162-36499162 on Build GRCh38
rs941946480 Chr.2: 36499159-36499159 on Build GRCh38
rs530408681 Chr.2: 36499467-36499467 on Build GRCh38
rs769261151 Chr.2: 36499196-36499196 on Build GRCh38
rs983124300 Chr.2: 36499128-36499128 on Build GRCh38
rs775214531 Chr.2: 36499341-36499341 on Build GRCh38
rs202109107 Chr.2: 36499254-36499254 on Build GRCh38
rs770825553 Chr.2: 36499318-36499318 on Build GRCh38
rs1038285944 Chr.2: 36499377-36499377 on Build GRCh38
rs374553015 Chr.2: 36499311-36499311 on Build GRCh38
rs950359584 Chr.2: 36499113-36499113 on Build GRCh38
rs950450335 Chr.2: 36499411-36499411 on Build GRCh38
rs748342817 Chr.2: 36499241-36499241 on Build GRCh38
rs921405667 Chr.2: 36499518-36499518 on Build GRCh38
rs769857707 Chr.2: 36499244-36499244 on Build GRCh38
rs767333044 Chr.2: 36499259-36499259 on Build GRCh38
rs774110531 Chr.2: 36499255-36499255 on Build GRCh38
rs776889204 Chr.2: 36499195-36499196 on Build GRCh38
rs921033151 Chr.2: 36499316-36499316 on Build GRCh38
rs371409342 Chr.2: 36499392-36499392 on Build GRCh38
rs766099530 Chr.2: 36499172-36499172 on Build GRCh38
rs1033286058 Chr.2: 36499449-36499449 on Build GRCh38
rs548647212 Chr.2: 36499483-36499483 on Build GRCh38
rs933724606 Chr.2: 36499356-36499356 on Build GRCh38
rs561133654 Chr.2: 36499178-36499178 on Build GRCh38
rs371361879 Chr.2: 36499280-36499280 on Build GRCh38
rs974671226 Chr.2: 36499440-36499440 on Build GRCh38
rs375356337 Chr.2: 36499310-36499310 on Build GRCh38
rs946015251 Chr.2: 36499251-36499251 on Build GRCh38
rs71437556 Chr.2: 36499539-36499539 on Build GRCh38
rs760357974 Chr.2: 36499351-36499351 on Build GRCh38
rs759367951 Chr.2: 36499256-36499256 on Build GRCh38
rs753346080 Chr.2: 36499266-36499266 on Build GRCh38
rs563303043 Chr.2: 36499127-36499127 on Build GRCh38
rs767029091 Chr.2: 36499281-36499281 on Build GRCh38
rs375715855 Chr.2: 36499209-36499209 on Build GRCh38
rs975589884 Chr.2: 36499496-36499496 on Build GRCh38
rs576945195 Chr.2: 36499114-36499114 on Build GRCh38
rs1002018883 Chr.2: 36499395-36499395 on Build GRCh38
rs529766756 Chr.2: 36499549-36499549 on Build GRCh38
rs749247802 Chr.2: 36499250-36499250 on Build GRCh38
rs773173229 Chr.2: 36499247-36499247 on Build GRCh38
rs538622396 Chr.2: 36499132-36499134 on Build GRCh38
rs755343317 Chr.2: 36499292-36499292 on Build GRCh38
rs200267829 Chr.2: 36499174-36499174 on Build GRCh38
rs184564899 Chr.2: 36499455-36499455 on Build GRCh38
rs777160324 Chr.2: 36499191-36499191 on Build GRCh38
rs770500528 Chr.2: 36499224-36499224 on Build GRCh38
rs967438346 Chr.2: 36499444-36499444 on Build GRCh38
rs776175356 Chr.2: 36499371-36499371 on Build GRCh38
rs752139638 Chr.2: 36499286-36499286 on Build GRCh38
rs139161726 Chr.2: 36499320-36499320 on Build GRCh38
rs886736965 Chr.2: 36499363-36499363 on Build GRCh38
rs1034427199 Chr.2: 36499166-36499166 on Build GRCh38
rs773882030 Chr.2: 36499326-36499326 on Build GRCh38
rs748207568 Chr.2: 36499206-36499209 on Build GRCh38
rs745332459 Chr.2: 36499327-36499327 on Build GRCh38
rs146523843 Chr.2: 36499181-36499181 on Build GRCh38
rs998842475 Chr.2: 36499445-36499445 on Build GRCh38
rs750874576 Chr.2: 36499203-36499203 on Build GRCh38
rs747574530 Chr.2: 36499179-36499180 on Build GRCh38
rs760248337 Chr.2: 36499263-36499263 on Build GRCh38
rs567155581 Chr.2: 36499534-36499534 on Build GRCh38
rs755216648 Chr.2: 36499237-36499237 on Build GRCh38
rs898375230 Chr.2: 36499160-36499160 on Build GRCh38
rs769862063 Chr.2: 36499270-36499270 on Build GRCh38
rs763233561 Chr.2: 36499365-36499366 on Build GRCh38
rs1016724792 Chr.2: 36499145-36499145 on Build GRCh38
rs781425552 Chr.2: 36499296-36499296 on Build GRCh38
rs771832173 Chr.2: 36499331-36499331 on Build GRCh38
rs761366438 Chr.2: 36499386-36499386 on Build GRCh38
rs780557382 Chr.2: 36499171-36499172 on Build GRCh38
rs748337826 Chr.2: 36499307-36499307 on Build GRCh38
rs377625523 Chr.2: 36499262-36499262 on Build GRCh38
rs763655624 Chr.2: 36499264-36499264 on Build GRCh38
rs1004097024 Chr.2: 36499135-36499135 on Build GRCh38
rs1024731168 Chr.2: 36499432-36499432 on Build GRCh38
rs565003825 Chr.2: 36499273-36499273 on Build GRCh38
rs781432598 Chr.2: 36499238-36499238 on Build GRCh38
rs59890798 Chr.2: 36499357-36499357 on Build GRCh38
rs987721000 Chr.2: 36499495-36499495 on Build GRCh38
rs756361822 Chr.2: 36499309-36499309 on Build GRCh38
rs1033452294 Chr.2: 36499401-36499401 on Build GRCh38

View Full Product Details