Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61180980-61181506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321300.1 NM_001321301.1 NM_001321302.1 NM_001321303.1 NM_001321304.1 NM_001321305.1 NM_152392.4 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479829 c.354C>T p.D118D Chr.2: 61181399-61181399 on Build GRCh38
5521319 c.275-8C>A p.? Chr.2: 61181312-61181312 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770917990 Chr.2: 61181324-61181324 on Build GRCh38
rs567980527 Chr.2: 61181062-61181062 on Build GRCh38
rs1025621873 Chr.2: 61181374-61181374 on Build GRCh38
rs775646582 Chr.2: 61181073-61181073 on Build GRCh38
rs764422599 Chr.2: 61181424-61181424 on Build GRCh38
rs976652053 Chr.2: 61181031-61181032 on Build GRCh38
rs935552979 Chr.2: 61181281-61181281 on Build GRCh38
rs917018156 Chr.2: 61181187-61181187 on Build GRCh38
rs972676326 Chr.2: 61181145-61181145 on Build GRCh38
rs747214894 Chr.2: 61181417-61181417 on Build GRCh38
rs147817052 Chr.2: 61181475-61181475 on Build GRCh38
rs984836403 Chr.2: 61181041-61181041 on Build GRCh38
rs374237302 Chr.2: 61181439-61181439 on Build GRCh38
rs550033758 Chr.2: 61181006-61181006 on Build GRCh38
rs554496539 Chr.2: 61181128-61181128 on Build GRCh38
rs758090041 Chr.2: 61181358-61181358 on Build GRCh38
rs753839333 Chr.2: 61181279-61181279 on Build GRCh38
rs534289362 Chr.2: 61181293-61181293 on Build GRCh38
rs887114066 Chr.2: 61181393-61181393 on Build GRCh38
rs772659705 Chr.2: 61181021-61181021 on Build GRCh38
rs1018815489 Chr.2: 61181022-61181022 on Build GRCh38
rs901166914 Chr.2: 61181056-61181056 on Build GRCh38
rs558829521 Chr.2: 61181304-61181304 on Build GRCh38
rs953551133 Chr.2: 61181330-61181330 on Build GRCh38
rs903557127 Chr.2: 61181247-61181247 on Build GRCh38
rs1056693960 Chr.2: 61181178-61181178 on Build GRCh38
rs181890681 Chr.2: 61181359-61181359 on Build GRCh38
rs953592893 Chr.2: 61181032-61181032 on Build GRCh38
rs1043427400 Chr.2: 61181245-61181245 on Build GRCh38
rs951253812 Chr.2: 61181113-61181113 on Build GRCh38
rs572910051 Chr.2: 61181257-61181257 on Build GRCh38
rs918490504 Chr.2: 61181151-61181151 on Build GRCh38
rs746853238 Chr.2: 61181031-61181031 on Build GRCh38
rs893973752 Chr.2: 61181292-61181292 on Build GRCh38
rs754930959 Chr.2: 61181283-61181283 on Build GRCh38
rs754776464 Chr.2: 61181061-61181061 on Build GRCh38
rs12328566 Chr.2: 61181449-61181449 on Build GRCh38
rs940644947 Chr.2: 61181096-61181096 on Build GRCh38
rs984050460 Chr.2: 61181127-61181127 on Build GRCh38
rs1016742158 Chr.2: 61181130-61181130 on Build GRCh38
rs564987913 Chr.2: 61181089-61181089 on Build GRCh38
rs747981867 Chr.2: 61181310-61181310 on Build GRCh38
rs902083505 Chr.2: 61181289-61181289 on Build GRCh38
rs748073674 Chr.2: 61181196-61181196 on Build GRCh38
rs966258988 Chr.2: 61181030-61181030 on Build GRCh38
rs567696673 Chr.2: 61181312-61181320 on Build GRCh38
rs761091803 Chr.2: 61181325-61181325 on Build GRCh38
rs972057319 Chr.2: 61181431-61181431 on Build GRCh38
rs1029110758 Chr.2: 61181032-61181032 on Build GRCh38
rs778167480 Chr.2: 61181371-61181371 on Build GRCh38
rs958788048 Chr.2: 61181133-61181133 on Build GRCh38
rs977077778 Chr.2: 61181208-61181208 on Build GRCh38
rs924295535 Chr.2: 61181265-61181265 on Build GRCh38
rs998623702 Chr.2: 61181072-61181072 on Build GRCh38
rs369627210 Chr.2: 61181083-61181083 on Build GRCh38
rs548176904 Chr.2: 61181301-61181301 on Build GRCh38
rs918545497 Chr.2: 61181437-61181437 on Build GRCh38
rs1016899617 Chr.2: 61181442-61181442 on Build GRCh38
rs535381816 Chr.2: 61181068-61181068 on Build GRCh38
rs999745556 Chr.2: 61181008-61181008 on Build GRCh38
rs984888733 Chr.2: 61181340-61181340 on Build GRCh38
rs991249282 Chr.2: 61181165-61181165 on Build GRCh38
rs949732336 Chr.2: 61181199-61181199 on Build GRCh38

View Full Product Details