Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61184629-61185139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321300.1 NM_001321301.1 NM_001321302.1 NM_001321303.1 NM_001321304.1 NM_001321305.1 NM_152392.4 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5458990 c.564+3A>G p.? Chr.2: 61184803-61184803 on Build GRCh38
1022052 c.481A>G p.T161A Chr.2: 61184717-61184717 on Build GRCh38
4095033 c.15G>A p.T5T Chr.2: 61184740-61184740 on Build GRCh38
3054270 c.485C>T p.T162M Chr.2: 61184721-61184721 on Build GRCh38
5354152 c.25G>A p.A9T Chr.2: 61184750-61184750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752723116 Chr.2: 61184748-61184748 on Build GRCh38
rs759082886 Chr.2: 61184817-61184817 on Build GRCh38
rs375582313 Chr.2: 61184676-61184676 on Build GRCh38
rs376685659 Chr.2: 61184939-61184939 on Build GRCh38
rs557138733 Chr.2: 61185008-61185008 on Build GRCh38
rs757073837 Chr.2: 61184756-61184756 on Build GRCh38
rs776108536 Chr.2: 61184739-61184739 on Build GRCh38
rs549252701 Chr.2: 61184902-61184902 on Build GRCh38
rs375483746 Chr.2: 61184846-61184846 on Build GRCh38
rs762982954 Chr.2: 61184751-61184751 on Build GRCh38
rs764154281 Chr.2: 61184829-61184829 on Build GRCh38
rs148994739 Chr.2: 61184800-61184800 on Build GRCh38
rs745988566 Chr.2: 61184718-61184718 on Build GRCh38
rs975067658 Chr.2: 61185052-61185052 on Build GRCh38
rs778219054 Chr.2: 61184862-61184862 on Build GRCh38
rs373856736 Chr.2: 61184722-61184722 on Build GRCh38
rs781585026 Chr.2: 61184784-61184784 on Build GRCh38
rs115642769 Chr.2: 61184863-61184863 on Build GRCh38
rs1020540119 Chr.2: 61184709-61184709 on Build GRCh38
rs747306729 Chr.2: 61184683-61184683 on Build GRCh38
rs368015953 Chr.2: 61184738-61184738 on Build GRCh38
rs527744468 Chr.2: 61184788-61184788 on Build GRCh38
rs571166446 Chr.2: 61184932-61184932 on Build GRCh38
rs896213352 Chr.2: 61184667-61184667 on Build GRCh38
rs372305344 Chr.2: 61184818-61184818 on Build GRCh38
rs762482356 Chr.2: 61184827-61184827 on Build GRCh38
rs769710888 Chr.2: 61184720-61184720 on Build GRCh38
rs944738217 Chr.2: 61184855-61184855 on Build GRCh38
rs1049436369 Chr.2: 61184896-61184896 on Build GRCh38
rs184310344 Chr.2: 61185020-61185020 on Build GRCh38
rs753678340 Chr.2: 61184832-61184832 on Build GRCh38
rs769165399 Chr.2: 61184807-61184807 on Build GRCh38
rs368581345 Chr.2: 61184813-61184813 on Build GRCh38
rs930620448 Chr.2: 61185083-61185083 on Build GRCh38
rs764633305 Chr.2: 61184741-61184741 on Build GRCh38
rs767183327 Chr.2: 61184850-61184850 on Build GRCh38
rs986274031 Chr.2: 61184824-61184824 on Build GRCh38
rs781495132 Chr.2: 61184696-61184696 on Build GRCh38
rs116428488 Chr.2: 61184740-61184740 on Build GRCh38
rs919377048 Chr.2: 61185081-61185081 on Build GRCh38
rs1019767009 Chr.2: 61185037-61185037 on Build GRCh38
rs903504346 Chr.2: 61184940-61184940 on Build GRCh38
rs771612138 Chr.2: 61184924-61184924 on Build GRCh38
rs930693064 Chr.2: 61184871-61184871 on Build GRCh38
rs764149818 Chr.2: 61184793-61184793 on Build GRCh38
rs7606748 Chr.2: 61184910-61184910 on Build GRCh38
rs554819327 Chr.2: 61185038-61185038 on Build GRCh38
rs573061076 Chr.2: 61185095-61185095 on Build GRCh38
rs569049262 Chr.2: 61185009-61185009 on Build GRCh38
rs541045635 Chr.2: 61184681-61184682 on Build GRCh38
rs148380855 Chr.2: 61184695-61184695 on Build GRCh38
rs878866571 Chr.2: 61184659-61184659 on Build GRCh38
rs751489602 Chr.2: 61184841-61184841 on Build GRCh38
rs894403633 Chr.2: 61184996-61184996 on Build GRCh38
rs984750472 Chr.2: 61185111-61185111 on Build GRCh38
rs756048689 Chr.2: 61184895-61184895 on Build GRCh38
rs778262724 Chr.2: 61184846-61184847 on Build GRCh38
rs559507245 Chr.2: 61184721-61184721 on Build GRCh38
rs1011406942 Chr.2: 61185003-61185003 on Build GRCh38
rs749864378 Chr.2: 61184803-61184803 on Build GRCh38
rs774234297 Chr.2: 61184823-61184823 on Build GRCh38
rs1055670016 Chr.2: 61184974-61184974 on Build GRCh38
rs374476804 Chr.2: 61184812-61184812 on Build GRCh38
rs7606756 Chr.2: 61184925-61184925 on Build GRCh38
rs756830463 Chr.2: 61184840-61184840 on Build GRCh38
rs745594486 Chr.2: 61184849-61184854 on Build GRCh38
rs926626969 Chr.2: 61185099-61185099 on Build GRCh38
rs769075363 Chr.2: 61184729-61184729 on Build GRCh38
rs767484568 Chr.2: 61184671-61184672 on Build GRCh38
rs920898089 Chr.2: 61185085-61185085 on Build GRCh38
rs371391326 Chr.2: 61184753-61184753 on Build GRCh38
rs998646049 Chr.2: 61185036-61185036 on Build GRCh38
rs1000561399 Chr.2: 61184945-61184945 on Build GRCh38
rs988450399 Chr.2: 61184911-61184920 on Build GRCh38
rs756255243 Chr.2: 61184798-61184798 on Build GRCh38
rs972132709 Chr.2: 61185048-61185048 on Build GRCh38
rs150899176 Chr.2: 61184909-61184909 on Build GRCh38
rs774684282 Chr.2: 61184734-61184734 on Build GRCh38
rs771433334 Chr.2: 61184982-61184982 on Build GRCh38
rs750585998 Chr.2: 61184794-61184794 on Build GRCh38
rs774205352 Chr.2: 61185001-61185002 on Build GRCh38
rs563529587 Chr.2: 61184749-61184749 on Build GRCh38
rs755910325 Chr.2: 61184707-61184708 on Build GRCh38
rs118129500 Chr.2: 61184959-61184959 on Build GRCh38
rs905130601 Chr.2: 61184900-61184900 on Build GRCh38
rs377625930 Chr.2: 61184677-61184677 on Build GRCh38
rs751399704 Chr.2: 61184755-61184755 on Build GRCh38

View Full Product Details