Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61188001-61188504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
UniGene
Hs.655602
Gene Chromosome Location
Chr.2: 61177418-61191210 on Build GRCh38
Transcripts
NM_001321305.1 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

Gene Symbol
USP34
Entrez Gene ID
9736
Gene Name
ubiquitin specific peptidase 34
Gene Aliases
-
UniGene
Hs.644708
Gene Chromosome Location
Chr.2: 61187455-61470714 on Build GRCh38
Transcripts
NM_001321305.1 XM_011532541.2 XM_011532543.2 XM_011532545.2 XM_017003377.1 XM_017003378.1 NM_014709.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922962 c.10435G>C p.D3479H Chr.2: 61188308-61188308 on Build GRCh38
1582539 c.10480C>T p.Q3494* Chr.2: 61188263-61188263 on Build GRCh38
6158735 c.10327A>G p.R3443G Chr.2: 61188416-61188416 on Build GRCh38
5726901 c.10314G>A p.Q3438Q Chr.2: 61188429-61188429 on Build GRCh38
5354623 c.10300C>T p.H3434Y Chr.2: 61188443-61188443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537190557 Chr.2: 61188226-61188226 on Build GRCh38
rs758747458 Chr.2: 61188242-61188242 on Build GRCh38
rs373276020 Chr.2: 61188408-61188408 on Build GRCh38
rs746161609 Chr.2: 61188054-61188055 on Build GRCh38
rs183972127 Chr.2: 61188363-61188363 on Build GRCh38
rs751900335 Chr.2: 61188100-61188100 on Build GRCh38
rs947078808 Chr.2: 61188339-61188339 on Build GRCh38
rs763955723 Chr.2: 61188099-61188099 on Build GRCh38
rs750629756 Chr.2: 61188197-61188197 on Build GRCh38
rs762318901 Chr.2: 61188428-61188428 on Build GRCh38
rs772430552 Chr.2: 61188061-61188062 on Build GRCh38
rs753614529 Chr.2: 61188434-61188434 on Build GRCh38
rs769393778 Chr.2: 61188159-61188159 on Build GRCh38
rs745731950 Chr.2: 61188130-61188130 on Build GRCh38
rs764453553 Chr.2: 61188380-61188380 on Build GRCh38
rs772901471 Chr.2: 61188070-61188070 on Build GRCh38
rs372729949 Chr.2: 61188143-61188143 on Build GRCh38
rs765693555 Chr.2: 61188295-61188295 on Build GRCh38
rs776907742 Chr.2: 61188240-61188240 on Build GRCh38
rs900169166 Chr.2: 61188134-61188134 on Build GRCh38
rs377379006 Chr.2: 61188137-61188137 on Build GRCh38
rs376246838 Chr.2: 61188379-61188379 on Build GRCh38
rs771829839 Chr.2: 61188349-61188349 on Build GRCh38
rs746923658 Chr.2: 61188477-61188477 on Build GRCh38
rs749687408 Chr.2: 61188063-61188063 on Build GRCh38
rs760230196 Chr.2: 61188072-61188072 on Build GRCh38
rs776793397 Chr.2: 61188360-61188360 on Build GRCh38
rs775656101 Chr.2: 61188411-61188411 on Build GRCh38
rs751910261 Chr.2: 61188331-61188331 on Build GRCh38
rs1010295293 Chr.2: 61188202-61188202 on Build GRCh38
rs797020579 Chr.2: 61188425-61188425 on Build GRCh38
rs552559192 Chr.2: 61188326-61188326 on Build GRCh38
rs760404288 Chr.2: 61188356-61188356 on Build GRCh38
rs983151235 Chr.2: 61188328-61188328 on Build GRCh38
rs933000623 Chr.2: 61188383-61188383 on Build GRCh38
rs776008043 Chr.2: 61188078-61188078 on Build GRCh38
rs753213727 Chr.2: 61188319-61188319 on Build GRCh38
rs770540687 Chr.2: 61188074-61188074 on Build GRCh38
rs201379721 Chr.2: 61188193-61188193 on Build GRCh38
rs764697278 Chr.2: 61188422-61188422 on Build GRCh38
rs767981755 Chr.2: 61188429-61188429 on Build GRCh38
rs187252476 Chr.2: 61188449-61188449 on Build GRCh38
rs774971871 Chr.2: 61188423-61188423 on Build GRCh38
rs775723756 Chr.2: 61188072-61188073 on Build GRCh38
rs1044491199 Chr.2: 61188469-61188469 on Build GRCh38
rs755848631 Chr.2: 61188332-61188332 on Build GRCh38
rs770219215 Chr.2: 61188056-61188056 on Build GRCh38
rs773365619 Chr.2: 61188264-61188264 on Build GRCh38
rs779896422 Chr.2: 61188388-61188388 on Build GRCh38
rs898706944 Chr.2: 61188421-61188421 on Build GRCh38
rs373091962 Chr.2: 61188132-61188132 on Build GRCh38
rs762982938 Chr.2: 61188165-61188165 on Build GRCh38
rs746837716 Chr.2: 61188405-61188405 on Build GRCh38
rs375982399 Chr.2: 61188144-61188144 on Build GRCh38
rs369821987 Chr.2: 61188372-61188372 on Build GRCh38
rs757660239 Chr.2: 61188387-61188387 on Build GRCh38
rs759027175 Chr.2: 61188090-61188090 on Build GRCh38
rs753927540 Chr.2: 61188276-61188276 on Build GRCh38
rs760884483 Chr.2: 61188269-61188269 on Build GRCh38
rs753518855 Chr.2: 61188389-61188389 on Build GRCh38
rs748841847 Chr.2: 61188334-61188334 on Build GRCh38
rs377232800 Chr.2: 61188065-61188065 on Build GRCh38
rs767323813 Chr.2: 61188263-61188263 on Build GRCh38
rs966628459 Chr.2: 61188027-61188030 on Build GRCh38
rs368138866 Chr.2: 61188333-61188333 on Build GRCh38
rs72884976 Chr.2: 61188402-61188402 on Build GRCh38
rs375896839 Chr.2: 61188403-61188403 on Build GRCh38
rs200812795 Chr.2: 61188262-61188262 on Build GRCh38
rs143842551 Chr.2: 61188284-61188284 on Build GRCh38
rs761623093 Chr.2: 61188192-61188192 on Build GRCh38
rs762856863 Chr.2: 61188451-61188452 on Build GRCh38
rs756235625 Chr.2: 61188205-61188205 on Build GRCh38
rs921589212 Chr.2: 61188365-61188365 on Build GRCh38
rs749064258 Chr.2: 61188043-61188043 on Build GRCh38
rs769483650 Chr.2: 61188254-61188254 on Build GRCh38
rs1047405124 Chr.2: 61188082-61188082 on Build GRCh38
rs779243938 Chr.2: 61188133-61188133 on Build GRCh38
rs747919040 Chr.2: 61188089-61188089 on Build GRCh38
rs1015777060 Chr.2: 61188180-61188180 on Build GRCh38
rs375952464 Chr.2: 61188419-61188419 on Build GRCh38
rs373240678 Chr.2: 61188058-61188058 on Build GRCh38
rs749773425 Chr.2: 61188125-61188125 on Build GRCh38
rs762877050 Chr.2: 61188098-61188098 on Build GRCh38
rs777520011 Chr.2: 61188404-61188404 on Build GRCh38
rs763250723 Chr.2: 61188361-61188361 on Build GRCh38
rs745508469 Chr.2: 61188246-61188246 on Build GRCh38
rs756179609 Chr.2: 61188115-61188115 on Build GRCh38
rs745896985 Chr.2: 61188054-61188054 on Build GRCh38
rs750494102 Chr.2: 61188113-61188113 on Build GRCh38
rs911750526 Chr.2: 61188036-61188037 on Build GRCh38
rs765934041 Chr.2: 61188455-61188456 on Build GRCh38
rs184080617 Chr.2: 61188176-61188176 on Build GRCh38
rs766433361 Chr.2: 61188212-61188212 on Build GRCh38
rs764060181 Chr.2: 61188167-61188167 on Build GRCh38
rs747071492 Chr.2: 61188040-61188040 on Build GRCh38
rs891280632 Chr.2: 61188149-61188149 on Build GRCh38
rs768739686 Chr.2: 61188258-61188258 on Build GRCh38
rs769595243 Chr.2: 61188132-61188133 on Build GRCh38
rs201356428 Chr.2: 61188095-61188095 on Build GRCh38
rs764737207 Chr.2: 61188443-61188443 on Build GRCh38
rs757429225 Chr.2: 61188101-61188101 on Build GRCh38
rs372028992 Chr.2: 61188272-61188272 on Build GRCh38
rs370358517 Chr.2: 61188454-61188454 on Build GRCh38
rs201798314 Chr.2: 61188313-61188313 on Build GRCh38
rs768251669 Chr.2: 61188336-61188336 on Build GRCh38
rs774308916 Chr.2: 61188260-61188260 on Build GRCh38
rs14170 Chr.2: 61188357-61188357 on Build GRCh38
rs370142738 Chr.2: 61188161-61188161 on Build GRCh38
rs757097700 Chr.2: 61188478-61188478 on Build GRCh38
rs928482124 Chr.2: 61188033-61188033 on Build GRCh38
rs781420169 Chr.2: 61188107-61188107 on Build GRCh38
rs765335041 Chr.2: 61188375-61188375 on Build GRCh38
rs754664502 Chr.2: 61188439-61188439 on Build GRCh38
rs751996549 Chr.2: 61188381-61188381 on Build GRCh38
rs748481815 Chr.2: 61188237-61188237 on Build GRCh38
rs371602191 Chr.2: 61188337-61188337 on Build GRCh38
rs371587110 Chr.2: 61188213-61188213 on Build GRCh38
rs188614804 Chr.2: 61188243-61188243 on Build GRCh38
rs561532901 Chr.2: 61188162-61188162 on Build GRCh38
rs749974086 Chr.2: 61188432-61188434 on Build GRCh38
rs369630143 Chr.2: 61188407-61188407 on Build GRCh38
rs200566849 Chr.2: 61188462-61188462 on Build GRCh38
rs907571442 Chr.2: 61188344-61188344 on Build GRCh38
rs148154008 Chr.2: 61188139-61188139 on Build GRCh38
rs773063379 Chr.2: 61188353-61188353 on Build GRCh38
rs779331964 Chr.2: 61188230-61188230 on Build GRCh38
rs192182020 Chr.2: 61188257-61188257 on Build GRCh38
rs780581064 Chr.2: 61188123-61188123 on Build GRCh38
rs748104342 Chr.2: 61188342-61188342 on Build GRCh38
rs752273114 Chr.2: 61188445-61188445 on Build GRCh38
rs754504021 Chr.2: 61188397-61188397 on Build GRCh38

View Full Product Details