Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61826395-61826931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM161A
Entrez Gene ID
84140
Gene Name
family with sequence similarity 161 member A
Gene Aliases
RP28
UniGene
Hs.440466
Gene Chromosome Location
Chr.2: 61800240-61856074 on Build GRCh38
Transcripts
NM_001201543.1 NM_032180.2 NR_037710.1 XM_017005072.1 XM_017005073.1 XM_017005074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682666 c.1531delA p.I511fs*1 Chr.2: 61826580-61826580 on Build GRCh38
1022129 c.1617G>A p.E539E Chr.2: 61826494-61826494 on Build GRCh38
1022131 c.1519G>T p.E507* Chr.2: 61826592-61826592 on Build GRCh38
5623904 c.2110G>A p.E704K Chr.2: 61826496-61826496 on Build GRCh38
6564726 c.2019A>T p.E673D Chr.2: 61826587-61826587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962442172 Chr.2: 61826661-61826661 on Build GRCh38
rs755891868 Chr.2: 61826412-61826412 on Build GRCh38
rs941194514 Chr.2: 61826763-61826763 on Build GRCh38
rs926098895 Chr.2: 61826636-61826636 on Build GRCh38
rs368402244 Chr.2: 61826590-61826590 on Build GRCh38
rs201436856 Chr.2: 61826742-61826742 on Build GRCh38
rs774262388 Chr.2: 61826628-61826628 on Build GRCh38
rs755427067 Chr.2: 61826556-61826556 on Build GRCh38
rs72380346 Chr.2: 61826412-61826414 on Build GRCh38
rs200040100 Chr.2: 61826743-61826743 on Build GRCh38
rs563015622 Chr.2: 61826768-61826768 on Build GRCh38
rs762666633 Chr.2: 61826616-61826616 on Build GRCh38
rs375275247 Chr.2: 61826644-61826644 on Build GRCh38
rs966473607 Chr.2: 61826870-61826870 on Build GRCh38
rs576190170 Chr.2: 61826651-61826651 on Build GRCh38
rs927645332 Chr.2: 61826710-61826710 on Build GRCh38
rs897217301 Chr.2: 61826776-61826776 on Build GRCh38
rs998875938 Chr.2: 61826586-61826586 on Build GRCh38
rs998251316 Chr.2: 61826785-61826785 on Build GRCh38
rs750019920 Chr.2: 61826544-61826544 on Build GRCh38
rs879158086 Chr.2: 61826599-61826599 on Build GRCh38
rs866016324 Chr.2: 61826656-61826656 on Build GRCh38
rs749622564 Chr.2: 61826553-61826553 on Build GRCh38
rs751454162 Chr.2: 61826517-61826517 on Build GRCh38
rs771089237 Chr.2: 61826600-61826600 on Build GRCh38
rs752502114 Chr.2: 61826615-61826615 on Build GRCh38
rs888295419 Chr.2: 61826540-61826540 on Build GRCh38
rs773397179 Chr.2: 61826461-61826461 on Build GRCh38
rs989602235 Chr.2: 61826645-61826645 on Build GRCh38
rs763811079 Chr.2: 61826622-61826622 on Build GRCh38
rs1007056265 Chr.2: 61826816-61826816 on Build GRCh38
rs774003829 Chr.2: 61826460-61826460 on Build GRCh38
rs563390775 Chr.2: 61826680-61826680 on Build GRCh38
rs925070366 Chr.2: 61826751-61826751 on Build GRCh38
rs397770644 Chr.2: 61826745-61826746 on Build GRCh38
rs937883870 Chr.2: 61826716-61826716 on Build GRCh38
rs774744427 Chr.2: 61826438-61826438 on Build GRCh38
rs376530799 Chr.2: 61826498-61826498 on Build GRCh38
rs756054713 Chr.2: 61826620-61826620 on Build GRCh38
rs780565463 Chr.2: 61826416-61826416 on Build GRCh38
rs981755820 Chr.2: 61826703-61826703 on Build GRCh38
rs77562614 Chr.2: 61826484-61826484 on Build GRCh38
rs1017027409 Chr.2: 61826647-61826647 on Build GRCh38
rs750533047 Chr.2: 61826814-61826814 on Build GRCh38
rs764049454 Chr.2: 61826502-61826502 on Build GRCh38
rs781110661 Chr.2: 61826565-61826566 on Build GRCh38
rs998192755 Chr.2: 61826574-61826574 on Build GRCh38
rs957893140 Chr.2: 61826834-61826834 on Build GRCh38
rs779412827 Chr.2: 61826578-61826578 on Build GRCh38
rs1055067867 Chr.2: 61826737-61826737 on Build GRCh38
rs769132179 Chr.2: 61826653-61826653 on Build GRCh38
rs775239997 Chr.2: 61826496-61826496 on Build GRCh38
rs772056519 Chr.2: 61826441-61826441 on Build GRCh38
rs779902438 Chr.2: 61826551-61826551 on Build GRCh38
rs913645310 Chr.2: 61826667-61826667 on Build GRCh38
rs954282082 Chr.2: 61826608-61826608 on Build GRCh38
rs138464813 Chr.2: 61826542-61826542 on Build GRCh38
rs142802290 Chr.2: 61826847-61826847 on Build GRCh38
rs766627808 Chr.2: 61826490-61826490 on Build GRCh38
rs1033230698 Chr.2: 61826821-61826821 on Build GRCh38
rs370157006 Chr.2: 61826459-61826459 on Build GRCh38
rs11392751 Chr.2: 61826743-61826744 on Build GRCh38
rs945159813 Chr.2: 61826681-61826681 on Build GRCh38
rs755888023 Chr.2: 61826651-61826651 on Build GRCh38
rs375067034 Chr.2: 61826549-61826549 on Build GRCh38
rs766206495 Chr.2: 61826543-61826543 on Build GRCh38
rs1029588921 Chr.2: 61826794-61826794 on Build GRCh38
rs1025766874 Chr.2: 61826684-61826684 on Build GRCh38
rs748587957 Chr.2: 61826588-61826588 on Build GRCh38
rs61743113 Chr.2: 61826429-61826429 on Build GRCh38
rs548583143 Chr.2: 61826818-61826818 on Build GRCh38
rs191862300 Chr.2: 61826541-61826541 on Build GRCh38
rs370603613 Chr.2: 61826595-61826595 on Build GRCh38
rs558080743 Chr.2: 61826480-61826482 on Build GRCh38
rs530255784 Chr.2: 61826799-61826799 on Build GRCh38
rs748220974 Chr.2: 61826440-61826440 on Build GRCh38
rs527984324 Chr.2: 61826867-61826867 on Build GRCh38
rs769265045 Chr.2: 61826434-61826434 on Build GRCh38
rs869269610 Chr.2: 61826744-61826745 on Build GRCh38
rs748147923 Chr.2: 61826569-61826572 on Build GRCh38

View Full Product Details