Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68819033-68819522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP25
Entrez Gene ID
9938
Gene Name
Rho GTPase activating protein 25
Gene Aliases
HEL-S-308, KAIA0053
UniGene
Hs.531807
Gene Chromosome Location
Chr.2: 68710505-68826833 on Build GRCh38
Transcripts
NM_001007231.2 NM_001166276.1 NM_001166277.1 NM_014882.2 XM_005264675.2 XM_005264676.2 XM_011533206.2 XM_011533207.2 XM_011533208.2 XM_011533209.2 XM_011533210.2 XM_017005423.1 XM_017005424.1 XM_017005425.1 XM_017005426.1 XM_017005427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3407941 c.1176T>C p.S392S Chr.2: 68819295-68819295 on Build GRCh38
1668992 c.1039T>G p.F347V Chr.2: 68819179-68819179 on Build GRCh38
1494942 c.1075G>A p.D359N Chr.2: 68819194-68819194 on Build GRCh38
3407943 c.1155T>C p.S385S Chr.2: 68819295-68819295 on Build GRCh38
7356190 c.1033A>G p.M345V Chr.2: 68819152-68819152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753579313 Chr.2: 68819374-68819374 on Build GRCh38
rs150740740 Chr.2: 68819124-68819124 on Build GRCh38
rs771627403 Chr.2: 68819172-68819172 on Build GRCh38
rs551401562 Chr.2: 68819253-68819253 on Build GRCh38
rs776009822 Chr.2: 68819333-68819333 on Build GRCh38
rs926072441 Chr.2: 68819406-68819406 on Build GRCh38
rs762343958 Chr.2: 68819113-68819113 on Build GRCh38
rs763025368 Chr.2: 68819216-68819216 on Build GRCh38
rs531604178 Chr.2: 68819136-68819136 on Build GRCh38
rs747654803 Chr.2: 68819166-68819166 on Build GRCh38
rs770685966 Chr.2: 68819306-68819306 on Build GRCh38
rs762041392 Chr.2: 68819363-68819363 on Build GRCh38
rs772939730 Chr.2: 68819175-68819175 on Build GRCh38
rs561976079 Chr.2: 68819447-68819447 on Build GRCh38
rs936084320 Chr.2: 68819421-68819421 on Build GRCh38
rs777158951 Chr.2: 68819314-68819314 on Build GRCh38
rs202221071 Chr.2: 68819203-68819203 on Build GRCh38
rs2271833 Chr.2: 68819424-68819424 on Build GRCh38
rs760085818 Chr.2: 68819072-68819072 on Build GRCh38
rs777934913 Chr.2: 68819101-68819101 on Build GRCh38
rs780834306 Chr.2: 68819440-68819440 on Build GRCh38
rs370176484 Chr.2: 68819368-68819368 on Build GRCh38
rs755847429 Chr.2: 68819108-68819108 on Build GRCh38
rs554864462 Chr.2: 68819054-68819054 on Build GRCh38
rs765273339 Chr.2: 68819076-68819076 on Build GRCh38
rs143413666 Chr.2: 68819153-68819153 on Build GRCh38
rs758380401 Chr.2: 68819404-68819404 on Build GRCh38
rs899799149 Chr.2: 68819154-68819154 on Build GRCh38
rs763359758 Chr.2: 68819335-68819335 on Build GRCh38
rs367906670 Chr.2: 68819288-68819288 on Build GRCh38
rs754744039 Chr.2: 68819376-68819376 on Build GRCh38
rs777105770 Chr.2: 68819191-68819191 on Build GRCh38
rs764317075 Chr.2: 68819227-68819227 on Build GRCh38
rs757436657 Chr.2: 68819238-68819238 on Build GRCh38
rs78992846 Chr.2: 68819152-68819152 on Build GRCh38
rs1004388629 Chr.2: 68819209-68819209 on Build GRCh38
rs777475622 Chr.2: 68819414-68819414 on Build GRCh38
rs754902466 Chr.2: 68819251-68819251 on Build GRCh38
rs967593730 Chr.2: 68819340-68819340 on Build GRCh38
rs1033344671 Chr.2: 68819276-68819276 on Build GRCh38
rs576454977 Chr.2: 68819230-68819230 on Build GRCh38
rs926018697 Chr.2: 68819073-68819073 on Build GRCh38
rs751066163 Chr.2: 68819422-68819422 on Build GRCh38
rs149839883 Chr.2: 68819200-68819200 on Build GRCh38
rs904038387 Chr.2: 68819053-68819053 on Build GRCh38
rs372036911 Chr.2: 68819470-68819470 on Build GRCh38
rs746207665 Chr.2: 68819318-68819318 on Build GRCh38
rs764196308 Chr.2: 68819336-68819336 on Build GRCh38
rs766734098 Chr.2: 68819316-68819316 on Build GRCh38
rs752800516 Chr.2: 68819087-68819087 on Build GRCh38
rs764925329 Chr.2: 68819395-68819395 on Build GRCh38
rs775468442 Chr.2: 68819206-68819206 on Build GRCh38
rs757259867 Chr.2: 68819480-68819480 on Build GRCh38
rs778843724 Chr.2: 68819254-68819254 on Build GRCh38
rs746701704 Chr.2: 68819303-68819303 on Build GRCh38
rs746235124 Chr.2: 68819478-68819478 on Build GRCh38
rs375730981 Chr.2: 68819297-68819297 on Build GRCh38
rs371873808 Chr.2: 68819245-68819245 on Build GRCh38
rs753762761 Chr.2: 68819082-68819083 on Build GRCh38
rs779140073 Chr.2: 68819351-68819351 on Build GRCh38
rs758574634 Chr.2: 68819098-68819098 on Build GRCh38
rs147150060 Chr.2: 68819184-68819184 on Build GRCh38
rs562552839 Chr.2: 68819243-68819243 on Build GRCh38
rs750227054 Chr.2: 68819103-68819103 on Build GRCh38
rs17604346 Chr.2: 68819274-68819274 on Build GRCh38
rs751843881 Chr.2: 68819234-68819234 on Build GRCh38
rs770211030 Chr.2: 68819332-68819332 on Build GRCh38
rs752747750 Chr.2: 68819396-68819396 on Build GRCh38

View Full Product Details