Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70832514-70833038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD207
Entrez Gene ID
50489
Gene Name
CD207 molecule
Gene Aliases
CLEC4K
UniGene
Hs.199731
Gene Chromosome Location
Chr.2: 70825248-70860787 on Build GRCh38
Transcripts
NM_015717.4 XM_011532874.1 XM_011532875.2 XM_011532876.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3724397 c.613_614insG p.N205fs*14 Chr.2: 70833003-70833004 on Build GRCh38
1022550 c.649T>C p.Y217H Chr.2: 70832968-70832968 on Build GRCh38
4095383 c.630T>C p.S210S Chr.2: 70832987-70832987 on Build GRCh38
6542696 c.674C>A p.S225Y Chr.2: 70832943-70832943 on Build GRCh38
6880514 c.697G>A p.V233M Chr.2: 70832920-70832920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542384792 Chr.2: 70832876-70832876 on Build GRCh38
rs979122257 Chr.2: 70832654-70832654 on Build GRCh38
rs559383492 Chr.2: 70832879-70832879 on Build GRCh38
rs781970590 Chr.2: 70832922-70832922 on Build GRCh38
rs782227601 Chr.2: 70832861-70832861 on Build GRCh38
rs782816989 Chr.2: 70832969-70832969 on Build GRCh38
rs10204437 Chr.2: 70832632-70832632 on Build GRCh38
rs782225816 Chr.2: 70832976-70832976 on Build GRCh38
rs782137807 Chr.2: 70832884-70832884 on Build GRCh38
rs535590748 Chr.2: 70832687-70832687 on Build GRCh38
rs781824963 Chr.2: 70832579-70832579 on Build GRCh38
rs782617460 Chr.2: 70832975-70832975 on Build GRCh38
rs781901575 Chr.2: 70832890-70832890 on Build GRCh38
rs782515098 Chr.2: 70832978-70832978 on Build GRCh38
rs143527696 Chr.2: 70832678-70832678 on Build GRCh38
rs988827126 Chr.2: 70832594-70832594 on Build GRCh38
rs782391351 Chr.2: 70832926-70832926 on Build GRCh38
rs6712863 Chr.2: 70832906-70832906 on Build GRCh38
rs17006436 Chr.2: 70832980-70832980 on Build GRCh38
rs1010315927 Chr.2: 70832840-70832840 on Build GRCh38
rs930320017 Chr.2: 70832793-70832793 on Build GRCh38
rs782627815 Chr.2: 70832904-70832904 on Build GRCh38
rs781862128 Chr.2: 70832880-70832880 on Build GRCh38
rs782559974 Chr.2: 70832766-70832766 on Build GRCh38
rs781942625 Chr.2: 70832869-70832869 on Build GRCh38
rs782247148 Chr.2: 70832925-70832925 on Build GRCh38
rs781919980 Chr.2: 70832992-70832992 on Build GRCh38
rs147182001 Chr.2: 70832824-70832824 on Build GRCh38
rs782565816 Chr.2: 70832920-70832920 on Build GRCh38
rs184811667 Chr.2: 70832788-70832788 on Build GRCh38
rs573751884 Chr.2: 70832866-70832866 on Build GRCh38
rs782073024 Chr.2: 70832749-70832749 on Build GRCh38
rs1020317527 Chr.2: 70832860-70832860 on Build GRCh38
rs782592632 Chr.2: 70832854-70832854 on Build GRCh38
rs968688434 Chr.2: 70832643-70832643 on Build GRCh38
rs1049152149 Chr.2: 70832797-70832797 on Build GRCh38
rs901310113 Chr.2: 70832827-70832827 on Build GRCh38
rs376010760 Chr.2: 70832959-70832959 on Build GRCh38
rs1036072354 Chr.2: 70832759-70832759 on Build GRCh38
rs782417032 Chr.2: 70832985-70832985 on Build GRCh38
rs782427845 Chr.2: 70833014-70833014 on Build GRCh38
rs563353094 Chr.2: 70832865-70832865 on Build GRCh38
rs781850322 Chr.2: 70832960-70832960 on Build GRCh38
rs115440071 Chr.2: 70832822-70832822 on Build GRCh38
rs533675173 Chr.2: 70832972-70832972 on Build GRCh38
rs374676643 Chr.2: 70832918-70832918 on Build GRCh38
rs547167390 Chr.2: 70832728-70832728 on Build GRCh38
rs115972324 Chr.2: 70832748-70832748 on Build GRCh38
rs782792485 Chr.2: 70832885-70832885 on Build GRCh38
rs782020363 Chr.2: 70832936-70832936 on Build GRCh38
rs782291774 Chr.2: 70832823-70832823 on Build GRCh38
rs992695147 Chr.2: 70832733-70832733 on Build GRCh38
rs782697700 Chr.2: 70832939-70832939 on Build GRCh38
rs541916783 Chr.2: 70832659-70832659 on Build GRCh38
rs782163497 Chr.2: 70832937-70832937 on Build GRCh38
rs782512345 Chr.2: 70832858-70832858 on Build GRCh38
rs11891027 Chr.2: 70832800-70832800 on Build GRCh38
rs782482178 Chr.2: 70832901-70832901 on Build GRCh38
rs372167399 Chr.2: 70832945-70832945 on Build GRCh38
rs367897796 Chr.2: 70832899-70832899 on Build GRCh38
rs782750570 Chr.2: 70832562-70832562 on Build GRCh38
rs782104857 Chr.2: 70833015-70833015 on Build GRCh38
rs782643609 Chr.2: 70832979-70832979 on Build GRCh38
rs376939757 Chr.2: 70832745-70832745 on Build GRCh38
rs940427218 Chr.2: 70832757-70832757 on Build GRCh38

View Full Product Details