Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70833945-70834437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD207
Entrez Gene ID
50489
Gene Name
CD207 molecule
Gene Aliases
CLEC4K
UniGene
Hs.199731
Gene Chromosome Location
Chr.2: 70825248-70860787 on Build GRCh38
Transcripts
NM_015717.4 XM_011532874.1 XM_011532875.2 XM_011532876.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938838 c.191-4C>T p.? Chr.2: 70834024-70834024 on Build GRCh38
1409220 c.266G>A p.S89N Chr.2: 70833945-70833945 on Build GRCh38
7412864 c.233T>C p.V78A Chr.2: 70833978-70833978 on Build GRCh38
3582880 c.217G>A p.D73N Chr.2: 70833994-70833994 on Build GRCh38
5025645 c.204G>T p.M68I Chr.2: 70834007-70834007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760229899 Chr.2: 70834375-70834375 on Build GRCh38
rs17662453 Chr.2: 70833977-70833977 on Build GRCh38
rs760081403 Chr.2: 70834257-70834257 on Build GRCh38
rs79233139 Chr.2: 70834378-70834378 on Build GRCh38
rs959031901 Chr.2: 70834348-70834348 on Build GRCh38
rs782056607 Chr.2: 70834395-70834395 on Build GRCh38
rs997293730 Chr.2: 70834242-70834242 on Build GRCh38
rs561534679 Chr.2: 70834191-70834191 on Build GRCh38
rs72911708 Chr.2: 70834014-70834014 on Build GRCh38
rs199819493 Chr.2: 70833975-70833975 on Build GRCh38
rs781919872 Chr.2: 70833980-70833980 on Build GRCh38
rs377027699 Chr.2: 70834030-70834030 on Build GRCh38
rs553273697 Chr.2: 70834182-70834182 on Build GRCh38
rs909179277 Chr.2: 70834153-70834153 on Build GRCh38
rs1013625552 Chr.2: 70834353-70834353 on Build GRCh38
rs1023215418 Chr.2: 70834368-70834368 on Build GRCh38
rs781819743 Chr.2: 70834062-70834062 on Build GRCh38
rs540099793 Chr.2: 70834070-70834070 on Build GRCh38
rs971579431 Chr.2: 70834370-70834370 on Build GRCh38
rs544983849 Chr.2: 70834198-70834198 on Build GRCh38
rs1049542548 Chr.2: 70834140-70834140 on Build GRCh38
rs371003487 Chr.2: 70834035-70834035 on Build GRCh38
rs544252498 Chr.2: 70834269-70834269 on Build GRCh38
rs782041008 Chr.2: 70834042-70834042 on Build GRCh38
rs781984976 Chr.2: 70834015-70834015 on Build GRCh38
rs139574661 Chr.2: 70834388-70834388 on Build GRCh38
rs902291985 Chr.2: 70834267-70834267 on Build GRCh38
rs782717720 Chr.2: 70834061-70834061 on Build GRCh38
rs558734445 Chr.2: 70834256-70834256 on Build GRCh38
rs781818915 Chr.2: 70834311-70834311 on Build GRCh38
rs782194035 Chr.2: 70833986-70833986 on Build GRCh38
rs567505928 Chr.2: 70834156-70834156 on Build GRCh38
rs957871402 Chr.2: 70834002-70834002 on Build GRCh38
rs774857827 Chr.2: 70834190-70834190 on Build GRCh38
rs782057983 Chr.2: 70834059-70834059 on Build GRCh38
rs17719042 Chr.2: 70834268-70834268 on Build GRCh38
rs144732266 Chr.2: 70834218-70834218 on Build GRCh38
rs782651346 Chr.2: 70833974-70833974 on Build GRCh38
rs919323466 Chr.2: 70834078-70834078 on Build GRCh38
rs189619883 Chr.2: 70834146-70834146 on Build GRCh38
rs929382255 Chr.2: 70834139-70834139 on Build GRCh38
rs763818568 Chr.2: 70834410-70834410 on Build GRCh38
rs149783212 Chr.2: 70834083-70834083 on Build GRCh38
rs536561002 Chr.2: 70834180-70834180 on Build GRCh38

View Full Product Details