Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 72496366-72496870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6B
Entrez Gene ID
23233
Gene Name
exocyst complex component 6B
Gene Aliases
SEC15B, SEC15L2
UniGene
Hs.303454
Gene Chromosome Location
Chr.2: 72175984-72826042 on Build GRCh38
Transcripts
NM_001321729.1 NM_001321730.1 NM_001321731.1 NM_001321733.1 NM_001321734.1 NM_015189.2 NR_135773.1 NR_135774.1 XM_005264224.1 XM_011532711.2 XM_011532712.2 XM_017003641.1 XM_017003642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037062 c.1367C>T p.P456L Chr.2: 72496530-72496530 on Build GRCh38
5601445 c.12G>A p.K4K Chr.2: 72496496-72496496 on Build GRCh38
6435290 c.4T>A p.Y2N Chr.2: 72496504-72496504 on Build GRCh38
3583085 c.1413A>G p.Q471Q Chr.2: 72496484-72496484 on Build GRCh38
1632109 c.1338-2A>G p.? Chr.2: 72496561-72496561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007045825 Chr.2: 72496633-72496633 on Build GRCh38
rs968480205 Chr.2: 72496478-72496478 on Build GRCh38
rs763515222 Chr.2: 72496490-72496490 on Build GRCh38
rs554999483 Chr.2: 72496583-72496583 on Build GRCh38
rs753069474 Chr.2: 72496518-72496518 on Build GRCh38
rs537524016 Chr.2: 72496626-72496626 on Build GRCh38
rs1015172440 Chr.2: 72496421-72496421 on Build GRCh38
rs904265377 Chr.2: 72496671-72496671 on Build GRCh38
rs777702053 Chr.2: 72496437-72496437 on Build GRCh38
rs200752375 Chr.2: 72496606-72496606 on Build GRCh38
rs995091488 Chr.2: 72496816-72496816 on Build GRCh38
rs895513354 Chr.2: 72496710-72496710 on Build GRCh38
rs554413279 Chr.2: 72496461-72496461 on Build GRCh38
rs752955636 Chr.2: 72496605-72496606 on Build GRCh38
rs745859425 Chr.2: 72496568-72496588 on Build GRCh38
rs1043291958 Chr.2: 72496648-72496648 on Build GRCh38
rs374459655 Chr.2: 72496481-72496481 on Build GRCh38
rs754067186 Chr.2: 72496523-72496523 on Build GRCh38
rs1013943132 Chr.2: 72496719-72496719 on Build GRCh38
rs954363977 Chr.2: 72496436-72496436 on Build GRCh38
rs986227463 Chr.2: 72496576-72496576 on Build GRCh38
rs969969470 Chr.2: 72496722-72496722 on Build GRCh38
rs765719239 Chr.2: 72496454-72496454 on Build GRCh38
rs538884972 Chr.2: 72496456-72496456 on Build GRCh38
rs768633033 Chr.2: 72496424-72496424 on Build GRCh38
rs558809149 Chr.2: 72496673-72496673 on Build GRCh38
rs897152393 Chr.2: 72496611-72496611 on Build GRCh38
rs1052356889 Chr.2: 72496605-72496605 on Build GRCh38
rs746947544 Chr.2: 72496534-72496534 on Build GRCh38
rs184302368 Chr.2: 72496800-72496800 on Build GRCh38
rs758697899 Chr.2: 72496575-72496575 on Build GRCh38
rs770869353 Chr.2: 72496767-72496767 on Build GRCh38
rs375239014 Chr.2: 72496600-72496600 on Build GRCh38
rs934228253 Chr.2: 72496622-72496622 on Build GRCh38
rs746645705 Chr.2: 72496606-72496606 on Build GRCh38
rs955904244 Chr.2: 72496677-72496680 on Build GRCh38
rs536601645 Chr.2: 72496492-72496492 on Build GRCh38
rs77317487 Chr.2: 72496739-72496739 on Build GRCh38
rs145555079 Chr.2: 72496601-72496601 on Build GRCh38
rs755084255 Chr.2: 72496525-72496525 on Build GRCh38
rs756495001 Chr.2: 72496507-72496507 on Build GRCh38
rs776621349 Chr.2: 72496449-72496449 on Build GRCh38
rs368707427 Chr.2: 72496491-72496491 on Build GRCh38
rs71416746 Chr.2: 72496635-72496635 on Build GRCh38
rs189300636 Chr.2: 72496818-72496818 on Build GRCh38
rs373678197 Chr.2: 72496467-72496467 on Build GRCh38
rs376461076 Chr.2: 72496406-72496406 on Build GRCh38
rs557166858 Chr.2: 72496618-72496618 on Build GRCh38
rs560350703 Chr.2: 72496796-72496796 on Build GRCh38
rs767892582 Chr.2: 72496493-72496493 on Build GRCh38
rs370902491 Chr.2: 72496445-72496445 on Build GRCh38
rs992729198 Chr.2: 72496685-72496685 on Build GRCh38
rs1028275048 Chr.2: 72496819-72496819 on Build GRCh38
rs909761560 Chr.2: 72496744-72496744 on Build GRCh38
rs934005098 Chr.2: 72496842-72496842 on Build GRCh38
rs746989343 Chr.2: 72496440-72496440 on Build GRCh38
rs904350292 Chr.2: 72496667-72496667 on Build GRCh38
rs778921080 Chr.2: 72496529-72496529 on Build GRCh38
rs993155291 Chr.2: 72496560-72496560 on Build GRCh38
rs745607076 Chr.2: 72496589-72496589 on Build GRCh38
rs376652191 Chr.2: 72496630-72496630 on Build GRCh38
rs139290455 Chr.2: 72496468-72496468 on Build GRCh38
rs371861513 Chr.2: 72496506-72496506 on Build GRCh38
rs192743152 Chr.2: 72496674-72496674 on Build GRCh38
rs941411038 Chr.2: 72496764-72496764 on Build GRCh38
rs1024658393 Chr.2: 72496692-72496692 on Build GRCh38

View Full Product Details