Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74135305-74135773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLA3
Entrez Gene ID
388962
Gene Name
bolA family member 3
Gene Aliases
MMDS2
UniGene
Hs.61472
Gene Chromosome Location
Chr.2: 74135401-74147912 on Build GRCh38
Transcripts
NM_001035505.1 NM_212552.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343703 c.271C>A p.P91T Chr.2: 74135557-74135557 on Build GRCh38
1022911 c.313C>T p.P105S Chr.2: 74135604-74135604 on Build GRCh38
6612269 c.259-1G>T p.? Chr.2: 74135659-74135659 on Build GRCh38
1022910 c.314C>T p.P105L Chr.2: 74135603-74135603 on Build GRCh38
4874558 c.224C>T p.P75L Chr.2: 74135604-74135604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187615861 Chr.2: 74135430-74135430 on Build GRCh38
rs763511539 Chr.2: 74135491-74135491 on Build GRCh38
rs766621227 Chr.2: 74135558-74135558 on Build GRCh38
rs558987463 Chr.2: 74135330-74135331 on Build GRCh38
rs555683516 Chr.2: 74135744-74135744 on Build GRCh38
rs766637713 Chr.2: 74135662-74135662 on Build GRCh38
rs780663816 Chr.2: 74135474-74135474 on Build GRCh38
rs146584766 Chr.2: 74135720-74135720 on Build GRCh38
rs769219293 Chr.2: 74135620-74135620 on Build GRCh38
rs908781045 Chr.2: 74135628-74135628 on Build GRCh38
rs142868057 Chr.2: 74135676-74135676 on Build GRCh38
rs998761768 Chr.2: 74135359-74135359 on Build GRCh38
rs150626228 Chr.2: 74135598-74135598 on Build GRCh38
rs1049228537 Chr.2: 74135460-74135460 on Build GRCh38
rs780684324 Chr.2: 74135466-74135466 on Build GRCh38
rs191936501 Chr.2: 74135589-74135589 on Build GRCh38
rs2308245 Chr.2: 74135337-74135338 on Build GRCh38
rs750269271 Chr.2: 74135588-74135588 on Build GRCh38
rs886056325 Chr.2: 74135403-74135403 on Build GRCh38
rs200389155 Chr.2: 74135339-74135339 on Build GRCh38
rs72540771 Chr.2: 74135336-74135337 on Build GRCh38
rs941523861 Chr.2: 74135642-74135642 on Build GRCh38
rs777509576 Chr.2: 74135580-74135580 on Build GRCh38
rs202046520 Chr.2: 74135621-74135621 on Build GRCh38
rs180789482 Chr.2: 74135404-74135404 on Build GRCh38
rs756966193 Chr.2: 74135586-74135586 on Build GRCh38
rs774782921 Chr.2: 74135495-74135495 on Build GRCh38
rs923922886 Chr.2: 74135362-74135362 on Build GRCh38
rs774960372 Chr.2: 74135487-74135487 on Build GRCh38
rs761041660 Chr.2: 74135651-74135651 on Build GRCh38
rs776860491 Chr.2: 74135674-74135674 on Build GRCh38
rs769039782 Chr.2: 74135492-74135492 on Build GRCh38
rs148674363 Chr.2: 74135630-74135630 on Build GRCh38
rs751495007 Chr.2: 74135691-74135691 on Build GRCh38
rs773696418 Chr.2: 74135649-74135649 on Build GRCh38
rs762034480 Chr.2: 74135500-74135500 on Build GRCh38
rs759490061 Chr.2: 74135675-74135675 on Build GRCh38
rs992122474 Chr.2: 74135740-74135740 on Build GRCh38
rs549931538 Chr.2: 74135624-74135624 on Build GRCh38
rs923996415 Chr.2: 74135693-74135693 on Build GRCh38
rs751396525 Chr.2: 74135585-74135585 on Build GRCh38
rs376876176 Chr.2: 74135600-74135600 on Build GRCh38
rs769336989 Chr.2: 74135486-74135486 on Build GRCh38
rs745416387 Chr.2: 74135485-74135485 on Build GRCh38
rs753960028 Chr.2: 74135565-74135565 on Build GRCh38
rs77807718 Chr.2: 74135332-74135332 on Build GRCh38
rs146336257 Chr.2: 74135597-74135597 on Build GRCh38
rs35136039 Chr.2: 74135332-74135333 on Build GRCh38
rs566060433 Chr.2: 74135699-74135699 on Build GRCh38
rs886056326 Chr.2: 74135562-74135562 on Build GRCh38
rs1012917997 Chr.2: 74135418-74135418 on Build GRCh38
rs535272966 Chr.2: 74135436-74135436 on Build GRCh38
rs147523003 Chr.2: 74135335-74135336 on Build GRCh38
rs543245755 Chr.2: 74135502-74135502 on Build GRCh38
rs758410967 Chr.2: 74135572-74135572 on Build GRCh38
rs895368250 Chr.2: 74135408-74135408 on Build GRCh38
rs866949521 Chr.2: 74135622-74135622 on Build GRCh38
rs561184018 Chr.2: 74135504-74135504 on Build GRCh38
rs957222446 Chr.2: 74135442-74135442 on Build GRCh38
rs577621123 Chr.2: 74135331-74135331 on Build GRCh38
rs72387988 Chr.2: 74135333-74135334 on Build GRCh38
rs567877959 Chr.2: 74135384-74135384 on Build GRCh38
rs200938488 Chr.2: 74135635-74135635 on Build GRCh38

View Full Product Details