Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74361073-74361575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN1
Entrez Gene ID
1639
Gene Name
dynactin subunit 1
Gene Aliases
DAP-150, DP-150, P135
UniGene
Hs.516111
Gene Chromosome Location
Chr.2: 74361154-74392087 on Build GRCh38
Transcripts
NM_001135040.2 NM_001135041.2 NM_001190836.1 NM_001190837.1 NM_004082.4 NM_023019.3 NR_033935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026381 c.3775C>T p.R1259* Chr.2: 74361561-74361561 on Build GRCh38
6768740 c.3781C>T p.R1261W Chr.2: 74361555-74361555 on Build GRCh38
3939163 c.3774G>A p.Q1258Q Chr.2: 74361562-74361562 on Build GRCh38
4655317 c.3772C>A p.Q1258K Chr.2: 74361564-74361564 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758979848 Chr.2: 74361319-74361319 on Build GRCh38
rs1051686630 Chr.2: 74361500-74361500 on Build GRCh38
rs985777861 Chr.2: 74361295-74361295 on Build GRCh38
rs753974651 Chr.2: 74361515-74361515 on Build GRCh38
rs745491164 Chr.2: 74361461-74361461 on Build GRCh38
rs377080829 Chr.2: 74361487-74361487 on Build GRCh38
rs7570481 Chr.2: 74361141-74361141 on Build GRCh38
rs544852465 Chr.2: 74361253-74361253 on Build GRCh38
rs1053169737 Chr.2: 74361458-74361458 on Build GRCh38
rs533691355 Chr.2: 74361325-74361325 on Build GRCh38
rs777649011 Chr.2: 74361377-74361377 on Build GRCh38
rs549059722 Chr.2: 74361416-74361416 on Build GRCh38
rs768719210 Chr.2: 74361202-74361202 on Build GRCh38
rs748065208 Chr.2: 74361374-74361374 on Build GRCh38
rs538932463 Chr.2: 74361540-74361540 on Build GRCh38
rs948687816 Chr.2: 74361124-74361124 on Build GRCh38
rs947292936 Chr.2: 74361312-74361312 on Build GRCh38
rs756299486 Chr.2: 74361429-74361429 on Build GRCh38
rs745906832 Chr.2: 74361242-74361242 on Build GRCh38
rs200676625 Chr.2: 74361493-74361493 on Build GRCh38
rs771866623 Chr.2: 74361375-74361375 on Build GRCh38
rs773699546 Chr.2: 74361183-74361183 on Build GRCh38
rs17009890 Chr.2: 74361227-74361227 on Build GRCh38
rs773669685 Chr.2: 74361511-74361511 on Build GRCh38
rs963066378 Chr.2: 74361376-74361376 on Build GRCh38
rs755013572 Chr.2: 74361526-74361526 on Build GRCh38
rs763470976 Chr.2: 74361400-74361400 on Build GRCh38
rs777509420 Chr.2: 74361448-74361448 on Build GRCh38
rs774703501 Chr.2: 74361491-74361491 on Build GRCh38
rs560344779 Chr.2: 74361512-74361512 on Build GRCh38
rs762282945 Chr.2: 74361492-74361492 on Build GRCh38
rs767833292 Chr.2: 74361419-74361419 on Build GRCh38
rs779040394 Chr.2: 74361369-74361369 on Build GRCh38
rs776019307 Chr.2: 74361398-74361398 on Build GRCh38
rs757513753 Chr.2: 74361354-74361354 on Build GRCh38
rs368049781 Chr.2: 74361450-74361450 on Build GRCh38
rs544736810 Chr.2: 74361106-74361106 on Build GRCh38
rs140263124 Chr.2: 74361260-74361260 on Build GRCh38
rs529485093 Chr.2: 74361217-74361217 on Build GRCh38
rs998780750 Chr.2: 74361338-74361338 on Build GRCh38
rs1000218255 Chr.2: 74361269-74361269 on Build GRCh38
rs971879611 Chr.2: 74361237-74361237 on Build GRCh38
rs763358994 Chr.2: 74361326-74361326 on Build GRCh38
rs746888495 Chr.2: 74361449-74361449 on Build GRCh38
rs1030107900 Chr.2: 74361505-74361505 on Build GRCh38
rs917118890 Chr.2: 74361422-74361422 on Build GRCh38
rs1006433114 Chr.2: 74361405-74361405 on Build GRCh38
rs1007184904 Chr.2: 74361363-74361363 on Build GRCh38
rs751431467 Chr.2: 74361533-74361533 on Build GRCh38
rs374006190 Chr.2: 74361508-74361508 on Build GRCh38
rs776143255 Chr.2: 74361385-74361385 on Build GRCh38
rs753923005 Chr.2: 74361441-74361441 on Build GRCh38
rs185694177 Chr.2: 74361394-74361394 on Build GRCh38
rs774992924 Chr.2: 74361472-74361472 on Build GRCh38
rs772783174 Chr.2: 74361271-74361271 on Build GRCh38
rs1029876124 Chr.2: 74361342-74361342 on Build GRCh38
rs3177776 Chr.2: 74361165-74361165 on Build GRCh38
rs910189036 Chr.2: 74361305-74361305 on Build GRCh38
rs11555696 Chr.2: 74361478-74361478 on Build GRCh38
rs562207772 Chr.2: 74361161-74361161 on Build GRCh38
rs770516270 Chr.2: 74361322-74361322 on Build GRCh38
rs750593731 Chr.2: 74361359-74361359 on Build GRCh38
rs1023864913 Chr.2: 74361123-74361123 on Build GRCh38
rs766653950 Chr.2: 74361513-74361513 on Build GRCh38
rs764449369 Chr.2: 74361407-74361407 on Build GRCh38
rs756362040 Chr.2: 74361248-74361248 on Build GRCh38
rs889980270 Chr.2: 74361517-74361517 on Build GRCh38
rs750648692 Chr.2: 74361426-74361426 on Build GRCh38
rs779676968 Chr.2: 74361553-74361553 on Build GRCh38
rs146083590 Chr.2: 74361537-74361537 on Build GRCh38
rs770445860 Chr.2: 74361306-74361306 on Build GRCh38
rs768025820 Chr.2: 74361555-74361555 on Build GRCh38
rs745876269 Chr.2: 74361189-74361189 on Build GRCh38
rs764549033 Chr.2: 74361331-74361331 on Build GRCh38
rs1017280550 Chr.2: 74361372-74361372 on Build GRCh38
rs755766984 Chr.2: 74361548-74361548 on Build GRCh38
rs756246243 Chr.2: 74361368-74361368 on Build GRCh38
rs371589609 Chr.2: 74361451-74361451 on Build GRCh38
rs769386100 Chr.2: 74361462-74361462 on Build GRCh38
rs1016440744 Chr.2: 74361154-74361154 on Build GRCh38
rs142053202 Chr.2: 74361332-74361332 on Build GRCh38
rs748010130 Chr.2: 74361247-74361247 on Build GRCh38
rs760410729 Chr.2: 74361289-74361289 on Build GRCh38
rs567051449 Chr.2: 74361442-74361442 on Build GRCh38
rs757129747 Chr.2: 74361169-74361169 on Build GRCh38
rs770626130 Chr.2: 74361382-74361382 on Build GRCh38
rs781204224 Chr.2: 74361355-74361355 on Build GRCh38
rs746727753 Chr.2: 74361381-74361381 on Build GRCh38
rs772261569 Chr.2: 74361253-74361261 on Build GRCh38
rs151150356 Chr.2: 74361396-74361396 on Build GRCh38
rs553822174 Chr.2: 74361554-74361554 on Build GRCh38
rs962484837 Chr.2: 74361206-74361206 on Build GRCh38
rs780309814 Chr.2: 74361439-74361439 on Build GRCh38
rs776222478 Chr.2: 74361317-74361317 on Build GRCh38

View Full Product Details