Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74553787-74554274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK1
Entrez Gene ID
1796
Gene Name
docking protein 1
Gene Aliases
P62DOK, pp62
UniGene
Hs.103854
Gene Chromosome Location
Chr.2: 74549020-74557551 on Build GRCh38
Transcripts
NM_001197260.1 NM_001318866.1 NM_001318867.1 NM_001318868.1 NM_001318869.1 NM_001381.4 NM_001289164.1 NM_032603.3 XM_011533134.2

Gene Symbol
LOXL3
Entrez Gene ID
84695
Gene Name
lysyl oxidase like 3
Gene Aliases
LOXL
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74532258-74555709 on Build GRCh38
Transcripts
NM_001197260.1 NM_001318866.1 NM_001318867.1 NM_001318868.1 NM_001318869.1 NM_001381.4 NM_001289164.1 NM_032603.3 XM_011533134.2

SNP Detail

SNP ID Chromosome Location
rs988847262 Chr.2: 74554112-74554112 on Build GRCh38
rs567408062 Chr.2: 74553817-74553817 on Build GRCh38
rs996075314 Chr.2: 74553980-74553981 on Build GRCh38
rs953736889 Chr.2: 74554003-74554003 on Build GRCh38
rs886486017 Chr.2: 74553867-74553867 on Build GRCh38
rs549983859 Chr.2: 74554028-74554028 on Build GRCh38
rs764423076 Chr.2: 74553943-74553944 on Build GRCh38
rs113018584 Chr.2: 74553944-74553944 on Build GRCh38
rs1016978989 Chr.2: 74554064-74554064 on Build GRCh38
rs1052501327 Chr.2: 74554226-74554226 on Build GRCh38
rs957286882 Chr.2: 74554041-74554041 on Build GRCh38
rs571371548 Chr.2: 74554092-74554092 on Build GRCh38
rs553655021 Chr.2: 74554161-74554161 on Build GRCh38
rs781370619 Chr.2: 74554135-74554135 on Build GRCh38
rs202031999 Chr.2: 74553943-74553943 on Build GRCh38
rs972514912 Chr.2: 74554123-74554123 on Build GRCh38
rs1029262690 Chr.2: 74553995-74553995 on Build GRCh38
rs930617110 Chr.2: 74553827-74553827 on Build GRCh38
rs925316646 Chr.2: 74554185-74554185 on Build GRCh38
rs918319859 Chr.2: 74554128-74554128 on Build GRCh38
rs999600156 Chr.2: 74553823-74553823 on Build GRCh38
rs960556286 Chr.2: 74554133-74554133 on Build GRCh38
rs1044093875 Chr.2: 74553815-74553815 on Build GRCh38
rs755257892 Chr.2: 74553856-74553856 on Build GRCh38
rs151276951 Chr.2: 74554104-74554104 on Build GRCh38
rs530332428 Chr.2: 74553862-74553862 on Build GRCh38
rs900363561 Chr.2: 74553913-74553913 on Build GRCh38
rs186141934 Chr.2: 74553838-74553838 on Build GRCh38
rs867183682 Chr.2: 74554138-74554138 on Build GRCh38
rs77436581 Chr.2: 74554248-74554248 on Build GRCh38
rs538899929 Chr.2: 74554156-74554156 on Build GRCh38
rs531506448 Chr.2: 74554048-74554050 on Build GRCh38
rs947925304 Chr.2: 74554153-74554153 on Build GRCh38
rs935317226 Chr.2: 74554188-74554188 on Build GRCh38
rs902043197 Chr.2: 74553868-74553868 on Build GRCh38
rs1019950785 Chr.2: 74554126-74554126 on Build GRCh38
rs997627200 Chr.2: 74553969-74553969 on Build GRCh38
rs962395208 Chr.2: 74554074-74554074 on Build GRCh38
rs11688169 Chr.2: 74554006-74554006 on Build GRCh38
rs985147936 Chr.2: 74554043-74554043 on Build GRCh38

View Full Product Details