Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79744249-79744791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA2
Entrez Gene ID
1496
Gene Name
catenin alpha 2
Gene Aliases
CAP-R, CAPR, CT114, CTNR
UniGene
Hs.167368
Gene Chromosome Location
Chr.2: 79512934-80648868 on Build GRCh38
Transcripts
NM_001164883.1 NM_001282597.2 NM_001282598.1 NM_004389.3 XM_011532555.2 XM_011532556.2 XM_017003403.1 XM_017003404.1 XM_017003405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758044 c.351A>C p.Q117H Chr.2: 79744533-79744533 on Build GRCh38
5914904 c.343G>A p.E115K Chr.2: 79744525-79744525 on Build GRCh38
6159106 c.197A>T p.Q66L Chr.2: 79744481-79744481 on Build GRCh38
3000679 c.289C>T p.R97C Chr.2: 79744573-79744573 on Build GRCh38
375040 c.172C>T p.H58Y Chr.2: 79744456-79744456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10167435 Chr.2: 79744289-79744289 on Build GRCh38
rs749396664 Chr.2: 79744457-79744457 on Build GRCh38
rs918399234 Chr.2: 79744685-79744685 on Build GRCh38
rs777188641 Chr.2: 79744624-79744624 on Build GRCh38
rs746060871 Chr.2: 79744685-79744686 on Build GRCh38
rs191974108 Chr.2: 79744326-79744326 on Build GRCh38
rs771428803 Chr.2: 79744533-79744533 on Build GRCh38
rs544674695 Chr.2: 79744679-79744679 on Build GRCh38
rs375648846 Chr.2: 79744308-79744308 on Build GRCh38
rs376745664 Chr.2: 79744574-79744574 on Build GRCh38
rs925522323 Chr.2: 79744661-79744661 on Build GRCh38
rs761552911 Chr.2: 79744526-79744526 on Build GRCh38
rs775095830 Chr.2: 79744630-79744630 on Build GRCh38
rs775044854 Chr.2: 79744547-79744547 on Build GRCh38
rs776430147 Chr.2: 79744501-79744501 on Build GRCh38
rs149416893 Chr.2: 79744768-79744768 on Build GRCh38
rs115094550 Chr.2: 79744694-79744694 on Build GRCh38
rs771412365 Chr.2: 79744613-79744613 on Build GRCh38
rs747386866 Chr.2: 79744612-79744612 on Build GRCh38
rs200093329 Chr.2: 79744518-79744518 on Build GRCh38
rs748265137 Chr.2: 79744597-79744597 on Build GRCh38
rs751405871 Chr.2: 79744552-79744552 on Build GRCh38
rs772576068 Chr.2: 79744354-79744356 on Build GRCh38
rs761060937 Chr.2: 79744502-79744502 on Build GRCh38
rs1023781680 Chr.2: 79744467-79744467 on Build GRCh38
rs755217215 Chr.2: 79744459-79744459 on Build GRCh38
rs769618732 Chr.2: 79744556-79744556 on Build GRCh38
rs545090625 Chr.2: 79744683-79744684 on Build GRCh38
rs370052135 Chr.2: 79744534-79744534 on Build GRCh38
rs556012450 Chr.2: 79744611-79744611 on Build GRCh38
rs533353778 Chr.2: 79744687-79744687 on Build GRCh38
rs779424622 Chr.2: 79744305-79744305 on Build GRCh38
rs752522908 Chr.2: 79744363-79744363 on Build GRCh38
rs573582994 Chr.2: 79744374-79744374 on Build GRCh38
rs767623788 Chr.2: 79744572-79744572 on Build GRCh38
rs886305812 Chr.2: 79744720-79744720 on Build GRCh38
rs769137882 Chr.2: 79744628-79744628 on Build GRCh38
rs775867046 Chr.2: 79744368-79744368 on Build GRCh38
rs747471556 Chr.2: 79744530-79744530 on Build GRCh38
rs183893244 Chr.2: 79744625-79744625 on Build GRCh38
rs955654943 Chr.2: 79744595-79744595 on Build GRCh38
rs979695577 Chr.2: 79744658-79744658 on Build GRCh38
rs138888063 Chr.2: 79744587-79744587 on Build GRCh38
rs762689721 Chr.2: 79744549-79744549 on Build GRCh38
rs377364195 Chr.2: 79744519-79744519 on Build GRCh38
rs757294238 Chr.2: 79744419-79744419 on Build GRCh38
rs773262596 Chr.2: 79744562-79744562 on Build GRCh38
rs941208005 Chr.2: 79744663-79744663 on Build GRCh38
rs373460375 Chr.2: 79744365-79744365 on Build GRCh38
rs896170322 Chr.2: 79744716-79744716 on Build GRCh38
rs17017796 Chr.2: 79744351-79744351 on Build GRCh38
rs150332729 Chr.2: 79744343-79744343 on Build GRCh38
rs374216983 Chr.2: 79744684-79744685 on Build GRCh38
rs373818447 Chr.2: 79744573-79744573 on Build GRCh38
rs773644253 Chr.2: 79744527-79744527 on Build GRCh38
rs764938006 Chr.2: 79744546-79744546 on Build GRCh38
rs1033170641 Chr.2: 79744542-79744542 on Build GRCh38
rs763757402 Chr.2: 79744550-79744550 on Build GRCh38
rs780217471 Chr.2: 79744426-79744426 on Build GRCh38
rs922592327 Chr.2: 79744278-79744278 on Build GRCh38
rs947670295 Chr.2: 79744742-79744742 on Build GRCh38
rs374360278 Chr.2: 79744686-79744686 on Build GRCh38
rs1044828839 Chr.2: 79744763-79744763 on Build GRCh38
rs200888828 Chr.2: 79744563-79744563 on Build GRCh38
rs758676012 Chr.2: 79744598-79744598 on Build GRCh38
rs759102402 Chr.2: 79744545-79744545 on Build GRCh38
rs779169912 Chr.2: 79744575-79744575 on Build GRCh38
rs564459787 Chr.2: 79744686-79744686 on Build GRCh38
rs896126282 Chr.2: 79744373-79744373 on Build GRCh38
rs1050260312 Chr.2: 79744290-79744290 on Build GRCh38
rs779076049 Chr.2: 79744498-79744498 on Build GRCh38
rs748517537 Chr.2: 79744515-79744515 on Build GRCh38
rs751493653 Chr.2: 79744404-79744404 on Build GRCh38

View Full Product Details