Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85401441-85401948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPG
Entrez Gene ID
822
Gene Name
capping actin protein, gelsolin like
Gene Aliases
AFCP, HEL-S-66, MCP
UniGene
Hs.516155
Gene Chromosome Location
Chr.2: 85394748-85418467 on Build GRCh38
Transcripts
NM_001256139.1 NM_001256140.1 NM_001320732.1 NM_001320733.1 NM_001320734.1 NM_001747.3 XM_011533122.1 XM_011533123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583748 c.188T>G p.L63R Chr.2: 85401793-85401793 on Build GRCh38
5973907 c.211C>T p.R71W Chr.2: 85401669-85401669 on Build GRCh38
6159030 c.133G>T p.G45W Chr.2: 85401848-85401848 on Build GRCh38
3583749 c.136G>T p.D46Y Chr.2: 85401845-85401845 on Build GRCh38
5781336 c.351+3C>T p.? Chr.2: 85401526-85401526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762696731 Chr.2: 85401636-85401636 on Build GRCh38
rs891699379 Chr.2: 85401676-85401676 on Build GRCh38
rs778301507 Chr.2: 85401886-85401886 on Build GRCh38
rs1044665190 Chr.2: 85401716-85401716 on Build GRCh38
rs773528263 Chr.2: 85401567-85401567 on Build GRCh38
rs2229668 Chr.2: 85401860-85401860 on Build GRCh38
rs779149972 Chr.2: 85401561-85401561 on Build GRCh38
rs764211779 Chr.2: 85401759-85401759 on Build GRCh38
rs527935851 Chr.2: 85401920-85401920 on Build GRCh38
rs771699453 Chr.2: 85401827-85401827 on Build GRCh38
rs756599713 Chr.2: 85401876-85401876 on Build GRCh38
rs373269135 Chr.2: 85401850-85401850 on Build GRCh38
rs759583335 Chr.2: 85401764-85401764 on Build GRCh38
rs752595362 Chr.2: 85401891-85401892 on Build GRCh38
rs755099333 Chr.2: 85401603-85401603 on Build GRCh38
rs964821692 Chr.2: 85401556-85401556 on Build GRCh38
rs768722003 Chr.2: 85401552-85401552 on Build GRCh38
rs149746691 Chr.2: 85401533-85401533 on Build GRCh38
rs754174223 Chr.2: 85401915-85401915 on Build GRCh38
rs763059649 Chr.2: 85401755-85401755 on Build GRCh38
rs750757077 Chr.2: 85401774-85401774 on Build GRCh38
rs780277550 Chr.2: 85401496-85401496 on Build GRCh38
rs766855151 Chr.2: 85401908-85401908 on Build GRCh38
rs367763988 Chr.2: 85401565-85401565 on Build GRCh38
rs751243644 Chr.2: 85401592-85401592 on Build GRCh38
rs756006551 Chr.2: 85401668-85401668 on Build GRCh38
rs760621075 Chr.2: 85401747-85401747 on Build GRCh38
rs747761634 Chr.2: 85401826-85401826 on Build GRCh38
rs747754392 Chr.2: 85401888-85401888 on Build GRCh38
rs763900464 Chr.2: 85401639-85401639 on Build GRCh38
rs1009015112 Chr.2: 85401785-85401785 on Build GRCh38
rs761040798 Chr.2: 85401503-85401503 on Build GRCh38
rs760943110 Chr.2: 85401907-85401907 on Build GRCh38
rs747619453 Chr.2: 85401710-85401710 on Build GRCh38
rs1018937424 Chr.2: 85401831-85401831 on Build GRCh38
rs765040897 Chr.2: 85401516-85401516 on Build GRCh38
rs762641569 Chr.2: 85401583-85401583 on Build GRCh38
rs956323297 Chr.2: 85401587-85401587 on Build GRCh38
rs142150224 Chr.2: 85401877-85401877 on Build GRCh38
rs777441461 Chr.2: 85401890-85401890 on Build GRCh38
rs746350508 Chr.2: 85401633-85401633 on Build GRCh38
rs770680588 Chr.2: 85401840-85401840 on Build GRCh38
rs773757356 Chr.2: 85401501-85401501 on Build GRCh38
rs138195193 Chr.2: 85401857-85401857 on Build GRCh38
rs368493475 Chr.2: 85401490-85401490 on Build GRCh38
rs549982416 Chr.2: 85401741-85401741 on Build GRCh38
rs776366363 Chr.2: 85401895-85401895 on Build GRCh38
rs748876636 Chr.2: 85401719-85401719 on Build GRCh38
rs758551430 Chr.2: 85401619-85401619 on Build GRCh38
rs755285701 Chr.2: 85401916-85401916 on Build GRCh38
rs74396419 Chr.2: 85401635-85401635 on Build GRCh38
rs777044133 Chr.2: 85401625-85401625 on Build GRCh38
rs773927919 Chr.2: 85401731-85401731 on Build GRCh38
rs757065726 Chr.2: 85401593-85401593 on Build GRCh38
rs772994075 Chr.2: 85401739-85401739 on Build GRCh38
rs138976851 Chr.2: 85401849-85401849 on Build GRCh38
rs767415106 Chr.2: 85401888-85401889 on Build GRCh38
rs375004748 Chr.2: 85401609-85401609 on Build GRCh38
rs1053715341 Chr.2: 85401760-85401760 on Build GRCh38
rs777180092 Chr.2: 85401576-85401576 on Build GRCh38
rs775328091 Chr.2: 85401582-85401582 on Build GRCh38
rs753219092 Chr.2: 85401507-85401507 on Build GRCh38
rs747222141 Chr.2: 85401693-85401693 on Build GRCh38
rs758457470 Chr.2: 85401670-85401670 on Build GRCh38
rs371246622 Chr.2: 85401608-85401608 on Build GRCh38
rs373390653 Chr.2: 85401906-85401906 on Build GRCh38
rs145160691 Chr.2: 85401669-85401669 on Build GRCh38
rs750346858 Chr.2: 85401594-85401594 on Build GRCh38
rs921095576 Chr.2: 85401615-85401616 on Build GRCh38
rs140150295 Chr.2: 85401889-85401889 on Build GRCh38
rs780749454 Chr.2: 85401778-85401778 on Build GRCh38
rs776599208 Chr.2: 85401841-85401841 on Build GRCh38
rs771321418 Chr.2: 85401623-85401623 on Build GRCh38
rs145323184 Chr.2: 85401862-85401862 on Build GRCh38
rs905712438 Chr.2: 85401647-85401647 on Build GRCh38
rs747556915 Chr.2: 85401568-85401568 on Build GRCh38
rs774052021 Chr.2: 85401646-85401646 on Build GRCh38
rs375167913 Chr.2: 85401545-85401545 on Build GRCh38
rs746641476 Chr.2: 85401893-85401893 on Build GRCh38
rs11539104 Chr.2: 85401882-85401882 on Build GRCh38
rs772395120 Chr.2: 85401492-85401492 on Build GRCh38
rs749388241 Chr.2: 85401546-85401546 on Build GRCh38
rs908836213 Chr.2: 85401611-85401611 on Build GRCh38
rs565837522 Chr.2: 85401768-85401768 on Build GRCh38
rs770355071 Chr.2: 85401713-85401713 on Build GRCh38
rs777922930 Chr.2: 85401686-85401686 on Build GRCh38
rs754432574 Chr.2: 85401880-85401880 on Build GRCh38
rs745794232 Chr.2: 85401899-85401899 on Build GRCh38
rs761482009 Chr.2: 85401735-85401735 on Build GRCh38
rs768077096 Chr.2: 85401801-85401801 on Build GRCh38
rs369804332 Chr.2: 85401793-85401793 on Build GRCh38
rs751748879 Chr.2: 85401763-85401763 on Build GRCh38
rs775274344 Chr.2: 85401901-85401901 on Build GRCh38
rs748348412 Chr.2: 85401563-85401563 on Build GRCh38
rs567151587 Chr.2: 85401863-85401863 on Build GRCh38
rs767325894 Chr.2: 85401655-85401655 on Build GRCh38
rs779720878 Chr.2: 85401717-85401717 on Build GRCh38
rs35899313 Chr.2: 85401581-85401581 on Build GRCh38
rs772448274 Chr.2: 85401564-85401564 on Build GRCh38
rs781769439 Chr.2: 85401642-85401642 on Build GRCh38
rs749794313 Chr.2: 85401800-85401800 on Build GRCh38
rs199693005 Chr.2: 85401751-85401751 on Build GRCh38
rs2229669 Chr.2: 85401622-85401622 on Build GRCh38
rs750644909 Chr.2: 85401875-85401875 on Build GRCh38
rs755446246 Chr.2: 85401881-85401881 on Build GRCh38
rs771283910 Chr.2: 85401569-85401569 on Build GRCh38
rs760528962 Chr.2: 85401835-85401835 on Build GRCh38
rs768184767 Chr.2: 85401724-85401724 on Build GRCh38
rs367976290 Chr.2: 85401845-85401845 on Build GRCh38
rs1000920358 Chr.2: 85401788-85401788 on Build GRCh38
rs201082741 Chr.2: 85401515-85401515 on Build GRCh38
rs778447049 Chr.2: 85401821-85401821 on Build GRCh38
rs564215346 Chr.2: 85401697-85401697 on Build GRCh38
rs11539101 Chr.2: 85401674-85401674 on Build GRCh38
rs368233778 Chr.2: 85401517-85401517 on Build GRCh38
rs527327511 Chr.2: 85401595-85401595 on Build GRCh38
rs939782679 Chr.2: 85401660-85401660 on Build GRCh38
rs771795135 Chr.2: 85401737-85401737 on Build GRCh38
rs764009099 Chr.2: 85401591-85401591 on Build GRCh38
rs772840062 Chr.2: 85401833-85401833 on Build GRCh38
rs767686851 Chr.2: 85401904-85401904 on Build GRCh38
rs531742845 Chr.2: 85401712-85401712 on Build GRCh38
rs770733881 Chr.2: 85401894-85401894 on Build GRCh38
rs769711684 Chr.2: 85401900-85401900 on Build GRCh38
rs372064925 Chr.2: 85401548-85401548 on Build GRCh38
rs761995248 Chr.2: 85401902-85401902 on Build GRCh38
rs751535351 Chr.2: 85401543-85401543 on Build GRCh38
rs1013132391 Chr.2: 85401541-85401541 on Build GRCh38
rs757456618 Chr.2: 85401764-85401764 on Build GRCh38

View Full Product Details