Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85607942-85608444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf68
Entrez Gene ID
388969
Gene Name
chromosome 2 open reading frame 68
Gene Aliases
HCRCN81
UniGene
Hs.516159
Gene Chromosome Location
Chr.2: 85605253-85612073 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

Gene Symbol
USP39
Entrez Gene ID
10713
Gene Name
ubiquitin specific peptidase 39
Gene Aliases
65K, CGI-21, HSPC332, SAD1, SNRNP65
UniGene
Hs.654745
Gene Chromosome Location
Chr.2: 85602842-85649284 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

SNP Detail

SNP ID Chromosome Location
rs950167552 Chr.2: 85608179-85608179 on Build GRCh38
rs369846835 Chr.2: 85608415-85608416 on Build GRCh38
rs1027519825 Chr.2: 85608258-85608258 on Build GRCh38
rs1006475908 Chr.2: 85608412-85608413 on Build GRCh38
rs1055005137 Chr.2: 85608020-85608020 on Build GRCh38
rs751864005 Chr.2: 85608080-85608081 on Build GRCh38
rs889439387 Chr.2: 85608409-85608409 on Build GRCh38
rs777025586 Chr.2: 85608296-85608296 on Build GRCh38
rs571715467 Chr.2: 85608192-85608192 on Build GRCh38
rs192924318 Chr.2: 85608069-85608069 on Build GRCh38
rs890086523 Chr.2: 85608161-85608161 on Build GRCh38
rs141806397 Chr.2: 85608065-85608065 on Build GRCh38
rs545386121 Chr.2: 85608102-85608102 on Build GRCh38
rs924229066 Chr.2: 85608101-85608101 on Build GRCh38
rs935472184 Chr.2: 85608103-85608103 on Build GRCh38
rs192206692 Chr.2: 85608328-85608328 on Build GRCh38
rs973290181 Chr.2: 85608173-85608173 on Build GRCh38
rs985671420 Chr.2: 85608135-85608135 on Build GRCh38
rs184687326 Chr.2: 85608378-85608378 on Build GRCh38
rs114521018 Chr.2: 85608157-85608157 on Build GRCh38
rs744716 Chr.2: 85608091-85608091 on Build GRCh38
rs557367045 Chr.2: 85608136-85608136 on Build GRCh38
rs761262145 Chr.2: 85608264-85608264 on Build GRCh38
rs1023679943 Chr.2: 85608294-85608294 on Build GRCh38
rs1898528 Chr.2: 85608280-85608280 on Build GRCh38
rs1014846218 Chr.2: 85607970-85607970 on Build GRCh38
rs189473807 Chr.2: 85608407-85608407 on Build GRCh38
rs952824701 Chr.2: 85608059-85608059 on Build GRCh38
rs1047051094 Chr.2: 85608093-85608093 on Build GRCh38
rs1017160995 Chr.2: 85608414-85608414 on Build GRCh38
rs551104597 Chr.2: 85608038-85608038 on Build GRCh38
rs927277798 Chr.2: 85608225-85608225 on Build GRCh38
rs564469610 Chr.2: 85607975-85607975 on Build GRCh38
rs1898527 Chr.2: 85608278-85608278 on Build GRCh38
rs995061398 Chr.2: 85608255-85608255 on Build GRCh38
rs916615865 Chr.2: 85608068-85608068 on Build GRCh38
rs375349529 Chr.2: 85608345-85608345 on Build GRCh38
rs997873787 Chr.2: 85608100-85608100 on Build GRCh38
rs185068302 Chr.2: 85608237-85608237 on Build GRCh38
rs946588585 Chr.2: 85608271-85608271 on Build GRCh38
rs147187251 Chr.2: 85608235-85608235 on Build GRCh38
rs1011121767 Chr.2: 85608036-85608036 on Build GRCh38
rs918694477 Chr.2: 85608173-85608173 on Build GRCh38
rs567828986 Chr.2: 85608127-85608127 on Build GRCh38
rs749436960 Chr.2: 85608116-85608116 on Build GRCh38
rs981489762 Chr.2: 85608195-85608195 on Build GRCh38
rs553070536 Chr.2: 85608164-85608164 on Build GRCh38
rs982152076 Chr.2: 85608356-85608356 on Build GRCh38
rs779430615 Chr.2: 85608317-85608317 on Build GRCh38
rs541365956 Chr.2: 85608395-85608395 on Build GRCh38
rs973274746 Chr.2: 85607985-85607985 on Build GRCh38
rs549576879 Chr.2: 85608126-85608126 on Build GRCh38
rs751481652 Chr.2: 85607976-85607976 on Build GRCh38
rs188729629 Chr.2: 85608248-85608248 on Build GRCh38

View Full Product Details