Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85608663-85609207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf68
Entrez Gene ID
388969
Gene Name
chromosome 2 open reading frame 68
Gene Aliases
HCRCN81
UniGene
Hs.516159
Gene Chromosome Location
Chr.2: 85605253-85612073 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

Gene Symbol
USP39
Entrez Gene ID
10713
Gene Name
ubiquitin specific peptidase 39
Gene Aliases
65K, CGI-21, HSPC332, SAD1, SNRNP65
UniGene
Hs.654745
Gene Chromosome Location
Chr.2: 85602842-85649284 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408015 c.423G>A p.T141T Chr.2: 85609023-85609023 on Build GRCh38
1307048 c.453C>G p.L151L Chr.2: 85608993-85608993 on Build GRCh38
6442102 c.469G>A p.E157K Chr.2: 85608977-85608977 on Build GRCh38
1023382 c.461G>A p.R154H Chr.2: 85608985-85608985 on Build GRCh38
5496824 c.485G>A p.R162H Chr.2: 85608961-85608961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543824731 Chr.2: 85608940-85608940 on Build GRCh38
rs10170768 Chr.2: 85608742-85608742 on Build GRCh38
rs761640978 Chr.2: 85609047-85609047 on Build GRCh38
rs760596326 Chr.2: 85609066-85609066 on Build GRCh38
rs966187273 Chr.2: 85609142-85609142 on Build GRCh38
rs746389238 Chr.2: 85609104-85609104 on Build GRCh38
rs547310328 Chr.2: 85608931-85608931 on Build GRCh38
rs569335915 Chr.2: 85609088-85609088 on Build GRCh38
rs200445699 Chr.2: 85608730-85608730 on Build GRCh38
rs550907332 Chr.2: 85608726-85608729 on Build GRCh38
rs898148471 Chr.2: 85609123-85609123 on Build GRCh38
rs528498191 Chr.2: 85608952-85608952 on Build GRCh38
rs1047765295 Chr.2: 85609177-85609177 on Build GRCh38
rs369815691 Chr.2: 85608903-85608903 on Build GRCh38
rs769485684 Chr.2: 85609024-85609024 on Build GRCh38
rs369730273 Chr.2: 85608756-85608756 on Build GRCh38
rs1003670028 Chr.2: 85609032-85609032 on Build GRCh38
rs971802741 Chr.2: 85609147-85609147 on Build GRCh38
rs765248533 Chr.2: 85609071-85609071 on Build GRCh38
rs757299153 Chr.2: 85608946-85608946 on Build GRCh38
rs144427574 Chr.2: 85609111-85609111 on Build GRCh38
rs752757700 Chr.2: 85609072-85609072 on Build GRCh38
rs184488528 Chr.2: 85609003-85609003 on Build GRCh38
rs747486270 Chr.2: 85608905-85608905 on Build GRCh38
rs1023650571 Chr.2: 85608749-85608749 on Build GRCh38
rs755094987 Chr.2: 85608985-85608985 on Build GRCh38
rs774034423 Chr.2: 85609040-85609040 on Build GRCh38
rs970807097 Chr.2: 85608757-85608757 on Build GRCh38
rs758590518 Chr.2: 85608989-85608989 on Build GRCh38
rs912100100 Chr.2: 85609118-85609118 on Build GRCh38
rs758761543 Chr.2: 85609133-85609133 on Build GRCh38
rs759068950 Chr.2: 85608932-85608932 on Build GRCh38
rs759750067 Chr.2: 85609092-85609104 on Build GRCh38
rs1024118944 Chr.2: 85609006-85609006 on Build GRCh38
rs772285102 Chr.2: 85609114-85609114 on Build GRCh38
rs376446305 Chr.2: 85608902-85608902 on Build GRCh38
rs372058948 Chr.2: 85608850-85608850 on Build GRCh38
rs932110125 Chr.2: 85608835-85608835 on Build GRCh38
rs373678804 Chr.2: 85608958-85608958 on Build GRCh38
rs377491416 Chr.2: 85608979-85608979 on Build GRCh38
rs781679335 Chr.2: 85609096-85609096 on Build GRCh38
rs180693413 Chr.2: 85608747-85608747 on Build GRCh38
rs774755717 Chr.2: 85608985-85608986 on Build GRCh38
rs5832651 Chr.2: 85608729-85608729 on Build GRCh38
rs66822657 Chr.2: 85608702-85608702 on Build GRCh38
rs779050104 Chr.2: 85608986-85608986 on Build GRCh38
rs762514760 Chr.2: 85609033-85609033 on Build GRCh38
rs989708089 Chr.2: 85608818-85608818 on Build GRCh38
rs201140439 Chr.2: 85608974-85608974 on Build GRCh38
rs189456850 Chr.2: 85609077-85609077 on Build GRCh38
rs982505998 Chr.2: 85608768-85608768 on Build GRCh38
rs369747469 Chr.2: 85608987-85608987 on Build GRCh38
rs778006796 Chr.2: 85608990-85608990 on Build GRCh38
rs771305086 Chr.2: 85608941-85608941 on Build GRCh38
rs771589402 Chr.2: 85609010-85609010 on Build GRCh38
rs1031034717 Chr.2: 85608808-85608808 on Build GRCh38
rs374124839 Chr.2: 85608973-85608973 on Build GRCh38
rs971980144 Chr.2: 85608862-85608862 on Build GRCh38
rs958749349 Chr.2: 85608953-85608953 on Build GRCh38
rs956697443 Chr.2: 85609035-85609035 on Build GRCh38
rs994825756 Chr.2: 85608835-85608835 on Build GRCh38
rs769397836 Chr.2: 85608942-85608942 on Build GRCh38
rs549296186 Chr.2: 85608812-85608812 on Build GRCh38
rs1006528025 Chr.2: 85608770-85608773 on Build GRCh38
rs376820834 Chr.2: 85608961-85608961 on Build GRCh38
rs768311573 Chr.2: 85609037-85609037 on Build GRCh38
rs561558122 Chr.2: 85608837-85608837 on Build GRCh38
rs563016828 Chr.2: 85608692-85608692 on Build GRCh38
rs140476145 Chr.2: 85609094-85609094 on Build GRCh38
rs781777007 Chr.2: 85609013-85609013 on Build GRCh38
rs751393826 Chr.2: 85608821-85608821 on Build GRCh38
rs986285621 Chr.2: 85609112-85609112 on Build GRCh38
rs775005531 Chr.2: 85609030-85609030 on Build GRCh38
rs540995399 Chr.2: 85608701-85608702 on Build GRCh38
rs758580710 Chr.2: 85609075-85609075 on Build GRCh38
rs371091330 Chr.2: 85608962-85608962 on Build GRCh38
rs775071406 Chr.2: 85608950-85608950 on Build GRCh38
rs755211478 Chr.2: 85608898-85608898 on Build GRCh38
rs767273605 Chr.2: 85609049-85609049 on Build GRCh38
rs775330307 Chr.2: 85609161-85609161 on Build GRCh38
rs746075456 Chr.2: 85609023-85609023 on Build GRCh38
rs752611997 Chr.2: 85608923-85608923 on Build GRCh38
rs746372773 Chr.2: 85609021-85609021 on Build GRCh38
rs555272397 Chr.2: 85609107-85609107 on Build GRCh38

View Full Product Details