Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85611799-85612293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf68
Entrez Gene ID
388969
Gene Name
chromosome 2 open reading frame 68
Gene Aliases
HCRCN81
UniGene
Hs.516159
Gene Chromosome Location
Chr.2: 85605253-85612073 on Build GRCh38
Transcripts
NM_001013649.3 XM_005264305.3 NM_001256727.1 NM_001256728.1 NR_046347.1 XM_006711922.1 XM_011532488.1

Gene Symbol
USP39
Entrez Gene ID
10713
Gene Name
ubiquitin specific peptidase 39
Gene Aliases
65K, CGI-21, HSPC332, SAD1, SNRNP65
UniGene
Hs.654745
Gene Chromosome Location
Chr.2: 85602842-85649284 on Build GRCh38
Transcripts
NM_001013649.3 XM_005264305.3 NM_001256727.1 NM_001256728.1 NR_046347.1 XM_006711922.1 XM_011532488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5862926 c.52C>T p.R18W Chr.2: 85611933-85611933 on Build GRCh38
6615896 c.107+10_107+14delCGCCC p.? Chr.2: 85611864-85611868 on Build GRCh38
5862925 c.52C>T p.R18W Chr.2: 85611933-85611933 on Build GRCh38
6615897 c.107+10_107+14delCGCCC p.? Chr.2: 85611864-85611868 on Build GRCh38
3972379 c.23G>C p.R8P Chr.2: 85611962-85611962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572426500 Chr.2: 85611949-85611949 on Build GRCh38
rs982462801 Chr.2: 85612107-85612107 on Build GRCh38
rs762845589 Chr.2: 85612017-85612017 on Build GRCh38
rs964210418 Chr.2: 85611872-85611872 on Build GRCh38
rs1002457894 Chr.2: 85612010-85612010 on Build GRCh38
rs377439765 Chr.2: 85611930-85611930 on Build GRCh38
rs545756346 Chr.2: 85611958-85611958 on Build GRCh38
rs895911509 Chr.2: 85612018-85612018 on Build GRCh38
rs746104852 Chr.2: 85611830-85611830 on Build GRCh38
rs1023689618 Chr.2: 85611988-85611988 on Build GRCh38
rs554124077 Chr.2: 85611831-85611831 on Build GRCh38
rs781160277 Chr.2: 85611884-85611884 on Build GRCh38
rs565365810 Chr.2: 85611843-85611844 on Build GRCh38
rs780091412 Chr.2: 85611953-85611953 on Build GRCh38
rs377584220 Chr.2: 85611852-85611852 on Build GRCh38
rs926148422 Chr.2: 85611855-85611855 on Build GRCh38
rs775718792 Chr.2: 85612015-85612018 on Build GRCh38
rs772684723 Chr.2: 85612254-85612254 on Build GRCh38
rs778460121 Chr.2: 85612034-85612034 on Build GRCh38
rs772594273 Chr.2: 85611910-85611910 on Build GRCh38
rs761675911 Chr.2: 85612024-85612024 on Build GRCh38
rs771492705 Chr.2: 85611850-85611850 on Build GRCh38
rs965870209 Chr.2: 85612245-85612245 on Build GRCh38
rs537111251 Chr.2: 85612021-85612021 on Build GRCh38
rs753911648 Chr.2: 85612027-85612027 on Build GRCh38
rs778468143 Chr.2: 85611979-85611979 on Build GRCh38
rs765077330 Chr.2: 85611871-85611871 on Build GRCh38
rs774783380 Chr.2: 85611844-85611844 on Build GRCh38
rs964179799 Chr.2: 85612065-85612065 on Build GRCh38
rs748604317 Chr.2: 85611909-85611909 on Build GRCh38
rs769032984 Chr.2: 85611842-85611842 on Build GRCh38
rs375189288 Chr.2: 85611915-85611915 on Build GRCh38
rs1020330973 Chr.2: 85612044-85612044 on Build GRCh38
rs756164113 Chr.2: 85611892-85611892 on Build GRCh38
rs766085466 Chr.2: 85611861-85611861 on Build GRCh38
rs766449299 Chr.2: 85612025-85612025 on Build GRCh38
rs752636664 Chr.2: 85611874-85611874 on Build GRCh38
rs751467406 Chr.2: 85611948-85611948 on Build GRCh38
rs752836689 Chr.2: 85612035-85612035 on Build GRCh38
rs957555181 Chr.2: 85612218-85612218 on Build GRCh38
rs200797297 Chr.2: 85611820-85611820 on Build GRCh38
rs1036568227 Chr.2: 85612141-85612141 on Build GRCh38
rs760438050 Chr.2: 85611860-85611860 on Build GRCh38
rs745512379 Chr.2: 85612001-85612001 on Build GRCh38
rs748090010 Chr.2: 85611824-85611824 on Build GRCh38
rs1020119807 Chr.2: 85612232-85612232 on Build GRCh38
rs1001300566 Chr.2: 85612201-85612201 on Build GRCh38
rs1000410980 Chr.2: 85612056-85612056 on Build GRCh38
rs749701835 Chr.2: 85611833-85611833 on Build GRCh38
rs753522677 Chr.2: 85611862-85611862 on Build GRCh38
rs1033270316 Chr.2: 85612057-85612057 on Build GRCh38
rs1032323804 Chr.2: 85612212-85612212 on Build GRCh38
rs762366488 Chr.2: 85611845-85611845 on Build GRCh38
rs759213616 Chr.2: 85611934-85611934 on Build GRCh38
rs543992256 Chr.2: 85611825-85611826 on Build GRCh38
rs781319913 Chr.2: 85611827-85611827 on Build GRCh38
rs779179161 Chr.2: 85611970-85611970 on Build GRCh38
rs955330394 Chr.2: 85612090-85612090 on Build GRCh38
rs776225801 Chr.2: 85611996-85611996 on Build GRCh38
rs547788644 Chr.2: 85612217-85612218 on Build GRCh38
rs770623023 Chr.2: 85611990-85611990 on Build GRCh38
rs770537097 Chr.2: 85611924-85611924 on Build GRCh38
rs941317759 Chr.2: 85611951-85611951 on Build GRCh38
rs750452061 Chr.2: 85611891-85611891 on Build GRCh38
rs759291669 Chr.2: 85611866-85611866 on Build GRCh38
rs770241399 Chr.2: 85612221-85612221 on Build GRCh38
rs758299105 Chr.2: 85611878-85611878 on Build GRCh38
rs758672486 Chr.2: 85611977-85611977 on Build GRCh38
rs967651714 Chr.2: 85611825-85611825 on Build GRCh38
rs921664110 Chr.2: 85612093-85612093 on Build GRCh38
rs913063347 Chr.2: 85612054-85612054 on Build GRCh38
rs370381748 Chr.2: 85611956-85611956 on Build GRCh38
rs926558776 Chr.2: 85612149-85612149 on Build GRCh38
rs896612607 Chr.2: 85611978-85611978 on Build GRCh38
rs560456073 Chr.2: 85612188-85612188 on Build GRCh38
rs1038166933 Chr.2: 85611971-85611971 on Build GRCh38
rs532822784 Chr.2: 85612108-85612109 on Build GRCh38
rs562692517 Chr.2: 85612081-85612081 on Build GRCh38
rs923723406 Chr.2: 85611875-85611875 on Build GRCh38
rs976005720 Chr.2: 85612066-85612066 on Build GRCh38
rs371001267 Chr.2: 85612223-85612223 on Build GRCh38
rs933649291 Chr.2: 85611918-85611918 on Build GRCh38
rs370522883 Chr.2: 85611898-85611898 on Build GRCh38
rs973930444 Chr.2: 85612137-85612137 on Build GRCh38
rs896665115 Chr.2: 85612146-85612147 on Build GRCh38
rs957294912 Chr.2: 85612040-85612040 on Build GRCh38
rs373989144 Chr.2: 85611832-85611832 on Build GRCh38
rs748680740 Chr.2: 85612177-85612177 on Build GRCh38
rs762819333 Chr.2: 85611943-85611943 on Build GRCh38
rs548389797 Chr.2: 85612161-85612161 on Build GRCh38
rs931656162 Chr.2: 85612103-85612103 on Build GRCh38
rs1029396723 Chr.2: 85612082-85612082 on Build GRCh38
rs908456742 Chr.2: 85611941-85611941 on Build GRCh38
rs937839895 Chr.2: 85612154-85612154 on Build GRCh38
rs772231715 Chr.2: 85611991-85611992 on Build GRCh38
rs543596651 Chr.2: 85612055-85612055 on Build GRCh38
rs904998304 Chr.2: 85612008-85612008 on Build GRCh38
rs908258424 Chr.2: 85612112-85612112 on Build GRCh38
rs780294799 Chr.2: 85611893-85611893 on Build GRCh38
rs895892392 Chr.2: 85612176-85612176 on Build GRCh38
rs905246829 Chr.2: 85612190-85612190 on Build GRCh38
rs752471581 Chr.2: 85611942-85611942 on Build GRCh38
rs748499030 Chr.2: 85611976-85611976 on Build GRCh38
rs749789284 Chr.2: 85611826-85611830 on Build GRCh38
rs941036599 Chr.2: 85612120-85612120 on Build GRCh38
rs779180669 Chr.2: 85611848-85611849 on Build GRCh38
rs564172040 Chr.2: 85612011-85612011 on Build GRCh38
rs749503791 Chr.2: 85611897-85611897 on Build GRCh38
rs988625945 Chr.2: 85612051-85612051 on Build GRCh38
rs764065300 Chr.2: 85611946-85611946 on Build GRCh38
rs747515268 Chr.2: 85611985-85611985 on Build GRCh38
rs989152011 Chr.2: 85612187-85612187 on Build GRCh38
rs760315190 Chr.2: 85611923-85611923 on Build GRCh38
rs529805914 Chr.2: 85612146-85612146 on Build GRCh38
rs746243009 Chr.2: 85611959-85611988 on Build GRCh38
rs764957672 Chr.2: 85611939-85611939 on Build GRCh38
rs576104461 Chr.2: 85612012-85612012 on Build GRCh38
rs1000211109 Chr.2: 85612206-85612206 on Build GRCh38

View Full Product Details