Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85695819-85696361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNLY
Entrez Gene ID
10578
Gene Name
granulysin
Gene Aliases
D2S69E, LAG-2, LAG2, NKG5, TLA519
UniGene
Hs.105806
Gene Chromosome Location
Chr.2: 85694291-85698854 on Build GRCh38
Transcripts
NM_001302758.1 NM_006433.4 NM_012483.3 XM_005264084.2 XM_005264085.2 XM_005264087.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1483319 c.200G>T p.R67M Chr.2: 85696001-85696001 on Build GRCh38
5801972 c.171delA p.I57fs*5 Chr.2: 85696017-85696017 on Build GRCh38
6159020 c.219C>G p.V73V Chr.2: 85696020-85696020 on Build GRCh38
4536318 c.183G>A p.L61L Chr.2: 85696029-85696029 on Build GRCh38
6159021 c.174C>G p.V58V Chr.2: 85696020-85696020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051308077 Chr.2: 85696136-85696136 on Build GRCh38
rs757879664 Chr.2: 85696016-85696016 on Build GRCh38
rs948595774 Chr.2: 85696124-85696124 on Build GRCh38
rs116332076 Chr.2: 85695882-85695882 on Build GRCh38
rs924068284 Chr.2: 85696130-85696130 on Build GRCh38
rs143603515 Chr.2: 85696014-85696014 on Build GRCh38
rs1043989785 Chr.2: 85696191-85696191 on Build GRCh38
rs746623967 Chr.2: 85695935-85695935 on Build GRCh38
rs769554534 Chr.2: 85696252-85696252 on Build GRCh38
rs902004306 Chr.2: 85696292-85696292 on Build GRCh38
rs141685032 Chr.2: 85696052-85696052 on Build GRCh38
rs759843973 Chr.2: 85696079-85696079 on Build GRCh38
rs773452688 Chr.2: 85695992-85695992 on Build GRCh38
rs577851040 Chr.2: 85695959-85695959 on Build GRCh38
rs149567569 Chr.2: 85696105-85696105 on Build GRCh38
rs548624046 Chr.2: 85696156-85696156 on Build GRCh38
rs552917326 Chr.2: 85695919-85695919 on Build GRCh38
rs985936173 Chr.2: 85695876-85695876 on Build GRCh38
rs776856779 Chr.2: 85696078-85696078 on Build GRCh38
rs1007744253 Chr.2: 85696228-85696228 on Build GRCh38
rs187039953 Chr.2: 85696064-85696064 on Build GRCh38
rs771146670 Chr.2: 85696070-85696070 on Build GRCh38
rs566986921 Chr.2: 85695895-85695895 on Build GRCh38
rs61731721 Chr.2: 85696046-85696046 on Build GRCh38
rs751088830 Chr.2: 85696034-85696034 on Build GRCh38
rs569201261 Chr.2: 85695865-85695865 on Build GRCh38
rs761811741 Chr.2: 85695966-85695966 on Build GRCh38
rs968095355 Chr.2: 85696119-85696119 on Build GRCh38
rs764681570 Chr.2: 85696102-85696102 on Build GRCh38
rs372926915 Chr.2: 85695909-85695909 on Build GRCh38
rs953660988 Chr.2: 85695867-85695867 on Build GRCh38
rs755303248 Chr.2: 85696013-85696013 on Build GRCh38
rs1008105427 Chr.2: 85696051-85696051 on Build GRCh38
rs906984474 Chr.2: 85696291-85696291 on Build GRCh38
rs75718542 Chr.2: 85696186-85696186 on Build GRCh38
rs144103676 Chr.2: 85696131-85696131 on Build GRCh38
rs556915721 Chr.2: 85695999-85695999 on Build GRCh38
rs371187535 Chr.2: 85696103-85696103 on Build GRCh38
rs993836809 Chr.2: 85696247-85696252 on Build GRCh38
rs915328165 Chr.2: 85695915-85695915 on Build GRCh38
rs770673724 Chr.2: 85695939-85695939 on Build GRCh38
rs777594532 Chr.2: 85695932-85695932 on Build GRCh38
rs376808749 Chr.2: 85696093-85696093 on Build GRCh38
rs866114045 Chr.2: 85696147-85696147 on Build GRCh38
rs138334587 Chr.2: 85696010-85696010 on Build GRCh38
rs1022487925 Chr.2: 85695852-85695852 on Build GRCh38
rs757025306 Chr.2: 85696035-85696035 on Build GRCh38
rs769572875 Chr.2: 85696045-85696045 on Build GRCh38
rs571397536 Chr.2: 85696324-85696324 on Build GRCh38
rs780984153 Chr.2: 85696042-85696042 on Build GRCh38
rs946808938 Chr.2: 85695929-85695929 on Build GRCh38
rs939469282 Chr.2: 85696150-85696150 on Build GRCh38
rs779471141 Chr.2: 85695913-85695913 on Build GRCh38
rs776615109 Chr.2: 85695940-85695940 on Build GRCh38
rs772444111 Chr.2: 85696123-85696123 on Build GRCh38
rs189664077 Chr.2: 85696006-85696006 on Build GRCh38
rs1051786565 Chr.2: 85695982-85695982 on Build GRCh38
rs752975937 Chr.2: 85696090-85696090 on Build GRCh38
rs760741630 Chr.2: 85696068-85696068 on Build GRCh38
rs41290043 Chr.2: 85696058-85696058 on Build GRCh38
rs534320800 Chr.2: 85696185-85696185 on Build GRCh38
rs766576647 Chr.2: 85696007-85696007 on Build GRCh38
rs572625963 Chr.2: 85696043-85696043 on Build GRCh38
rs35146240 Chr.2: 85695991-85695991 on Build GRCh38
rs528473235 Chr.2: 85696110-85696110 on Build GRCh38
rs377237054 Chr.2: 85695931-85695931 on Build GRCh38
rs919193607 Chr.2: 85695907-85695907 on Build GRCh38
rs889338443 Chr.2: 85696200-85696200 on Build GRCh38
rs369766430 Chr.2: 85695963-85695963 on Build GRCh38
rs566946349 Chr.2: 85696168-85696168 on Build GRCh38
rs942993508 Chr.2: 85696166-85696166 on Build GRCh38
rs1042566586 Chr.2: 85695934-85695934 on Build GRCh38
rs773674970 Chr.2: 85696140-85696140 on Build GRCh38
rs368762529 Chr.2: 85695908-85695908 on Build GRCh38
rs115691465 Chr.2: 85695892-85695892 on Build GRCh38
rs1003520169 Chr.2: 85695972-85695972 on Build GRCh38
rs532676766 Chr.2: 85696264-85696264 on Build GRCh38
rs777469496 Chr.2: 85696032-85696032 on Build GRCh38
rs981781292 Chr.2: 85695884-85695884 on Build GRCh38
rs991212290 Chr.2: 85696107-85696107 on Build GRCh38

View Full Product Details