Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86441781-86442285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3714315 c.30G>C p.P10P Chr.2: 86442077-86442077 on Build GRCh38
1307064 c.70G>A p.D24N Chr.2: 86442117-86442117 on Build GRCh38
7161975 c.52C>T p.L18F Chr.2: 86442099-86442099 on Build GRCh38
359478 c.44G>T p.R15M Chr.2: 86442091-86442091 on Build GRCh38
1615186 c.99G>A p.W33* Chr.2: 86442146-86442146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944973213 Chr.2: 86441926-86441926 on Build GRCh38
rs768316363 Chr.2: 86442025-86442025 on Build GRCh38
rs755641599 Chr.2: 86442058-86442058 on Build GRCh38
rs1026902777 Chr.2: 86441871-86441871 on Build GRCh38
rs367622060 Chr.2: 86442031-86442031 on Build GRCh38
rs780124918 Chr.2: 86442036-86442036 on Build GRCh38
rs768867283 Chr.2: 86442091-86442091 on Build GRCh38
rs912359124 Chr.2: 86441881-86441881 on Build GRCh38
rs527564102 Chr.2: 86442083-86442083 on Build GRCh38
rs757148088 Chr.2: 86442243-86442243 on Build GRCh38
rs774584530 Chr.2: 86442173-86442173 on Build GRCh38
rs758228009 Chr.2: 86442219-86442219 on Build GRCh38
rs931386198 Chr.2: 86441816-86441816 on Build GRCh38
rs932418625 Chr.2: 86441907-86441907 on Build GRCh38
rs894628238 Chr.2: 86442224-86442224 on Build GRCh38
rs779816862 Chr.2: 86442016-86442016 on Build GRCh38
rs146747408 Chr.2: 86442071-86442071 on Build GRCh38
rs776006202 Chr.2: 86442189-86442189 on Build GRCh38
rs550283706 Chr.2: 86441828-86441828 on Build GRCh38
rs917242244 Chr.2: 86442211-86442211 on Build GRCh38
rs898002020 Chr.2: 86441939-86441939 on Build GRCh38
rs778556993 Chr.2: 86442062-86442062 on Build GRCh38
rs994930859 Chr.2: 86441958-86441958 on Build GRCh38
rs376742455 Chr.2: 86442116-86442116 on Build GRCh38
rs751299695 Chr.2: 86442238-86442238 on Build GRCh38
rs768071670 Chr.2: 86442048-86442048 on Build GRCh38
rs951988563 Chr.2: 86442165-86442165 on Build GRCh38
rs745968685 Chr.2: 86442080-86442080 on Build GRCh38
rs780837292 Chr.2: 86441997-86441997 on Build GRCh38
rs948919188 Chr.2: 86441896-86441896 on Build GRCh38
rs753404969 Chr.2: 86442060-86442060 on Build GRCh38
rs771998711 Chr.2: 86442076-86442076 on Build GRCh38
rs774584950 Chr.2: 86442093-86442093 on Build GRCh38
rs202106084 Chr.2: 86442164-86442164 on Build GRCh38
rs771054383 Chr.2: 86442030-86442030 on Build GRCh38
rs531970958 Chr.2: 86441809-86441809 on Build GRCh38
rs562131913 Chr.2: 86441929-86441929 on Build GRCh38
rs1000348074 Chr.2: 86441936-86441936 on Build GRCh38
rs763422784 Chr.2: 86442035-86442035 on Build GRCh38
rs759043440 Chr.2: 86442196-86442196 on Build GRCh38
rs762240995 Chr.2: 86442047-86442047 on Build GRCh38
rs766849470 Chr.2: 86442188-86442188 on Build GRCh38
rs1027813748 Chr.2: 86441964-86441964 on Build GRCh38
rs1008588478 Chr.2: 86442086-86442086 on Build GRCh38
rs753582424 Chr.2: 86442120-86442120 on Build GRCh38
rs1008537537 Chr.2: 86441970-86441970 on Build GRCh38
rs771705903 Chr.2: 86442239-86442240 on Build GRCh38
rs748576324 Chr.2: 86442179-86442179 on Build GRCh38
rs921379482 Chr.2: 86441815-86441815 on Build GRCh38
rs371719009 Chr.2: 86442033-86442033 on Build GRCh38
rs749459833 Chr.2: 86442089-86442089 on Build GRCh38
rs781722463 Chr.2: 86442202-86442202 on Build GRCh38
rs201831015 Chr.2: 86442185-86442185 on Build GRCh38
rs749133826 Chr.2: 86442022-86442022 on Build GRCh38
rs755067815 Chr.2: 86442256-86442256 on Build GRCh38
rs775819688 Chr.2: 86442085-86442085 on Build GRCh38
rs1051228260 Chr.2: 86441914-86441914 on Build GRCh38
rs1044940336 Chr.2: 86441920-86441920 on Build GRCh38
rs751759466 Chr.2: 86442141-86442141 on Build GRCh38
rs760305076 Chr.2: 86442117-86442117 on Build GRCh38
rs369307421 Chr.2: 86442184-86442184 on Build GRCh38
rs778952114 Chr.2: 86442027-86442027 on Build GRCh38
rs1036551109 Chr.2: 86441930-86441930 on Build GRCh38
rs984173347 Chr.2: 86441821-86441821 on Build GRCh38
rs752461053 Chr.2: 86442138-86442138 on Build GRCh38
rs896043584 Chr.2: 86441895-86441895 on Build GRCh38
rs758140046 Chr.2: 86442139-86442139 on Build GRCh38
rs547724608 Chr.2: 86441981-86441981 on Build GRCh38
rs754597618 Chr.2: 86442122-86442122 on Build GRCh38
rs921048468 Chr.2: 86441906-86441906 on Build GRCh38
rs770034100 Chr.2: 86442032-86442032 on Build GRCh38
rs953887080 Chr.2: 86441875-86441875 on Build GRCh38
rs749448727 Chr.2: 86442116-86442124 on Build GRCh38
rs764816560 Chr.2: 86442209-86442209 on Build GRCh38
rs756251886 Chr.2: 86442158-86442158 on Build GRCh38
rs768165732 Chr.2: 86442105-86442105 on Build GRCh38
rs985322047 Chr.2: 86441878-86441878 on Build GRCh38
rs936146129 Chr.2: 86442075-86442075 on Build GRCh38
rs764874323 Chr.2: 86442123-86442123 on Build GRCh38
rs908544414 Chr.2: 86441825-86441825 on Build GRCh38
rs939651275 Chr.2: 86442133-86442133 on Build GRCh38
rs777736062 Chr.2: 86442147-86442147 on Build GRCh38
rs1050241898 Chr.2: 86441854-86441863 on Build GRCh38
rs1015264894 Chr.2: 86441857-86441857 on Build GRCh38
rs1013429261 Chr.2: 86442263-86442263 on Build GRCh38
rs778182326 Chr.2: 86442030-86442030 on Build GRCh38
rs1012982881 Chr.2: 86441987-86441987 on Build GRCh38
rs373486115 Chr.2: 86442213-86442213 on Build GRCh38
rs376191355 Chr.2: 86442244-86442244 on Build GRCh38
rs529431175 Chr.2: 86441971-86441971 on Build GRCh38
rs140350293 Chr.2: 86442171-86442171 on Build GRCh38
rs746917369 Chr.2: 86442156-86442156 on Build GRCh38
rs780013893 Chr.2: 86442163-86442163 on Build GRCh38
rs372709230 Chr.2: 86442002-86442002 on Build GRCh38
rs995458866 Chr.2: 86441867-86441867 on Build GRCh38
rs1031386854 Chr.2: 86441949-86441949 on Build GRCh38
rs752168850 Chr.2: 86442045-86442045 on Build GRCh38
rs772538293 Chr.2: 86442182-86442182 on Build GRCh38
rs749349413 Chr.2: 86442254-86442254 on Build GRCh38
rs764332809 Chr.2: 86442038-86442038 on Build GRCh38
rs762229054 Chr.2: 86442099-86442099 on Build GRCh38
rs755178415 Chr.2: 86442181-86442181 on Build GRCh38
rs763870735 Chr.2: 86442225-86442225 on Build GRCh38
rs894591000 Chr.2: 86441978-86441978 on Build GRCh38
rs1003012442 Chr.2: 86441853-86441853 on Build GRCh38
rs765804308 Chr.2: 86442119-86442119 on Build GRCh38
rs1057335344 Chr.2: 86441883-86441883 on Build GRCh38
rs570618403 Chr.2: 86442187-86442187 on Build GRCh38
rs755833831 Chr.2: 86442006-86442006 on Build GRCh38
rs964603108 Chr.2: 86441972-86441972 on Build GRCh38
rs890784246 Chr.2: 86441922-86441922 on Build GRCh38
rs749984494 Chr.2: 86442053-86442053 on Build GRCh38
rs961000011 Chr.2: 86441860-86441860 on Build GRCh38
rs565952376 Chr.2: 86442028-86442028 on Build GRCh38

View Full Product Details