Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88557706-88558238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK3
Entrez Gene ID
9451
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
PEK, PERK, WRS
UniGene
Hs.591589
Gene Chromosome Location
Chr.2: 88556740-88627576 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Gene Symbol
LOC101928371
Entrez Gene ID
101928371
Gene Name
uncharacterized LOC101928371
Gene Aliases
-
UniGene
Hs.738045
Gene Chromosome Location
Chr.2: 88538720-88575610 on Build GRCh38
Transcripts
NM_001313915.1 NM_004836.6 XM_017005376.1 NR_110236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096190 c.3178C>A p.P1060T Chr.2: 88557909-88557909 on Build GRCh38
1483340 c.3264C>T p.L1088L Chr.2: 88557823-88557823 on Build GRCh38
1647361 c.3265A>G p.R1089G Chr.2: 88557822-88557822 on Build GRCh38
5494401 c.3205A>G p.I1069V Chr.2: 88557882-88557882 on Build GRCh38
79355 c.3193C>T p.R1065* Chr.2: 88557894-88557894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377370017 Chr.2: 88558052-88558052 on Build GRCh38
rs1040115805 Chr.2: 88558022-88558022 on Build GRCh38
rs1040042580 Chr.2: 88557743-88557743 on Build GRCh38
rs201004765 Chr.2: 88557949-88557949 on Build GRCh38
rs747073375 Chr.2: 88557770-88557770 on Build GRCh38
rs781096880 Chr.2: 88557986-88557990 on Build GRCh38
rs370635736 Chr.2: 88558120-88558120 on Build GRCh38
rs181248985 Chr.2: 88557979-88557979 on Build GRCh38
rs1020057863 Chr.2: 88558151-88558151 on Build GRCh38
rs907647455 Chr.2: 88557986-88557986 on Build GRCh38
rs757415634 Chr.2: 88557778-88557778 on Build GRCh38
rs10208681 Chr.2: 88557796-88557796 on Build GRCh38
rs572234262 Chr.2: 88557793-88557793 on Build GRCh38
rs1055169296 Chr.2: 88558108-88558108 on Build GRCh38
rs751877330 Chr.2: 88557902-88557902 on Build GRCh38
rs944423941 Chr.2: 88557996-88557996 on Build GRCh38
rs771437394 Chr.2: 88557980-88557980 on Build GRCh38
rs1032902320 Chr.2: 88557761-88557761 on Build GRCh38
rs370657508 Chr.2: 88557868-88557868 on Build GRCh38
rs752870446 Chr.2: 88557737-88557737 on Build GRCh38
rs995819504 Chr.2: 88558055-88558055 on Build GRCh38
rs765878566 Chr.2: 88557809-88557809 on Build GRCh38
rs560583783 Chr.2: 88558091-88558091 on Build GRCh38
rs769976164 Chr.2: 88558035-88558035 on Build GRCh38
rs372724944 Chr.2: 88557972-88557972 on Build GRCh38
rs757254462 Chr.2: 88557920-88557920 on Build GRCh38
rs780507890 Chr.2: 88557944-88557944 on Build GRCh38
rs773099851 Chr.2: 88557830-88557830 on Build GRCh38
rs764288224 Chr.2: 88557899-88557899 on Build GRCh38
rs747041290 Chr.2: 88557998-88557999 on Build GRCh38
rs754991725 Chr.2: 88557947-88557947 on Build GRCh38
rs376257254 Chr.2: 88557807-88557807 on Build GRCh38
rs1010525939 Chr.2: 88558137-88558137 on Build GRCh38
rs530253795 Chr.2: 88558042-88558042 on Build GRCh38
rs773003685 Chr.2: 88557815-88557815 on Build GRCh38
rs768435755 Chr.2: 88557804-88557804 on Build GRCh38
rs117769655 Chr.2: 88558109-88558109 on Build GRCh38
rs950504073 Chr.2: 88558005-88558005 on Build GRCh38
rs1034373472 Chr.2: 88558176-88558176 on Build GRCh38
rs535059276 Chr.2: 88557730-88557730 on Build GRCh38
rs957438194 Chr.2: 88557763-88557763 on Build GRCh38
rs779142799 Chr.2: 88557816-88557819 on Build GRCh38
rs751487624 Chr.2: 88557848-88557848 on Build GRCh38
rs190054983 Chr.2: 88557968-88557968 on Build GRCh38
rs762080456 Chr.2: 88557800-88557800 on Build GRCh38
rs765299033 Chr.2: 88557810-88557810 on Build GRCh38
rs1002927183 Chr.2: 88558169-88558169 on Build GRCh38
rs778395797 Chr.2: 88557977-88557977 on Build GRCh38
rs746319005 Chr.2: 88557933-88557933 on Build GRCh38
rs772812948 Chr.2: 88557985-88557985 on Build GRCh38
rs770232594 Chr.2: 88557794-88557794 on Build GRCh38
rs56120877 Chr.2: 88557934-88557934 on Build GRCh38
rs902654397 Chr.2: 88557734-88557734 on Build GRCh38
rs530708954 Chr.2: 88557808-88557808 on Build GRCh38
rs1010039552 Chr.2: 88557787-88557787 on Build GRCh38
rs879187024 Chr.2: 88557731-88557731 on Build GRCh38
rs946976281 Chr.2: 88558155-88558155 on Build GRCh38
rs187164681 Chr.2: 88558162-88558162 on Build GRCh38
rs755367718 Chr.2: 88558095-88558095 on Build GRCh38
rs776812138 Chr.2: 88557746-88557746 on Build GRCh38
rs766214245 Chr.2: 88557735-88557735 on Build GRCh38
rs775427890 Chr.2: 88558044-88558044 on Build GRCh38
rs928096682 Chr.2: 88558061-88558061 on Build GRCh38
rs767092678 Chr.2: 88557814-88557814 on Build GRCh38
rs141166027 Chr.2: 88557930-88557930 on Build GRCh38
rs893047719 Chr.2: 88558103-88558103 on Build GRCh38
rs145982613 Chr.2: 88557757-88557757 on Build GRCh38
rs766126107 Chr.2: 88557827-88557827 on Build GRCh38
rs143931342 Chr.2: 88557824-88557824 on Build GRCh38
rs866814797 Chr.2: 88557894-88557894 on Build GRCh38

View Full Product Details