Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96761600-96762096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
UniGene
Hs.175043
Gene Chromosome Location
Chr.2: 96760902-96811891 on Build GRCh38
Transcripts
NM_020184.3 XM_005263914.3 XM_005263915.3 XM_011510955.2 XM_011510956.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4624891 c.697G>A p.E233K Chr.2: 96761696-96761696 on Build GRCh38
3799289 c.808A>T p.M270L Chr.2: 96761807-96761807 on Build GRCh38
1293999 c.1088A>T p.Q363L Chr.2: 96762087-96762087 on Build GRCh38
4825971 c.862C>T p.Q288* Chr.2: 96761861-96761861 on Build GRCh38
5702994 c.812C>T p.A271V Chr.2: 96761811-96761811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370137332 Chr.2: 96761758-96761758 on Build GRCh38
rs779146988 Chr.2: 96761688-96761689 on Build GRCh38
rs56246512 Chr.2: 96761783-96761783 on Build GRCh38
rs1050712428 Chr.2: 96761620-96761620 on Build GRCh38
rs999284391 Chr.2: 96761848-96761848 on Build GRCh38
rs970585969 Chr.2: 96762047-96762047 on Build GRCh38
rs749703825 Chr.2: 96761762-96761762 on Build GRCh38
rs146923909 Chr.2: 96761642-96761642 on Build GRCh38
rs762759439 Chr.2: 96762075-96762075 on Build GRCh38
rs201014887 Chr.2: 96761977-96761977 on Build GRCh38
rs771213657 Chr.2: 96762016-96762016 on Build GRCh38
rs145147485 Chr.2: 96762020-96762020 on Build GRCh38
rs757875854 Chr.2: 96761869-96761869 on Build GRCh38
rs74552543 Chr.2: 96761970-96761970 on Build GRCh38
rs773120249 Chr.2: 96762001-96762001 on Build GRCh38
rs559372337 Chr.2: 96761809-96761809 on Build GRCh38
rs780881771 Chr.2: 96761938-96761938 on Build GRCh38
rs752693477 Chr.2: 96761866-96761866 on Build GRCh38
rs182308036 Chr.2: 96761701-96761701 on Build GRCh38
rs760464330 Chr.2: 96761688-96761688 on Build GRCh38
rs200939351 Chr.2: 96761639-96761639 on Build GRCh38
rs749650885 Chr.2: 96761941-96761941 on Build GRCh38
rs373657140 Chr.2: 96761926-96761926 on Build GRCh38
rs746662702 Chr.2: 96761749-96761749 on Build GRCh38
rs35310683 Chr.2: 96761805-96761806 on Build GRCh38
rs903670676 Chr.2: 96761845-96761845 on Build GRCh38
rs763543912 Chr.2: 96761891-96761891 on Build GRCh38
rs760772919 Chr.2: 96761802-96761802 on Build GRCh38
rs786205530 Chr.2: 96761733-96761733 on Build GRCh38
rs139582999 Chr.2: 96761962-96761962 on Build GRCh38
rs1014216906 Chr.2: 96761996-96761996 on Build GRCh38
rs765198380 Chr.2: 96761842-96761842 on Build GRCh38
rs367842804 Chr.2: 96761966-96761966 on Build GRCh38
rs754940734 Chr.2: 96761725-96761725 on Build GRCh38
rs774907198 Chr.2: 96761773-96761773 on Build GRCh38
rs750618555 Chr.2: 96761680-96761680 on Build GRCh38
rs775538413 Chr.2: 96762070-96762070 on Build GRCh38
rs751120696 Chr.2: 96761919-96761919 on Build GRCh38
rs770067674 Chr.2: 96761623-96761623 on Build GRCh38
rs769192111 Chr.2: 96761656-96761656 on Build GRCh38
rs146068349 Chr.2: 96762011-96762011 on Build GRCh38
rs763749179 Chr.2: 96761771-96761771 on Build GRCh38
rs776551771 Chr.2: 96761828-96761828 on Build GRCh38
rs773439754 Chr.2: 96761795-96761795 on Build GRCh38
rs765321091 Chr.2: 96761734-96761734 on Build GRCh38
rs766669677 Chr.2: 96761812-96761812 on Build GRCh38
rs1043975317 Chr.2: 96761801-96761801 on Build GRCh38
rs977087685 Chr.2: 96761776-96761776 on Build GRCh38
rs759468747 Chr.2: 96761833-96761833 on Build GRCh38
rs150269409 Chr.2: 96761787-96761787 on Build GRCh38
rs759393159 Chr.2: 96762051-96762051 on Build GRCh38
rs75267011 Chr.2: 96761706-96761706 on Build GRCh38
rs929526785 Chr.2: 96762061-96762061 on Build GRCh38
rs763483009 Chr.2: 96761653-96761653 on Build GRCh38
rs762874791 Chr.2: 96761711-96761711 on Build GRCh38
rs761941296 Chr.2: 96761671-96761671 on Build GRCh38
rs777425652 Chr.2: 96761746-96761746 on Build GRCh38
rs772062538 Chr.2: 96761794-96761794 on Build GRCh38
rs187371920 Chr.2: 96762058-96762058 on Build GRCh38
rs967031665 Chr.2: 96761743-96761743 on Build GRCh38
rs755344155 Chr.2: 96761948-96761948 on Build GRCh38
rs935793378 Chr.2: 96761796-96761796 on Build GRCh38
rs377727962 Chr.2: 96761755-96761755 on Build GRCh38
rs752249547 Chr.2: 96761740-96761740 on Build GRCh38
rs757849001 Chr.2: 96761744-96761744 on Build GRCh38
rs147976284 Chr.2: 96761687-96761687 on Build GRCh38
rs2680925 Chr.2: 96761745-96761745 on Build GRCh38

View Full Product Details