Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96875800-96876297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM178B
Entrez Gene ID
51252
Gene Name
family with sequence similarity 178 member B
Gene Aliases
-
UniGene
Hs.107922
Gene Chromosome Location
Chr.2: 96875882-96986600 on Build GRCh38
Transcripts
NM_001122646.2 NM_001172667.1 NM_016490.4 XM_011511294.2 XM_017004263.1 XM_017004264.1 XM_017004266.1 XM_017004267.1 XM_017004268.1

SNP Detail

SNP ID Chromosome Location
rs946537410 Chr.2: 96875955-96875955 on Build GRCh38
rs141220264 Chr.2: 96875865-96875865 on Build GRCh38
rs531045466 Chr.2: 96876181-96876181 on Build GRCh38
rs779594067 Chr.2: 96876129-96876129 on Build GRCh38
rs898700524 Chr.2: 96876064-96876064 on Build GRCh38
rs911282758 Chr.2: 96876042-96876042 on Build GRCh38
rs867314389 Chr.2: 96875964-96875964 on Build GRCh38
rs528984776 Chr.2: 96876206-96876206 on Build GRCh38
rs577955421 Chr.2: 96876023-96876023 on Build GRCh38
rs925303318 Chr.2: 96876086-96876086 on Build GRCh38
rs397872699 Chr.2: 96875958-96875959 on Build GRCh38
rs565215732 Chr.2: 96876142-96876142 on Build GRCh38
rs372668986 Chr.2: 96876128-96876128 on Build GRCh38
rs374120633 Chr.2: 96876229-96876229 on Build GRCh38
rs750373555 Chr.2: 96875911-96875911 on Build GRCh38
rs528469437 Chr.2: 96876114-96876114 on Build GRCh38
rs559708693 Chr.2: 96876113-96876113 on Build GRCh38
rs879332781 Chr.2: 96875867-96875867 on Build GRCh38
rs1023302091 Chr.2: 96875903-96875903 on Build GRCh38
rs968577780 Chr.2: 96876258-96876258 on Build GRCh38
rs34783494 Chr.2: 96875955-96875956 on Build GRCh38
rs567728841 Chr.2: 96876201-96876201 on Build GRCh38
rs151082865 Chr.2: 96875948-96875951 on Build GRCh38
rs558072230 Chr.2: 96875912-96875912 on Build GRCh38
rs954626774 Chr.2: 96876210-96876210 on Build GRCh38
rs1013089258 Chr.2: 96876070-96876071 on Build GRCh38
rs188095156 Chr.2: 96875843-96875843 on Build GRCh38
rs1015868263 Chr.2: 96875972-96875972 on Build GRCh38
rs962055156 Chr.2: 96875985-96875985 on Build GRCh38
rs985982582 Chr.2: 96876036-96876036 on Build GRCh38
rs372145532 Chr.2: 96875950-96875953 on Build GRCh38
rs778830304 Chr.2: 96875848-96875848 on Build GRCh38
rs190516844 Chr.2: 96876091-96876091 on Build GRCh38
rs1039628987 Chr.2: 96875956-96875956 on Build GRCh38
rs1023099123 Chr.2: 96876075-96876075 on Build GRCh38
rs933891287 Chr.2: 96875989-96875989 on Build GRCh38
rs779568762 Chr.2: 96876254-96876254 on Build GRCh38
rs376012535 Chr.2: 96876266-96876266 on Build GRCh38
rs550777963 Chr.2: 96876194-96876194 on Build GRCh38
rs903883373 Chr.2: 96875841-96875841 on Build GRCh38
rs1043761243 Chr.2: 96875830-96875830 on Build GRCh38
rs746681130 Chr.2: 96876198-96876198 on Build GRCh38
rs574103352 Chr.2: 96876069-96876069 on Build GRCh38
rs563663735 Chr.2: 96876045-96876045 on Build GRCh38
rs762032421 Chr.2: 96876061-96876061 on Build GRCh38
rs755471840 Chr.2: 96876243-96876243 on Build GRCh38
rs115638608 Chr.2: 96876099-96876099 on Build GRCh38
rs370548325 Chr.2: 96876273-96876273 on Build GRCh38
rs534925831 Chr.2: 96875877-96875877 on Build GRCh38
rs1051217818 Chr.2: 96876010-96876012 on Build GRCh38
rs187344562 Chr.2: 96876044-96876044 on Build GRCh38
rs200020272 Chr.2: 96875947-96875949 on Build GRCh38
rs914649606 Chr.2: 96876169-96876169 on Build GRCh38
rs933920116 Chr.2: 96876247-96876247 on Build GRCh38
rs896731669 Chr.2: 96876108-96876108 on Build GRCh38
rs879121128 Chr.2: 96876122-96876122 on Build GRCh38
rs757908136 Chr.2: 96876090-96876090 on Build GRCh38
rs1023215526 Chr.2: 96875832-96875832 on Build GRCh38
rs954529992 Chr.2: 96876033-96876033 on Build GRCh38
rs971999135 Chr.2: 96876006-96876006 on Build GRCh38
rs1053153871 Chr.2: 96875845-96875845 on Build GRCh38
rs181051296 Chr.2: 96875864-96875864 on Build GRCh38
rs1043859128 Chr.2: 96876047-96876047 on Build GRCh38
rs1032108493 Chr.2: 96876126-96876126 on Build GRCh38
rs1005924937 Chr.2: 96876175-96876175 on Build GRCh38
rs542710137 Chr.2: 96876088-96876088 on Build GRCh38
rs569718482 Chr.2: 96875833-96875833 on Build GRCh38
rs1030227128 Chr.2: 96876202-96876202 on Build GRCh38
rs766806605 Chr.2: 96876228-96876228 on Build GRCh38
rs923189581 Chr.2: 96876031-96876031 on Build GRCh38

View Full Product Details