Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96876194-96876699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM178B
Entrez Gene ID
51252
Gene Name
family with sequence similarity 178 member B
Gene Aliases
-
UniGene
Hs.107922
Gene Chromosome Location
Chr.2: 96875882-96986600 on Build GRCh38
Transcripts
NM_001122646.2 NM_001172667.1 NM_016490.4 XM_011511294.2 XM_017004263.1 XM_017004264.1 XM_017004266.1 XM_017004267.1 XM_017004268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6995378 c.2462delT p.F821fs*>7 Chr.2: 96876298-96876298 on Build GRCh38
5476946 c.328-7G>A p.? Chr.2: 96876315-96876315 on Build GRCh38
6269180 c.2452-10C>A p.? Chr.2: 96876318-96876318 on Build GRCh38
6995377 c.338delT p.F113fs*>7 Chr.2: 96876298-96876298 on Build GRCh38
5476947 c.2452-7G>A p.? Chr.2: 96876315-96876315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543151045 Chr.2: 96876426-96876426 on Build GRCh38
rs368492559 Chr.2: 96876327-96876327 on Build GRCh38
rs150741478 Chr.2: 96876338-96876338 on Build GRCh38
rs557125216 Chr.2: 96876395-96876395 on Build GRCh38
rs886806398 Chr.2: 96876303-96876303 on Build GRCh38
rs78960893 Chr.2: 96876385-96876385 on Build GRCh38
rs539874440 Chr.2: 96876361-96876361 on Build GRCh38
rs35886699 Chr.2: 96876485-96876485 on Build GRCh38
rs573715963 Chr.2: 96876406-96876406 on Build GRCh38
rs775416688 Chr.2: 96876304-96876304 on Build GRCh38
rs1051562883 Chr.2: 96876645-96876645 on Build GRCh38
rs892048788 Chr.2: 96876517-96876517 on Build GRCh38
rs749672041 Chr.2: 96876292-96876292 on Build GRCh38
rs1044595475 Chr.2: 96876340-96876340 on Build GRCh38
rs746843636 Chr.2: 96876550-96876550 on Build GRCh38
rs760694499 Chr.2: 96876605-96876605 on Build GRCh38
rs374120633 Chr.2: 96876229-96876229 on Build GRCh38
rs753995784 Chr.2: 96876671-96876671 on Build GRCh38
rs745677858 Chr.2: 96876295-96876295 on Build GRCh38
rs1044147943 Chr.2: 96876482-96876482 on Build GRCh38
rs765827824 Chr.2: 96876341-96876341 on Build GRCh38
rs770613818 Chr.2: 96876291-96876291 on Build GRCh38
rs956268248 Chr.2: 96876378-96876378 on Build GRCh38
rs968577780 Chr.2: 96876258-96876258 on Build GRCh38
rs374222221 Chr.2: 96876283-96876283 on Build GRCh38
rs1038291454 Chr.2: 96876581-96876581 on Build GRCh38
rs980956423 Chr.2: 96876491-96876491 on Build GRCh38
rs767107594 Chr.2: 96876316-96876316 on Build GRCh38
rs753326398 Chr.2: 96876343-96876343 on Build GRCh38
rs1053818623 Chr.2: 96876509-96876509 on Build GRCh38
rs957019393 Chr.2: 96876461-96876461 on Build GRCh38
rs564503615 Chr.2: 96876447-96876447 on Build GRCh38
rs747278382 Chr.2: 96876290-96876290 on Build GRCh38
rs939697123 Chr.2: 96876309-96876309 on Build GRCh38
rs756023523 Chr.2: 96876290-96876290 on Build GRCh38
rs779568762 Chr.2: 96876254-96876254 on Build GRCh38
rs376012535 Chr.2: 96876266-96876266 on Build GRCh38
rs775666545 Chr.2: 96876492-96876492 on Build GRCh38
rs553191190 Chr.2: 96876442-96876442 on Build GRCh38
rs942179942 Chr.2: 96876511-96876511 on Build GRCh38
rs1011756764 Chr.2: 96876549-96876549 on Build GRCh38
rs755471840 Chr.2: 96876243-96876243 on Build GRCh38
rs372312893 Chr.2: 96876313-96876313 on Build GRCh38
rs929031608 Chr.2: 96876612-96876612 on Build GRCh38
rs370548325 Chr.2: 96876273-96876273 on Build GRCh38
rs1030400202 Chr.2: 96876386-96876386 on Build GRCh38
rs561539502 Chr.2: 96876542-96876542 on Build GRCh38
rs933920116 Chr.2: 96876247-96876247 on Build GRCh38
rs376634214 Chr.2: 96876315-96876315 on Build GRCh38
rs955224130 Chr.2: 96876427-96876427 on Build GRCh38
rs778964055 Chr.2: 96876410-96876410 on Build GRCh38
rs201518010 Chr.2: 96876302-96876302 on Build GRCh38
rs531132434 Chr.2: 96876472-96876472 on Build GRCh38
rs764311073 Chr.2: 96876664-96876664 on Build GRCh38
rs564902825 Chr.2: 96876471-96876471 on Build GRCh38
rs572930545 Chr.2: 96876452-96876452 on Build GRCh38
rs760627486 Chr.2: 96876614-96876614 on Build GRCh38
rs890150629 Chr.2: 96876400-96876400 on Build GRCh38
rs948467457 Chr.2: 96876474-96876474 on Build GRCh38
rs1007238991 Chr.2: 96876677-96876677 on Build GRCh38
rs571548878 Chr.2: 96876377-96876377 on Build GRCh38
rs138990494 Chr.2: 96876661-96876662 on Build GRCh38
rs113939032 Chr.2: 96876644-96876644 on Build GRCh38
rs367617195 Chr.2: 96876312-96876312 on Build GRCh38
rs766806605 Chr.2: 96876228-96876228 on Build GRCh38
rs181737692 Chr.2: 96876460-96876460 on Build GRCh38

View Full Product Details