Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96893789-96894281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM178B
Entrez Gene ID
51252
Gene Name
family with sequence similarity 178 member B
Gene Aliases
-
UniGene
Hs.107922
Gene Chromosome Location
Chr.2: 96875882-96986600 on Build GRCh38
Transcripts
NM_001122646.2 NM_001172667.1 NM_016490.4 XM_011511294.2 XM_017004263.1 XM_017004264.1 XM_017004265.1 XM_017004266.1 XM_017004267.1 XM_017004268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2825964 c.2140C>G p.L714V Chr.2: 96894006-96894006 on Build GRCh38
5397625 c.2206G>C p.E736Q Chr.2: 96893940-96893940 on Build GRCh38
6659006 c.2190G>A p.Q730Q Chr.2: 96893956-96893956 on Build GRCh38
5648952 c.2196_2197delGG p.K732fs*2 Chr.2: 96893949-96893950 on Build GRCh38
6659005 c.66G>A p.Q22Q Chr.2: 96893956-96893956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759661411 Chr.2: 96893937-96893937 on Build GRCh38
rs762655112 Chr.2: 96893948-96893948 on Build GRCh38
rs549259876 Chr.2: 96894114-96894114 on Build GRCh38
rs147523420 Chr.2: 96893998-96893998 on Build GRCh38
rs755683983 Chr.2: 96893882-96893882 on Build GRCh38
rs937559751 Chr.2: 96894163-96894163 on Build GRCh38
rs921834286 Chr.2: 96894058-96894058 on Build GRCh38
rs765961295 Chr.2: 96894075-96894075 on Build GRCh38
rs376034495 Chr.2: 96894035-96894035 on Build GRCh38
rs993100559 Chr.2: 96894204-96894204 on Build GRCh38
rs540801170 Chr.2: 96893904-96893904 on Build GRCh38
rs753309128 Chr.2: 96894101-96894101 on Build GRCh38
rs145520273 Chr.2: 96894006-96894006 on Build GRCh38
rs528275707 Chr.2: 96894247-96894247 on Build GRCh38
rs1042155642 Chr.2: 96894215-96894215 on Build GRCh38
rs767595027 Chr.2: 96893992-96893992 on Build GRCh38
rs936117663 Chr.2: 96894246-96894246 on Build GRCh38
rs761705760 Chr.2: 96894047-96894047 on Build GRCh38
rs775842387 Chr.2: 96893946-96893946 on Build GRCh38
rs931890915 Chr.2: 96894099-96894099 on Build GRCh38
rs758209046 Chr.2: 96894210-96894210 on Build GRCh38
rs776648654 Chr.2: 96893921-96893921 on Build GRCh38
rs543295888 Chr.2: 96893865-96893865 on Build GRCh38
rs772553566 Chr.2: 96893902-96893902 on Build GRCh38
rs918222275 Chr.2: 96893888-96893888 on Build GRCh38
rs879927962 Chr.2: 96893817-96893817 on Build GRCh38
rs529247829 Chr.2: 96894043-96894043 on Build GRCh38
rs892989574 Chr.2: 96894137-96894137 on Build GRCh38
rs1025398381 Chr.2: 96894109-96894109 on Build GRCh38
rs771447269 Chr.2: 96893911-96893911 on Build GRCh38
rs1012461388 Chr.2: 96894222-96894222 on Build GRCh38
rs565767321 Chr.2: 96894189-96894189 on Build GRCh38
rs967131811 Chr.2: 96893940-96893940 on Build GRCh38
rs772012759 Chr.2: 96893983-96893984 on Build GRCh38
rs764023219 Chr.2: 96893949-96893949 on Build GRCh38
rs35352216 Chr.2: 96893813-96893813 on Build GRCh38
rs374140903 Chr.2: 96894209-96894209 on Build GRCh38
rs1042847780 Chr.2: 96894143-96894143 on Build GRCh38
rs747403258 Chr.2: 96894008-96894008 on Build GRCh38
rs1020941196 Chr.2: 96893930-96893930 on Build GRCh38
rs926977640 Chr.2: 96894117-96894118 on Build GRCh38
rs760072551 Chr.2: 96894074-96894074 on Build GRCh38
rs775546309 Chr.2: 96894040-96894040 on Build GRCh38
rs34913145 Chr.2: 96893879-96893880 on Build GRCh38
rs369630987 Chr.2: 96894036-96894036 on Build GRCh38
rs965084991 Chr.2: 96893811-96893811 on Build GRCh38
rs767505166 Chr.2: 96894049-96894049 on Build GRCh38
rs751522162 Chr.2: 96893954-96893954 on Build GRCh38
rs753566144 Chr.2: 96894004-96894004 on Build GRCh38
rs1006791055 Chr.2: 96894079-96894079 on Build GRCh38
rs201571176 Chr.2: 96893895-96893895 on Build GRCh38
rs774250251 Chr.2: 96894044-96894044 on Build GRCh38
rs1027749320 Chr.2: 96894238-96894238 on Build GRCh38
rs983176486 Chr.2: 96894027-96894027 on Build GRCh38
rs889692031 Chr.2: 96894073-96894073 on Build GRCh38
rs769854303 Chr.2: 96893941-96893941 on Build GRCh38
rs972863943 Chr.2: 96894106-96894106 on Build GRCh38
rs975801885 Chr.2: 96894053-96894053 on Build GRCh38
rs373884209 Chr.2: 96894071-96894071 on Build GRCh38
rs11677797 Chr.2: 96894022-96894022 on Build GRCh38
rs564185157 Chr.2: 96894000-96894000 on Build GRCh38
rs1040709972 Chr.2: 96893810-96893810 on Build GRCh38
rs773985178 Chr.2: 96893876-96893878 on Build GRCh38
rs965201956 Chr.2: 96894093-96894093 on Build GRCh38
rs983798499 Chr.2: 96894116-96894116 on Build GRCh38
rs761793924 Chr.2: 96893981-96893981 on Build GRCh38
rs1035669789 Chr.2: 96893950-96893950 on Build GRCh38
rs759364503 Chr.2: 96893979-96893987 on Build GRCh38
rs1019386453 Chr.2: 96894085-96894085 on Build GRCh38
rs543442848 Chr.2: 96894013-96894013 on Build GRCh38
rs984743827 Chr.2: 96893894-96893895 on Build GRCh38
rs1003947244 Chr.2: 96894194-96894194 on Build GRCh38
rs946762450 Chr.2: 96894142-96894142 on Build GRCh38
rs747561559 Chr.2: 96893908-96893908 on Build GRCh38
rs200012479 Chr.2: 96893890-96893890 on Build GRCh38
rs758666340 Chr.2: 96894005-96894005 on Build GRCh38
rs972531768 Chr.2: 96893850-96893850 on Build GRCh38

View Full Product Details