Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96928819-96929336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM178B
Entrez Gene ID
51252
Gene Name
family with sequence similarity 178 member B
Gene Aliases
-
UniGene
Hs.107922
Gene Chromosome Location
Chr.2: 96875882-96986600 on Build GRCh38
Transcripts
NM_001122646.2 XM_011511294.2 XM_017004263.1 XM_017004264.1 XM_017004265.1 XM_017004266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316311 c.1581C>A p.S527R Chr.2: 96929262-96929262 on Build GRCh38
1410117 c.1536C>T p.H512H Chr.2: 96929307-96929307 on Build GRCh38
1206204 c.1610C>A p.P537H Chr.2: 96929233-96929233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557825899 Chr.2: 96928981-96928981 on Build GRCh38
rs771785447 Chr.2: 96929184-96929184 on Build GRCh38
rs545253787 Chr.2: 96929144-96929144 on Build GRCh38
rs57641223 Chr.2: 96928975-96928975 on Build GRCh38
rs1006802632 Chr.2: 96928930-96928930 on Build GRCh38
rs763054048 Chr.2: 96929251-96929251 on Build GRCh38
rs759988596 Chr.2: 96929214-96929214 on Build GRCh38
rs933760408 Chr.2: 96929018-96929018 on Build GRCh38
rs544406353 Chr.2: 96929100-96929100 on Build GRCh38
rs557466155 Chr.2: 96928974-96928975 on Build GRCh38
rs565561921 Chr.2: 96928984-96928984 on Build GRCh38
rs375952198 Chr.2: 96929265-96929265 on Build GRCh38
rs1054386051 Chr.2: 96928961-96928961 on Build GRCh38
rs941503474 Chr.2: 96928991-96928991 on Build GRCh38
rs901579490 Chr.2: 96928978-96928978 on Build GRCh38
rs999993618 Chr.2: 96929272-96929272 on Build GRCh38
rs764132573 Chr.2: 96929257-96929257 on Build GRCh38
rs1018324869 Chr.2: 96929309-96929309 on Build GRCh38
rs980380496 Chr.2: 96928983-96928983 on Build GRCh38
rs752135352 Chr.2: 96928984-96928984 on Build GRCh38
rs957988905 Chr.2: 96929283-96929283 on Build GRCh38
rs183782343 Chr.2: 96929069-96929069 on Build GRCh38
rs886083264 Chr.2: 96929153-96929153 on Build GRCh38
rs774031706 Chr.2: 96929167-96929167 on Build GRCh38
rs889662386 Chr.2: 96928887-96928887 on Build GRCh38
rs755790846 Chr.2: 96929092-96929092 on Build GRCh38
rs941703338 Chr.2: 96929189-96929189 on Build GRCh38
rs963000870 Chr.2: 96929215-96929215 on Build GRCh38
rs972942141 Chr.2: 96928996-96928996 on Build GRCh38
rs191572585 Chr.2: 96929019-96929019 on Build GRCh38
rs766281364 Chr.2: 96928989-96928989 on Build GRCh38
rs903707295 Chr.2: 96928956-96928956 on Build GRCh38
rs942602321 Chr.2: 96929037-96929037 on Build GRCh38
rs903782068 Chr.2: 96929062-96929062 on Build GRCh38
rs918802354 Chr.2: 96929242-96929242 on Build GRCh38
rs993196085 Chr.2: 96928977-96928977 on Build GRCh38
rs1047488734 Chr.2: 96929148-96929148 on Build GRCh38
rs956154430 Chr.2: 96928971-96928971 on Build GRCh38
rs573490517 Chr.2: 96929142-96929142 on Build GRCh38
rs868261599 Chr.2: 96928980-96928980 on Build GRCh38
rs543345215 Chr.2: 96929096-96929096 on Build GRCh38
rs578019389 Chr.2: 96929013-96929013 on Build GRCh38
rs563141532 Chr.2: 96929295-96929295 on Build GRCh38
rs776201484 Chr.2: 96929236-96929236 on Build GRCh38
rs564234620 Chr.2: 96929194-96929194 on Build GRCh38
rs765390941 Chr.2: 96929141-96929144 on Build GRCh38
rs955293849 Chr.2: 96929254-96929254 on Build GRCh38
rs796270348 Chr.2: 96928983-96928983 on Build GRCh38
rs74492791 Chr.2: 96928979-96928979 on Build GRCh38
rs1031086863 Chr.2: 96929276-96929276 on Build GRCh38
rs370575893 Chr.2: 96928983-96928984 on Build GRCh38
rs958081052 Chr.2: 96929063-96929063 on Build GRCh38
rs934559513 Chr.2: 96928947-96928947 on Build GRCh38
rs1024300775 Chr.2: 96929106-96929106 on Build GRCh38
rs532978003 Chr.2: 96928866-96928866 on Build GRCh38
rs573203682 Chr.2: 96929298-96929298 on Build GRCh38
rs549489762 Chr.2: 96928934-96928934 on Build GRCh38
rs978229142 Chr.2: 96929024-96929024 on Build GRCh38
rs1017171352 Chr.2: 96929208-96929208 on Build GRCh38
rs569979638 Chr.2: 96928937-96928937 on Build GRCh38
rs918005122 Chr.2: 96928889-96928889 on Build GRCh38
rs561011925 Chr.2: 96928883-96928883 on Build GRCh38
rs999383549 Chr.2: 96928965-96928965 on Build GRCh38
rs867665982 Chr.2: 96928966-96928966 on Build GRCh38
rs116325579 Chr.2: 96929012-96929012 on Build GRCh38
rs1043600574 Chr.2: 96929061-96929061 on Build GRCh38
rs191288606 Chr.2: 96929299-96929299 on Build GRCh38
rs113414411 Chr.2: 96929226-96929226 on Build GRCh38
rs1050726064 Chr.2: 96928880-96928880 on Build GRCh38
rs373857264 Chr.2: 96929029-96929029 on Build GRCh38
rs186296857 Chr.2: 96929097-96929097 on Build GRCh38
rs532882213 Chr.2: 96929213-96929213 on Build GRCh38
rs747706354 Chr.2: 96929174-96929174 on Build GRCh38

View Full Product Details