Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97539772-97540269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36B
Entrez Gene ID
57730
Gene Name
ankyrin repeat domain 36B
Gene Aliases
KIAA1641
UniGene
Hs.532921
Gene Chromosome Location
Chr.2: 97492663-97590245 on Build GRCh38
Transcripts
NM_025190.3 XM_005263995.4 XM_006712661.1 XM_011511555.1 XM_011511556.1 XM_011511557.2 XM_011511558.1 XM_017004578.1 XM_017004579.1 XM_017004580.1 XM_017004581.1 XM_017004582.1 XM_017004583.1 XM_017004584.1 XM_017004585.1 XM_017004586.1 XM_017004587.1 XM_017004588.1 XM_017004589.1 XM_017004590.1 XM_017004591.1 XM_017004592.1 XM_017004593.1 XM_017004594.1 XM_017004595.1

SNP Detail

SNP ID Chromosome Location
rs763177225 Chr.2: 97540055-97540055 on Build GRCh38
rs768265577 Chr.2: 97540004-97540004 on Build GRCh38
rs765878296 Chr.2: 97540049-97540049 on Build GRCh38
rs750201549 Chr.2: 97540247-97540247 on Build GRCh38
rs199679277 Chr.2: 97539951-97539952 on Build GRCh38
rs752862930 Chr.2: 97540016-97540017 on Build GRCh38
rs191410935 Chr.2: 97540159-97540159 on Build GRCh38
rs188739692 Chr.2: 97539970-97539970 on Build GRCh38
rs771262129 Chr.2: 97540025-97540025 on Build GRCh38
rs560334033 Chr.2: 97540112-97540112 on Build GRCh38
rs546668314 Chr.2: 97539894-97539894 on Build GRCh38
rs771646047 Chr.2: 97540166-97540166 on Build GRCh38
rs757423022 Chr.2: 97540087-97540087 on Build GRCh38
rs764395062 Chr.2: 97540061-97540061 on Build GRCh38
rs747330873 Chr.2: 97540017-97540017 on Build GRCh38
rs767661695 Chr.2: 97540246-97540246 on Build GRCh38
rs750206294 Chr.2: 97540129-97540129 on Build GRCh38
rs552426780 Chr.2: 97540196-97540196 on Build GRCh38
rs181777503 Chr.2: 97539958-97539958 on Build GRCh38
rs546924269 Chr.2: 97540077-97540077 on Build GRCh38
rs756004385 Chr.2: 97540136-97540136 on Build GRCh38
rs778291448 Chr.2: 97540147-97540147 on Build GRCh38
rs776874398 Chr.2: 97540026-97540026 on Build GRCh38
rs781583750 Chr.2: 97540180-97540180 on Build GRCh38
rs1044467224 Chr.2: 97539931-97539931 on Build GRCh38
rs571090346 Chr.2: 97540202-97540202 on Build GRCh38
rs879142874 Chr.2: 97539951-97539951 on Build GRCh38
rs756293096 Chr.2: 97540130-97540133 on Build GRCh38
rs765406242 Chr.2: 97539885-97539886 on Build GRCh38
rs536823787 Chr.2: 97540214-97540214 on Build GRCh38
rs776201270 Chr.2: 97540207-97540207 on Build GRCh38
rs1002835040 Chr.2: 97540078-97540078 on Build GRCh38
rs907220566 Chr.2: 97539949-97539949 on Build GRCh38
rs760042886 Chr.2: 97540042-97540042 on Build GRCh38
rs762043947 Chr.2: 97540237-97540237 on Build GRCh38
rs748826802 Chr.2: 97539999-97539999 on Build GRCh38
rs754550388 Chr.2: 97540139-97540139 on Build GRCh38
rs575936878 Chr.2: 97539930-97539930 on Build GRCh38
rs548206329 Chr.2: 97539890-97539890 on Build GRCh38
rs2442235 Chr.2: 97540137-97540137 on Build GRCh38
rs774306102 Chr.2: 97540215-97540215 on Build GRCh38
rs561530001 Chr.2: 97540005-97540005 on Build GRCh38
rs186892734 Chr.2: 97540051-97540051 on Build GRCh38

View Full Product Details