Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127861496-127862024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMMECR1L
Entrez Gene ID
83607
Gene Name
AMMECR1 like
Gene Aliases
-
UniGene
Hs.546449
Gene Chromosome Location
Chr.2: 127861630-127886408 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511955.2 XM_017005446.1

Gene Symbol
LOC107985803
Entrez Gene ID
107985803
Gene Name
uncharacterized LOC107985803
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 127858310-127887187 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511955.2 XM_017005446.1

SNP Detail

SNP ID Chromosome Location
rs904988492 Chr.2: 127861584-127861584 on Build GRCh38
rs959456754 Chr.2: 127861942-127861942 on Build GRCh38
rs866410681 Chr.2: 127861776-127861776 on Build GRCh38
rs375931120 Chr.2: 127861774-127861774 on Build GRCh38
rs1024166757 Chr.2: 127861594-127861594 on Build GRCh38
rs961346965 Chr.2: 127861988-127861988 on Build GRCh38
rs768202409 Chr.2: 127861819-127861819 on Build GRCh38
rs982638334 Chr.2: 127861612-127861612 on Build GRCh38
rs574031134 Chr.2: 127861524-127861524 on Build GRCh38
rs76141505 Chr.2: 127861634-127861634 on Build GRCh38
rs533888311 Chr.2: 127861620-127861624 on Build GRCh38
rs1037228806 Chr.2: 127861735-127861735 on Build GRCh38
rs148516805 Chr.2: 127861882-127861882 on Build GRCh38
rs75229967 Chr.2: 127861840-127861840 on Build GRCh38
rs575628637 Chr.2: 127861765-127861765 on Build GRCh38
rs938012219 Chr.2: 127861670-127861670 on Build GRCh38
rs558669055 Chr.2: 127861622-127861624 on Build GRCh38
rs982680723 Chr.2: 127861538-127861538 on Build GRCh38
rs1033347190 Chr.2: 127861572-127861572 on Build GRCh38
rs922621164 Chr.2: 127861527-127861527 on Build GRCh38
rs964510916 Chr.2: 127861766-127861766 on Build GRCh38
rs1054102177 Chr.2: 127861852-127861852 on Build GRCh38
rs750910408 Chr.2: 127861706-127861706 on Build GRCh38
rs971258721 Chr.2: 127861884-127861884 on Build GRCh38
rs909910624 Chr.2: 127861986-127861986 on Build GRCh38
rs1010184318 Chr.2: 127861881-127861881 on Build GRCh38
rs758950365 Chr.2: 127861843-127861843 on Build GRCh38
rs555587074 Chr.2: 127861977-127861977 on Build GRCh38
rs1034187931 Chr.2: 127861925-127861925 on Build GRCh38
rs933952588 Chr.2: 127861528-127861528 on Build GRCh38
rs972995515 Chr.2: 127861990-127861990 on Build GRCh38
rs530662833 Chr.2: 127861866-127861866 on Build GRCh38
rs889485460 Chr.2: 127861545-127861545 on Build GRCh38
rs751819846 Chr.2: 127861559-127861563 on Build GRCh38
rs866267230 Chr.2: 127861923-127861923 on Build GRCh38
rs867442966 Chr.2: 127861757-127861757 on Build GRCh38
rs1000280971 Chr.2: 127861841-127861841 on Build GRCh38
rs149844266 Chr.2: 127861666-127861666 on Build GRCh38
rs373715072 Chr.2: 127861533-127861533 on Build GRCh38
rs1049982988 Chr.2: 127861794-127861794 on Build GRCh38
rs982734044 Chr.2: 127861861-127861861 on Build GRCh38
rs1001336069 Chr.2: 127861889-127861889 on Build GRCh38
rs773873566 Chr.2: 127861918-127861918 on Build GRCh38
rs565151188 Chr.2: 127861728-127861728 on Build GRCh38
rs762632149 Chr.2: 127861702-127861702 on Build GRCh38
rs60235980 Chr.2: 127861549-127861549 on Build GRCh38
rs145854431 Chr.2: 127861703-127861703 on Build GRCh38
rs545266692 Chr.2: 127861711-127861711 on Build GRCh38
rs934154544 Chr.2: 127861793-127861793 on Build GRCh38
rs971204616 Chr.2: 127861600-127861600 on Build GRCh38
rs371697604 Chr.2: 127861700-127861700 on Build GRCh38
rs988659066 Chr.2: 127861587-127861587 on Build GRCh38
rs761181706 Chr.2: 127861619-127861620 on Build GRCh38
rs896158602 Chr.2: 127861620-127861622 on Build GRCh38
rs1033176826 Chr.2: 127861729-127861729 on Build GRCh38
rs751111365 Chr.2: 127861927-127861929 on Build GRCh38
rs955911390 Chr.2: 127861583-127861583 on Build GRCh38
rs984547245 Chr.2: 127861968-127861968 on Build GRCh38

View Full Product Details