Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127874135-127874631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMMECR1L
Entrez Gene ID
83607
Gene Name
AMMECR1 like
Gene Aliases
-
UniGene
Hs.546449
Gene Chromosome Location
Chr.2: 127861630-127886408 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511956.2 XM_017005063.1 XM_017005446.1

Gene Symbol
LOC107985803
Entrez Gene ID
107985803
Gene Name
uncharacterized LOC107985803
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 127858310-127887187 on Build GRCh38
Transcripts
NM_001199140.1 NM_031445.2 XM_005263804.4 XM_005263806.4 XM_005263807.4 XM_011511952.2 XM_011511954.2 XM_011511956.2 XM_017005063.1 XM_017005446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4084750 c.56G>A p.C19Y Chr.2: 127874179-127874179 on Build GRCh38
5394300 c.90A>G p.G30G Chr.2: 127874145-127874145 on Build GRCh38
242926 c.60G>A p.G20G Chr.2: 127874175-127874175 on Build GRCh38
4449106 c.16_18delTGT p.C6delC Chr.2: 127874217-127874219 on Build GRCh38
7128461 c.30C>G p.L10L Chr.2: 127874205-127874205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750124824 Chr.2: 127874221-127874221 on Build GRCh38
rs781212755 Chr.2: 127874271-127874271 on Build GRCh38
rs554223578 Chr.2: 127874369-127874369 on Build GRCh38
rs181382599 Chr.2: 127874355-127874355 on Build GRCh38
rs982707289 Chr.2: 127874424-127874424 on Build GRCh38
rs577732956 Chr.2: 127874513-127874513 on Build GRCh38
rs751087606 Chr.2: 127874245-127874245 on Build GRCh38
rs555348249 Chr.2: 127874359-127874359 on Build GRCh38
rs771188762 Chr.2: 127874161-127874161 on Build GRCh38
rs775664977 Chr.2: 127874187-127874187 on Build GRCh38
rs1036949319 Chr.2: 127874192-127874192 on Build GRCh38
rs565823838 Chr.2: 127874442-127874442 on Build GRCh38
rs374401241 Chr.2: 127874448-127874448 on Build GRCh38
rs772645121 Chr.2: 127874185-127874185 on Build GRCh38
rs557504763 Chr.2: 127874499-127874499 on Build GRCh38
rs13418973 Chr.2: 127874342-127874342 on Build GRCh38
rs963887819 Chr.2: 127874388-127874388 on Build GRCh38
rs534834893 Chr.2: 127874445-127874445 on Build GRCh38
rs1055867705 Chr.2: 127874256-127874256 on Build GRCh38
rs757410529 Chr.2: 127874284-127874284 on Build GRCh38
rs909590678 Chr.2: 127874574-127874574 on Build GRCh38
rs749582865 Chr.2: 127874283-127874283 on Build GRCh38
rs201713365 Chr.2: 127874205-127874205 on Build GRCh38
rs1021150386 Chr.2: 127874402-127874402 on Build GRCh38
rs752395245 Chr.2: 127874275-127874275 on Build GRCh38
rs955248601 Chr.2: 127874430-127874430 on Build GRCh38
rs975590393 Chr.2: 127874407-127874407 on Build GRCh38
rs937514287 Chr.2: 127874528-127874528 on Build GRCh38
rs201727413 Chr.2: 127874240-127874240 on Build GRCh38
rs1051878102 Chr.2: 127874306-127874306 on Build GRCh38
rs1009812242 Chr.2: 127874399-127874399 on Build GRCh38
rs774911630 Chr.2: 127874172-127874172 on Build GRCh38
rs940938374 Chr.2: 127874498-127874498 on Build GRCh38
rs879030279 Chr.2: 127874447-127874447 on Build GRCh38
rs1014249934 Chr.2: 127874312-127874312 on Build GRCh38
rs527335942 Chr.2: 127874604-127874604 on Build GRCh38
rs200409725 Chr.2: 127874263-127874263 on Build GRCh38
rs188584936 Chr.2: 127874197-127874197 on Build GRCh38
rs374145656 Chr.2: 127874244-127874244 on Build GRCh38
rs1032819644 Chr.2: 127874371-127874371 on Build GRCh38
rs376566715 Chr.2: 127874282-127874282 on Build GRCh38
rs758733688 Chr.2: 127874349-127874349 on Build GRCh38
rs760031576 Chr.2: 127874181-127874181 on Build GRCh38
rs1056318484 Chr.2: 127874549-127874549 on Build GRCh38
rs765869626 Chr.2: 127874243-127874243 on Build GRCh38
rs780259169 Chr.2: 127874592-127874592 on Build GRCh38
rs895898868 Chr.2: 127874562-127874562 on Build GRCh38
rs959393408 Chr.2: 127874460-127874460 on Build GRCh38
rs970894016 Chr.2: 127874413-127874413 on Build GRCh38

View Full Product Details