Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 128267438-128267945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST1
Entrez Gene ID
9394
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases
HH15, HS6ST
UniGene
Hs.512841
Gene Chromosome Location
Chr.2: 128265480-128318597 on Build GRCh38
Transcripts
NM_004807.2

SNP Detail

SNP ID Chromosome Location
rs563049278 Chr.2: 128267513-128267513 on Build GRCh38
rs528006987 Chr.2: 128267707-128267707 on Build GRCh38
rs899402372 Chr.2: 128267717-128267717 on Build GRCh38
rs1033950470 Chr.2: 128267606-128267606 on Build GRCh38
rs181410190 Chr.2: 128267742-128267742 on Build GRCh38
rs568626790 Chr.2: 128267703-128267703 on Build GRCh38
rs376696493 Chr.2: 128267576-128267576 on Build GRCh38
rs11554550 Chr.2: 128267874-128267874 on Build GRCh38
rs866645720 Chr.2: 128267684-128267684 on Build GRCh38
rs539842525 Chr.2: 128267767-128267767 on Build GRCh38
rs4662597 Chr.2: 128267894-128267894 on Build GRCh38
rs995208928 Chr.2: 128267740-128267740 on Build GRCh38
rs58484518 Chr.2: 128267830-128267830 on Build GRCh38
rs71420833 Chr.2: 128267822-128267822 on Build GRCh38
rs541516135 Chr.2: 128267830-128267830 on Build GRCh38
rs571127751 Chr.2: 128267745-128267745 on Build GRCh38
rs528952598 Chr.2: 128267650-128267650 on Build GRCh38
rs758627319 Chr.2: 128267646-128267647 on Build GRCh38
rs535632794 Chr.2: 128267778-128267778 on Build GRCh38
rs556592096 Chr.2: 128267771-128267771 on Build GRCh38
rs34162250 Chr.2: 128267915-128267915 on Build GRCh38
rs185303149 Chr.2: 128267858-128267858 on Build GRCh38
rs529322529 Chr.2: 128267544-128267546 on Build GRCh38
rs548541461 Chr.2: 128267666-128267666 on Build GRCh38
rs3916055 Chr.2: 128267575-128267575 on Build GRCh38
rs1026607892 Chr.2: 128267741-128267741 on Build GRCh38
rs150392154 Chr.2: 128267481-128267481 on Build GRCh38
rs936581168 Chr.2: 128267835-128267835 on Build GRCh38
rs570200930 Chr.2: 128267821-128267821 on Build GRCh38
rs201711108 Chr.2: 128267837-128267837 on Build GRCh38
rs754610159 Chr.2: 128267829-128267830 on Build GRCh38
rs982497353 Chr.2: 128267790-128267790 on Build GRCh38
rs913912768 Chr.2: 128267868-128267868 on Build GRCh38
rs532780178 Chr.2: 128267479-128267479 on Build GRCh38
rs758172474 Chr.2: 128267480-128267480 on Build GRCh38
rs989514974 Chr.2: 128267840-128267840 on Build GRCh38
rs375280117 Chr.2: 128267567-128267567 on Build GRCh38
rs1019296444 Chr.2: 128267803-128267803 on Build GRCh38

View Full Product Details