Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131529354-131529857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC74A
Entrez Gene ID
90557
Gene Name
coiled-coil domain containing 74A
Gene Aliases
-
UniGene
Hs.351461
Gene Chromosome Location
Chr.2: 131522621-131533666 on Build GRCh38
Transcripts
NM_001258304.1 NM_001258305.1 NM_001258306.1 NM_138770.2 XM_005263835.3 XM_005263837.1 XM_006712828.3 XM_017005234.1 XM_017005235.1 XM_017005236.1 XM_017005237.1 XM_017005238.1 XM_017005239.1 XM_017005240.1 XM_017005241.1 XM_017005242.1 XM_017005243.1 XM_017005244.1 XM_017005245.1 XM_017005246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4392771 c.257A>G p.H86R Chr.2: 131529653-131529653 on Build GRCh38
5767781 c.271A>G p.M91V Chr.2: 131529667-131529667 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745877290 Chr.2: 131529649-131529649 on Build GRCh38
rs1047076786 Chr.2: 131529821-131529821 on Build GRCh38
rs747108435 Chr.2: 131529755-131529755 on Build GRCh38
rs931184026 Chr.2: 131529791-131529791 on Build GRCh38
rs556767141 Chr.2: 131529430-131529430 on Build GRCh38
rs755726356 Chr.2: 131529704-131529704 on Build GRCh38
rs376331795 Chr.2: 131529673-131529673 on Build GRCh38
rs867019104 Chr.2: 131529532-131529532 on Build GRCh38
rs754486449 Chr.2: 131529615-131529615 on Build GRCh38
rs746354133 Chr.2: 131529729-131529729 on Build GRCh38
rs750271391 Chr.2: 131529588-131529588 on Build GRCh38
rs745327222 Chr.2: 131529486-131529486 on Build GRCh38
rs759683851 Chr.2: 131529562-131529562 on Build GRCh38
rs749558078 Chr.2: 131529723-131529723 on Build GRCh38
rs879022376 Chr.2: 131529691-131529691 on Build GRCh38
rs772744906 Chr.2: 131529736-131529736 on Build GRCh38
rs768337007 Chr.2: 131529501-131529501 on Build GRCh38
rs368234271 Chr.2: 131529505-131529505 on Build GRCh38
rs935765037 Chr.2: 131529426-131529426 on Build GRCh38
rs762275349 Chr.2: 131529437-131529437 on Build GRCh38
rs780240415 Chr.2: 131529744-131529744 on Build GRCh38
rs777504044 Chr.2: 131529720-131529720 on Build GRCh38
rs781326391 Chr.2: 131529752-131529753 on Build GRCh38
rs771511669 Chr.2: 131529561-131529561 on Build GRCh38
rs1028647243 Chr.2: 131529513-131529513 on Build GRCh38
rs1019817186 Chr.2: 131529395-131529395 on Build GRCh38
rs974383301 Chr.2: 131529422-131529422 on Build GRCh38
rs755069175 Chr.2: 131529661-131529661 on Build GRCh38
rs201257947 Chr.2: 131529633-131529633 on Build GRCh38
rs749697554 Chr.2: 131529637-131529637 on Build GRCh38
rs1055821827 Chr.2: 131529460-131529460 on Build GRCh38
rs763586479 Chr.2: 131529527-131529527 on Build GRCh38
rs138266222 Chr.2: 131529665-131529665 on Build GRCh38
rs768245156 Chr.2: 131529540-131529540 on Build GRCh38
rs184436886 Chr.2: 131529423-131529423 on Build GRCh38
rs750971922 Chr.2: 131529610-131529610 on Build GRCh38
rs775861404 Chr.2: 131529570-131529570 on Build GRCh38
rs148172431 Chr.2: 131529537-131529537 on Build GRCh38
rs745403262 Chr.2: 131529400-131529400 on Build GRCh38
rs977485686 Chr.2: 131529745-131529745 on Build GRCh38
rs1020329704 Chr.2: 131529713-131529713 on Build GRCh38
rs900000089 Chr.2: 131529632-131529632 on Build GRCh38
rs752247211 Chr.2: 131529701-131529701 on Build GRCh38
rs778607894 Chr.2: 131529483-131529483 on Build GRCh38
rs907125483 Chr.2: 131529450-131529450 on Build GRCh38
rs757755448 Chr.2: 131529531-131529542 on Build GRCh38
rs566374576 Chr.2: 131529646-131529646 on Build GRCh38
rs753408416 Chr.2: 131529493-131529493 on Build GRCh38
rs779670281 Chr.2: 131529487-131529487 on Build GRCh38
rs770224931 Chr.2: 131529515-131529515 on Build GRCh38
rs542783411 Chr.2: 131529536-131529536 on Build GRCh38
rs550380545 Chr.2: 131529543-131529543 on Build GRCh38
rs771707842 Chr.2: 131529410-131529410 on Build GRCh38
rs754374796 Chr.2: 131529514-131529539 on Build GRCh38
rs527680246 Chr.2: 131529535-131529535 on Build GRCh38
rs115035364 Chr.2: 131529447-131529447 on Build GRCh38
rs539387343 Chr.2: 131529809-131529809 on Build GRCh38
rs201079511 Chr.2: 131529381-131529381 on Build GRCh38
rs765100368 Chr.2: 131529587-131529587 on Build GRCh38
rs764351209 Chr.2: 131529432-131529432 on Build GRCh38
rs551652544 Chr.2: 131529620-131529620 on Build GRCh38
rs765761438 Chr.2: 131529443-131529443 on Build GRCh38
rs755816070 Chr.2: 131529629-131529629 on Build GRCh38
rs755954256 Chr.2: 131529692-131529692 on Build GRCh38
rs758652522 Chr.2: 131529591-131529592 on Build GRCh38
rs116399863 Chr.2: 131529657-131529657 on Build GRCh38
rs1019702739 Chr.2: 131529499-131529499 on Build GRCh38
rs774936277 Chr.2: 131529647-131529647 on Build GRCh38
rs764407846 Chr.2: 131529661-131529661 on Build GRCh38
rs766993815 Chr.2: 131529682-131529682 on Build GRCh38
rs371047111 Chr.2: 131529726-131529726 on Build GRCh38
rs967169781 Chr.2: 131529714-131529714 on Build GRCh38
rs373694975 Chr.2: 131529728-131529728 on Build GRCh38
rs761035196 Chr.2: 131529578-131529578 on Build GRCh38
rs578251204 Chr.2: 131529445-131529445 on Build GRCh38
rs777029189 Chr.2: 131529433-131529433 on Build GRCh38
rs189269221 Chr.2: 131529553-131529553 on Build GRCh38
rs908509862 Chr.2: 131529823-131529823 on Build GRCh38
rs578209983 Chr.2: 131529781-131529781 on Build GRCh38
rs747787886 Chr.2: 131529624-131529624 on Build GRCh38
rs560072033 Chr.2: 131529536-131529537 on Build GRCh38
rs531656696 Chr.2: 131529539-131529539 on Build GRCh38
rs928481170 Chr.2: 131529734-131529734 on Build GRCh38
rs773687308 Chr.2: 131529554-131529554 on Build GRCh38
rs779018089 Chr.2: 131529484-131529484 on Build GRCh38
rs776937225 Chr.2: 131529504-131529504 on Build GRCh38
rs989904520 Chr.2: 131529442-131529442 on Build GRCh38
rs766358918 Chr.2: 131529604-131529604 on Build GRCh38
rs749739607 Chr.2: 131529559-131529559 on Build GRCh38
rs768698531 Chr.2: 131529764-131529764 on Build GRCh38
rs537552766 Chr.2: 131529634-131529634 on Build GRCh38
rs958508406 Chr.2: 131529753-131529753 on Build GRCh38
rs951565096 Chr.2: 131529386-131529386 on Build GRCh38
rs767791559 Chr.2: 131529563-131529563 on Build GRCh38
rs574189534 Chr.2: 131529393-131529393 on Build GRCh38
rs757220876 Chr.2: 131529482-131529482 on Build GRCh38
rs147307685 Chr.2: 131529653-131529653 on Build GRCh38
rs944157966 Chr.2: 131529834-131529834 on Build GRCh38
rs757133118 Chr.2: 131529538-131529538 on Build GRCh38
rs747223445 Chr.2: 131529481-131529484 on Build GRCh38
rs779214153 Chr.2: 131529817-131529817 on Build GRCh38
rs555032574 Chr.2: 131529655-131529655 on Build GRCh38
rs373190695 Chr.2: 131529669-131529669 on Build GRCh38
rs533065298 Chr.2: 131529601-131529601 on Build GRCh38
rs765639124 Chr.2: 131529534-131529535 on Build GRCh38
rs868318217 Chr.2: 131529490-131529490 on Build GRCh38
rs937253959 Chr.2: 131529541-131529541 on Build GRCh38
rs919863761 Chr.2: 131529788-131529788 on Build GRCh38
rs548558651 Chr.2: 131529475-131529475 on Build GRCh38
rs780185652 Chr.2: 131529628-131529658 on Build GRCh38
rs773710233 Chr.2: 131529521-131529521 on Build GRCh38
rs751728759 Chr.2: 131529465-131529465 on Build GRCh38
rs964230684 Chr.2: 131529415-131529415 on Build GRCh38
rs115803849 Chr.2: 131529455-131529455 on Build GRCh38

View Full Product Details