Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131751381-131751923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf27A
Entrez Gene ID
29798
Gene Name
chromosome 2 open reading frame 27A
Gene Aliases
C2orf27, C2orf27B
UniGene
Hs.635289
Gene Chromosome Location
Chr.2: 131722491-131767404 on Build GRCh38
Transcripts
NM_013310.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
475907 c.9C>T p.V3V Chr.2: 131751566-131751566 on Build GRCh38
3708893 c.28+4A>G p.? Chr.2: 131751589-131751589 on Build GRCh38
6221026 c.12G>C p.K4N Chr.2: 131751569-131751569 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760075202 Chr.2: 131751556-131751556 on Build GRCh38
rs199616954 Chr.2: 131751439-131751439 on Build GRCh38
rs1017901734 Chr.2: 131751779-131751779 on Build GRCh38
rs769519507 Chr.2: 131751540-131751540 on Build GRCh38
rs775567612 Chr.2: 131751575-131751575 on Build GRCh38
rs998121597 Chr.2: 131751581-131751581 on Build GRCh38
rs764379155 Chr.2: 131751589-131751589 on Build GRCh38
rs182796512 Chr.2: 131751549-131751549 on Build GRCh38
rs1056294079 Chr.2: 131751464-131751464 on Build GRCh38
rs200729462 Chr.2: 131751538-131751538 on Build GRCh38
rs968080279 Chr.2: 131751790-131751790 on Build GRCh38
rs368189563 Chr.2: 131751633-131751633 on Build GRCh38
rs918037279 Chr.2: 131751471-131751471 on Build GRCh38
rs773384041 Chr.2: 131751541-131751541 on Build GRCh38
rs755405355 Chr.2: 131751770-131751770 on Build GRCh38
rs751930446 Chr.2: 131751618-131751619 on Build GRCh38
rs58354513 Chr.2: 131751583-131751583 on Build GRCh38
rs372499598 Chr.2: 131751529-131751529 on Build GRCh38
rs1045109714 Chr.2: 131751524-131751524 on Build GRCh38
rs955140797 Chr.2: 131751727-131751727 on Build GRCh38
rs948101684 Chr.2: 131751477-131751477 on Build GRCh38
rs374086974 Chr.2: 131751566-131751566 on Build GRCh38
rs771303060 Chr.2: 131751592-131751592 on Build GRCh38
rs77516598 Chr.2: 131751820-131751820 on Build GRCh38
rs1006565312 Chr.2: 131751746-131751746 on Build GRCh38
rs760766383 Chr.2: 131751577-131751577 on Build GRCh38
rs758213107 Chr.2: 131751605-131751605 on Build GRCh38
rs956638669 Chr.2: 131751898-131751898 on Build GRCh38
rs774835258 Chr.2: 131751551-131751551 on Build GRCh38
rs780810790 Chr.2: 131751625-131751625 on Build GRCh38
rs923802910 Chr.2: 131751409-131751409 on Build GRCh38
rs979119503 Chr.2: 131751824-131751824 on Build GRCh38
rs935202823 Chr.2: 131751461-131751461 on Build GRCh38
rs754990020 Chr.2: 131751616-131751616 on Build GRCh38
rs376334655 Chr.2: 131751535-131751535 on Build GRCh38
rs201634213 Chr.2: 131751548-131751548 on Build GRCh38
rs747014440 Chr.2: 131751512-131751512 on Build GRCh38
rs529484725 Chr.2: 131751767-131751767 on Build GRCh38
rs199932707 Chr.2: 131751685-131751686 on Build GRCh38
rs398080607 Chr.2: 131751819-131751819 on Build GRCh38
rs923890931 Chr.2: 131751835-131751835 on Build GRCh38
rs138153832 Chr.2: 131751684-131751684 on Build GRCh38
rs766321531 Chr.2: 131751613-131751613 on Build GRCh38
rs201222308 Chr.2: 131751568-131751568 on Build GRCh38
rs767561675 Chr.2: 131751563-131751563 on Build GRCh38
rs188355772 Chr.2: 131751614-131751614 on Build GRCh38
rs372594336 Chr.2: 131751737-131751737 on Build GRCh38

View Full Product Details