Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 148937196-148937693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5C
Entrez Gene ID
3800
Gene Name
kinesin family member 5C
Gene Aliases
CDCBM2, KINN, NKHC, NKHC-2, NKHC2
UniGene
Hs.435557
Gene Chromosome Location
Chr.2: 148875223-149026759 on Build GRCh38
Transcripts
NM_004522.2 XM_017004062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
282255 c.348C>A p.I116I Chr.2: 148937340-148937340 on Build GRCh38
5907197 c.294G>A p.G98G Chr.2: 148937286-148937286 on Build GRCh38
3701952 c.71A>G p.Y24C Chr.2: 148937354-148937354 on Build GRCh38
5394465 c.304G>A p.D102N Chr.2: 148937296-148937296 on Build GRCh38
6418792 c.2G>T p.G1V Chr.2: 148937285-148937285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773279411 Chr.2: 148937294-148937294 on Build GRCh38
rs758384229 Chr.2: 148937338-148937338 on Build GRCh38
rs550581047 Chr.2: 148937392-148937392 on Build GRCh38
rs754456238 Chr.2: 148937366-148937366 on Build GRCh38
rs747801298 Chr.2: 148937387-148937387 on Build GRCh38
rs773743084 Chr.2: 148937268-148937268 on Build GRCh38
rs372501544 Chr.2: 148937275-148937275 on Build GRCh38
rs777116777 Chr.2: 148937257-148937257 on Build GRCh38
rs866132278 Chr.2: 148937427-148937427 on Build GRCh38
rs749852266 Chr.2: 148937402-148937402 on Build GRCh38
rs1057026810 Chr.2: 148937298-148937298 on Build GRCh38
rs916054154 Chr.2: 148937226-148937226 on Build GRCh38
rs926761118 Chr.2: 148937627-148937627 on Build GRCh38
rs770545145 Chr.2: 148937555-148937555 on Build GRCh38
rs779765780 Chr.2: 148937343-148937343 on Build GRCh38
rs541496957 Chr.2: 148937260-148937260 on Build GRCh38
rs928835301 Chr.2: 148937577-148937577 on Build GRCh38
rs972079013 Chr.2: 148937252-148937252 on Build GRCh38
rs930646346 Chr.2: 148937514-148937514 on Build GRCh38
rs966105620 Chr.2: 148937465-148937465 on Build GRCh38
rs561809053 Chr.2: 148937546-148937546 on Build GRCh38
rs375819301 Chr.2: 148937346-148937346 on Build GRCh38
rs759547162 Chr.2: 148937436-148937436 on Build GRCh38
rs759871280 Chr.2: 148937278-148937278 on Build GRCh38
rs779675787 Chr.2: 148937238-148937238 on Build GRCh38
rs530707663 Chr.2: 148937511-148937511 on Build GRCh38
rs973949974 Chr.2: 148937483-148937483 on Build GRCh38
rs373694182 Chr.2: 148937232-148937232 on Build GRCh38
rs763419879 Chr.2: 148937274-148937274 on Build GRCh38
rs752904856 Chr.2: 148937308-148937308 on Build GRCh38
rs528882173 Chr.2: 148937382-148937382 on Build GRCh38
rs878980616 Chr.2: 148937447-148937447 on Build GRCh38
rs750422140 Chr.2: 148937333-148937333 on Build GRCh38
rs889706144 Chr.2: 148937407-148937407 on Build GRCh38
rs140607607 Chr.2: 148937364-148937364 on Build GRCh38
rs548915702 Chr.2: 148937644-148937644 on Build GRCh38
rs930615403 Chr.2: 148937266-148937266 on Build GRCh38
rs755521821 Chr.2: 148937439-148937439 on Build GRCh38
rs939540594 Chr.2: 148937277-148937277 on Build GRCh38
rs1026319117 Chr.2: 148937485-148937485 on Build GRCh38
rs985100809 Chr.2: 148937225-148937225 on Build GRCh38
rs1015842744 Chr.2: 148937449-148937449 on Build GRCh38
rs746720526 Chr.2: 148937240-148937240 on Build GRCh38
rs1008039012 Chr.2: 148937421-148937421 on Build GRCh38
rs773544630 Chr.2: 148937398-148937398 on Build GRCh38
rs771424772 Chr.2: 148937412-148937412 on Build GRCh38
rs369951019 Chr.2: 148937243-148937243 on Build GRCh38
rs981604153 Chr.2: 148937557-148937557 on Build GRCh38
rs763044028 Chr.2: 148937507-148937507 on Build GRCh38
rs972168569 Chr.2: 148937470-148937470 on Build GRCh38
rs767636994 Chr.2: 148937299-148937299 on Build GRCh38
rs545025418 Chr.2: 148937430-148937430 on Build GRCh38
rs773772474 Chr.2: 148937586-148937586 on Build GRCh38
rs916124067 Chr.2: 148937519-148937519 on Build GRCh38
rs562625899 Chr.2: 148937420-148937420 on Build GRCh38
rs757025401 Chr.2: 148937325-148937325 on Build GRCh38
rs949004189 Chr.2: 148937548-148937548 on Build GRCh38
rs187403886 Chr.2: 148937437-148937437 on Build GRCh38
rs770151934 Chr.2: 148937261-148937261 on Build GRCh38

View Full Product Details