Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 148941851-148942356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5C
Entrez Gene ID
3800
Gene Name
kinesin family member 5C
Gene Aliases
CDCBM2, KINN, NKHC, NKHC-2, NKHC2
UniGene
Hs.435557
Gene Chromosome Location
Chr.2: 148875223-149026759 on Build GRCh38
Transcripts
NM_004522.2 XM_017004062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354226 c.155-1G>A p.? Chr.2: 148941934-148941934 on Build GRCh38
5155435 c.501+6A>C p.? Chr.2: 148941996-148941996 on Build GRCh38
5680101 c.163A>G p.T55A Chr.2: 148941943-148941943 on Build GRCh38
5860211 c.175G>T p.V59F Chr.2: 148941955-148941955 on Build GRCh38
6434242 c.491C>A p.P164Q Chr.2: 148941980-148941980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779304805 Chr.2: 148942024-148942024 on Build GRCh38
rs947856866 Chr.2: 148942064-148942064 on Build GRCh38
rs1037560627 Chr.2: 148942088-148942088 on Build GRCh38
rs965363784 Chr.2: 148941901-148941901 on Build GRCh38
rs1055032189 Chr.2: 148942253-148942253 on Build GRCh38
rs753439237 Chr.2: 148942007-148942007 on Build GRCh38
rs1046349929 Chr.2: 148942113-148942113 on Build GRCh38
rs1011650874 Chr.2: 148942199-148942199 on Build GRCh38
rs749658964 Chr.2: 148941885-148941885 on Build GRCh38
rs891901751 Chr.2: 148942194-148942194 on Build GRCh38
rs1022102476 Chr.2: 148942227-148942227 on Build GRCh38
rs766012984 Chr.2: 148942212-148942212 on Build GRCh38
rs771181796 Chr.2: 148942266-148942266 on Build GRCh38
rs759499744 Chr.2: 148941973-148941973 on Build GRCh38
rs940481735 Chr.2: 148942052-148942052 on Build GRCh38
rs751495278 Chr.2: 148941966-148941966 on Build GRCh38
rs766995170 Chr.2: 148941980-148941980 on Build GRCh38
rs762866665 Chr.2: 148941955-148941955 on Build GRCh38
rs772616475 Chr.2: 148942028-148942028 on Build GRCh38
rs557396273 Chr.2: 148942012-148942012 on Build GRCh38
rs1000071698 Chr.2: 148942327-148942327 on Build GRCh38
rs78597522 Chr.2: 148942185-148942185 on Build GRCh38
rs895190612 Chr.2: 148942284-148942284 on Build GRCh38
rs375278207 Chr.2: 148941945-148941945 on Build GRCh38
rs906415272 Chr.2: 148942131-148942131 on Build GRCh38
rs554948713 Chr.2: 148942127-148942127 on Build GRCh38
rs761031427 Chr.2: 148941939-148941939 on Build GRCh38
rs746344928 Chr.2: 148941911-148941911 on Build GRCh38
rs766239754 Chr.2: 148941964-148941964 on Build GRCh38
rs902782471 Chr.2: 148942302-148942302 on Build GRCh38
rs575065922 Chr.2: 148942211-148942211 on Build GRCh38
rs13411910 Chr.2: 148941998-148941998 on Build GRCh38
rs267598915 Chr.2: 148941961-148941961 on Build GRCh38
rs572195275 Chr.2: 148941994-148941994 on Build GRCh38
rs575753436 Chr.2: 148942056-148942056 on Build GRCh38
rs374838707 Chr.2: 148942043-148942043 on Build GRCh38
rs752248579 Chr.2: 148941990-148941990 on Build GRCh38
rs923483326 Chr.2: 148941941-148941941 on Build GRCh38
rs73970610 Chr.2: 148942289-148942289 on Build GRCh38
rs779307872 Chr.2: 148941902-148941902 on Build GRCh38
rs775613404 Chr.2: 148941937-148941937 on Build GRCh38
rs746410979 Chr.2: 148942025-148942025 on Build GRCh38
rs537638140 Chr.2: 148942104-148942104 on Build GRCh38
rs776923833 Chr.2: 148941949-148941949 on Build GRCh38
rs371397019 Chr.2: 148942008-148942008 on Build GRCh38
rs1021778756 Chr.2: 148941881-148941882 on Build GRCh38
rs149625146 Chr.2: 148942292-148942292 on Build GRCh38
rs560712796 Chr.2: 148942261-148942261 on Build GRCh38
rs931444824 Chr.2: 148942209-148942209 on Build GRCh38
rs750002957 Chr.2: 148941918-148941918 on Build GRCh38
rs1002910243 Chr.2: 148942158-148942158 on Build GRCh38
rs542202861 Chr.2: 148942217-148942217 on Build GRCh38
rs780060876 Chr.2: 148942034-148942034 on Build GRCh38

View Full Product Details