Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157284152-157284651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT5
Entrez Gene ID
11227
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Gene Aliases
GALNAC-T5, GALNACT5
UniGene
Hs.269027
Gene Chromosome Location
Chr.2: 157257548-157318491 on Build GRCh38
Transcripts
NM_014568.1 XM_017003237.1 XM_017003238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5860328 c.1588G>A p.E530K Chr.2: 157284415-157284415 on Build GRCh38
4902498 c.1483C>T p.L495F Chr.2: 157284310-157284310 on Build GRCh38
716234 c.1495A>T p.S499C Chr.2: 157284322-157284322 on Build GRCh38
5928285 c.1564C>T p.R522C Chr.2: 157284391-157284391 on Build GRCh38
32456 c.1521A>T p.E507D Chr.2: 157284348-157284348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769504678 Chr.2: 157284283-157284283 on Build GRCh38
rs954223280 Chr.2: 157284560-157284560 on Build GRCh38
rs955130663 Chr.2: 157284524-157284524 on Build GRCh38
rs1030425525 Chr.2: 157284504-157284504 on Build GRCh38
rs2289942 Chr.2: 157284205-157284205 on Build GRCh38
rs79679728 Chr.2: 157284618-157284618 on Build GRCh38
rs768470109 Chr.2: 157284247-157284247 on Build GRCh38
rs757305711 Chr.2: 157284369-157284369 on Build GRCh38
rs764503542 Chr.2: 157284463-157284463 on Build GRCh38
rs781104495 Chr.2: 157284393-157284393 on Build GRCh38
rs747926566 Chr.2: 157284260-157284260 on Build GRCh38
rs772769659 Chr.2: 157284448-157284448 on Build GRCh38
rs949195889 Chr.2: 157284621-157284621 on Build GRCh38
rs184966944 Chr.2: 157284579-157284579 on Build GRCh38
rs714649 Chr.2: 157284580-157284580 on Build GRCh38
rs761114901 Chr.2: 157284337-157284337 on Build GRCh38
rs779011189 Chr.2: 157284389-157284389 on Build GRCh38
rs886477725 Chr.2: 157284332-157284332 on Build GRCh38
rs773848921 Chr.2: 157284439-157284439 on Build GRCh38
rs764905095 Chr.2: 157284484-157284485 on Build GRCh38
rs751466241 Chr.2: 157284390-157284390 on Build GRCh38
rs755937233 Chr.2: 157284493-157284493 on Build GRCh38
rs768002617 Chr.2: 157284309-157284309 on Build GRCh38
rs754037778 Chr.2: 157284466-157284466 on Build GRCh38
rs946502526 Chr.2: 157284179-157284179 on Build GRCh38
rs79136790 Chr.2: 157284459-157284459 on Build GRCh38
rs559962630 Chr.2: 157284445-157284445 on Build GRCh38
rs148501546 Chr.2: 157284432-157284432 on Build GRCh38
rs776106957 Chr.2: 157284450-157284450 on Build GRCh38
rs769557092 Chr.2: 157284409-157284409 on Build GRCh38
rs966807873 Chr.2: 157284576-157284576 on Build GRCh38
rs530726426 Chr.2: 157284523-157284523 on Build GRCh38
rs753167525 Chr.2: 157284325-157284325 on Build GRCh38
rs754968130 Chr.2: 157284391-157284391 on Build GRCh38
rs752584505 Chr.2: 157284492-157284492 on Build GRCh38
rs761396886 Chr.2: 157284368-157284369 on Build GRCh38
rs529341755 Chr.2: 157284612-157284612 on Build GRCh38
rs746914925 Chr.2: 157284245-157284245 on Build GRCh38
rs774754859 Chr.2: 157284297-157284297 on Build GRCh38
rs189685278 Chr.2: 157284199-157284199 on Build GRCh38
rs747978590 Chr.2: 157284406-157284406 on Build GRCh38
rs754067832 Chr.2: 157284368-157284368 on Build GRCh38
rs759099839 Chr.2: 157284441-157284441 on Build GRCh38
rs996317049 Chr.2: 157284465-157284465 on Build GRCh38
rs979103148 Chr.2: 157284577-157284577 on Build GRCh38
rs896584901 Chr.2: 157284210-157284210 on Build GRCh38
rs765365003 Chr.2: 157284473-157284473 on Build GRCh38
rs748934341 Chr.2: 157284423-157284423 on Build GRCh38
rs1006211792 Chr.2: 157284351-157284351 on Build GRCh38
rs999590287 Chr.2: 157284264-157284264 on Build GRCh38
rs139539481 Chr.2: 157284290-157284290 on Build GRCh38
rs770356046 Chr.2: 157284438-157284438 on Build GRCh38
rs764448608 Chr.2: 157284358-157284358 on Build GRCh38
rs2289941 Chr.2: 157284567-157284567 on Build GRCh38
rs750584805 Chr.2: 157284485-157284485 on Build GRCh38
rs777316011 Chr.2: 157284415-157284415 on Build GRCh38
rs760864607 Chr.2: 157284617-157284617 on Build GRCh38
rs714648 Chr.2: 157284559-157284559 on Build GRCh38
rs776352250 Chr.2: 157284259-157284259 on Build GRCh38
rs6759356 Chr.2: 157284294-157284294 on Build GRCh38
rs758443100 Chr.2: 157284489-157284489 on Build GRCh38
rs551076276 Chr.2: 157284352-157284352 on Build GRCh38
rs779903603 Chr.2: 157284232-157284232 on Build GRCh38
rs181245010 Chr.2: 157284296-157284296 on Build GRCh38
rs762027970 Chr.2: 157284471-157284471 on Build GRCh38
rs760026471 Chr.2: 157284306-157284306 on Build GRCh38
rs1012498274 Chr.2: 157284388-157284388 on Build GRCh38
rs769864861 Chr.2: 157284223-157284223 on Build GRCh38
rs937882215 Chr.2: 157284586-157284586 on Build GRCh38

View Full Product Details