Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 158338572-158339115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC148
Entrez Gene ID
130940
Gene Name
coiled-coil domain containing 148
Gene Aliases
-
UniGene
Hs.668597
Gene Chromosome Location
Chr.2: 158171356-158456753 on Build GRCh38
Transcripts
NM_001301684.1 NM_001301685.1 NM_138803.3 XM_005246319.3 XM_011510638.2 XM_017003392.1 XM_017003393.1 XM_017003394.1 XM_017003395.1 XM_017003396.1 XM_017003397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5767744 c.701A>T p.K234M Chr.2: 158338789-158338789 on Build GRCh38
3798069 c.672G>C p.L224F Chr.2: 158338818-158338818 on Build GRCh38
7359801 c.663C>A p.Y221* Chr.2: 158338827-158338827 on Build GRCh38
1691190 c.575G>A p.S192N Chr.2: 158338997-158338997 on Build GRCh38
3757691 c.720G>A p.Q240Q Chr.2: 158338770-158338770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754266027 Chr.2: 158338794-158338794 on Build GRCh38
rs767988980 Chr.2: 158338946-158338946 on Build GRCh38
rs757456150 Chr.2: 158338722-158338722 on Build GRCh38
rs914641178 Chr.2: 158338889-158338889 on Build GRCh38
rs16842890 Chr.2: 158338770-158338770 on Build GRCh38
rs889169397 Chr.2: 158338599-158338599 on Build GRCh38
rs769594233 Chr.2: 158338630-158338630 on Build GRCh38
rs763388934 Chr.2: 158338978-158338978 on Build GRCh38
rs117168817 Chr.2: 158338629-158338629 on Build GRCh38
rs201834765 Chr.2: 158338748-158338748 on Build GRCh38
rs142481955 Chr.2: 158338744-158338744 on Build GRCh38
rs766681923 Chr.2: 158338982-158338982 on Build GRCh38
rs942738800 Chr.2: 158338606-158338606 on Build GRCh38
rs927634236 Chr.2: 158338733-158338733 on Build GRCh38
rs763523827 Chr.2: 158338882-158338882 on Build GRCh38
rs371463737 Chr.2: 158338888-158338888 on Build GRCh38
rs761193577 Chr.2: 158338790-158338790 on Build GRCh38
rs11686762 Chr.2: 158338713-158338713 on Build GRCh38
rs759016308 Chr.2: 158338695-158338695 on Build GRCh38
rs773020595 Chr.2: 158338756-158338756 on Build GRCh38
rs780530986 Chr.2: 158339045-158339045 on Build GRCh38
rs141574148 Chr.2: 158338873-158338873 on Build GRCh38
rs767436755 Chr.2: 158338990-158338990 on Build GRCh38
rs777833058 Chr.2: 158338892-158338892 on Build GRCh38
rs894750211 Chr.2: 158339077-158339077 on Build GRCh38
rs367936711 Chr.2: 158338661-158338661 on Build GRCh38
rs759665333 Chr.2: 158338989-158338989 on Build GRCh38
rs753191402 Chr.2: 158338703-158338703 on Build GRCh38
rs552025563 Chr.2: 158338726-158338726 on Build GRCh38
rs778628228 Chr.2: 158338937-158338937 on Build GRCh38
rs969135906 Chr.2: 158338710-158338710 on Build GRCh38
rs371269896 Chr.2: 158338985-158338985 on Build GRCh38
rs751118092 Chr.2: 158338690-158338690 on Build GRCh38
rs777550315 Chr.2: 158338836-158338836 on Build GRCh38
rs762363909 Chr.2: 158338952-158338952 on Build GRCh38
rs766481003 Chr.2: 158338882-158338882 on Build GRCh38
rs770707281 Chr.2: 158338850-158338850 on Build GRCh38
rs771590238 Chr.2: 158338859-158338859 on Build GRCh38
rs747632099 Chr.2: 158339049-158339049 on Build GRCh38
rs1040349319 Chr.2: 158338616-158338616 on Build GRCh38
rs549989208 Chr.2: 158338863-158338863 on Build GRCh38
rs749112227 Chr.2: 158338900-158338900 on Build GRCh38
rs112804540 Chr.2: 158339039-158339039 on Build GRCh38
rs757142860 Chr.2: 158339001-158339001 on Build GRCh38
rs753792696 Chr.2: 158338997-158338997 on Build GRCh38
rs754041768 Chr.2: 158338718-158338718 on Build GRCh38
rs774645951 Chr.2: 158339083-158339083 on Build GRCh38
rs112134546 Chr.2: 158338849-158338849 on Build GRCh38
rs779259308 Chr.2: 158338729-158338729 on Build GRCh38
rs777003362 Chr.2: 158338872-158338872 on Build GRCh38
rs765641765 Chr.2: 158338874-158338874 on Build GRCh38
rs769501490 Chr.2: 158338750-158338750 on Build GRCh38
rs764707328 Chr.2: 158338867-158338867 on Build GRCh38
rs745548518 Chr.2: 158338939-158338939 on Build GRCh38
rs770316020 Chr.2: 158338953-158338953 on Build GRCh38
rs768238108 Chr.2: 158339042-158339042 on Build GRCh38
rs557543093 Chr.2: 158338976-158338976 on Build GRCh38
rs200460550 Chr.2: 158338782-158338782 on Build GRCh38
rs144579063 Chr.2: 158338801-158338801 on Build GRCh38
rs1010757372 Chr.2: 158338948-158338948 on Build GRCh38
rs182534397 Chr.2: 158338654-158338654 on Build GRCh38
rs561338945 Chr.2: 158338623-158338623 on Build GRCh38
rs778134729 Chr.2: 158338714-158338714 on Build GRCh38
rs61746487 Chr.2: 158339050-158339050 on Build GRCh38
rs1041932009 Chr.2: 158338659-158338659 on Build GRCh38
rs771945616 Chr.2: 158338712-158338716 on Build GRCh38
rs779765881 Chr.2: 158339008-158339008 on Build GRCh38
rs746858205 Chr.2: 158338926-158338930 on Build GRCh38
rs139201830 Chr.2: 158338796-158338796 on Build GRCh38
rs758655252 Chr.2: 158338881-158338881 on Build GRCh38
rs750609933 Chr.2: 158338808-158338808 on Build GRCh38
rs750698110 Chr.2: 158338878-158338878 on Build GRCh38
rs370640297 Chr.2: 158338759-158338759 on Build GRCh38
rs1035725105 Chr.2: 158338724-158338724 on Build GRCh38
rs771134571 Chr.2: 158339056-158339056 on Build GRCh38
rs777210859 Chr.2: 158338950-158338950 on Build GRCh38
rs1035135080 Chr.2: 158339033-158339033 on Build GRCh38
rs768079688 Chr.2: 158338702-158338702 on Build GRCh38
rs771749465 Chr.2: 158338940-158338940 on Build GRCh38
rs763479278 Chr.2: 158339052-158339052 on Build GRCh38
rs773753646 Chr.2: 158338973-158338973 on Build GRCh38
rs372038223 Chr.2: 158338792-158338792 on Build GRCh38
rs186616119 Chr.2: 158338945-158338945 on Build GRCh38
rs773979843 Chr.2: 158338771-158338771 on Build GRCh38
rs369988339 Chr.2: 158338685-158338685 on Build GRCh38
rs746007716 Chr.2: 158338730-158338730 on Build GRCh38
rs751709913 Chr.2: 158338884-158338884 on Build GRCh38
rs773995905 Chr.2: 158338856-158338856 on Build GRCh38
rs745463750 Chr.2: 158338858-158338858 on Build GRCh38
rs201088412 Chr.2: 158338823-158338823 on Build GRCh38
rs370762751 Chr.2: 158338843-158338843 on Build GRCh38
rs772278221 Chr.2: 158338732-158338732 on Build GRCh38
rs778787632 Chr.2: 158339002-158339002 on Build GRCh38
rs201223553 Chr.2: 158338778-158338778 on Build GRCh38
rs377043763 Chr.2: 158339089-158339089 on Build GRCh38
rs757056617 Chr.2: 158338920-158338920 on Build GRCh38
rs1009324109 Chr.2: 158338615-158338615 on Build GRCh38
rs376842792 Chr.2: 158339005-158339005 on Build GRCh38
rs969568758 Chr.2: 158338962-158338962 on Build GRCh38
rs779853044 Chr.2: 158338911-158338911 on Build GRCh38
rs375854120 Chr.2: 158338833-158338833 on Build GRCh38
rs748068879 Chr.2: 158338827-158338827 on Build GRCh38

View Full Product Details