Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159320079-159320623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554055 c.6470T>G p.F2157C Chr.2: 159320302-159320302 on Build GRCh38
4085876 c.6374T>C p.F2125S Chr.2: 159320398-159320398 on Build GRCh38
1400332 c.6479delA p.K2160fs*8 Chr.2: 159320293-159320293 on Build GRCh38
7430233 c.6444A>G p.R2148R Chr.2: 159320328-159320328 on Build GRCh38
4455042 c.6486A>T p.T2162T Chr.2: 159320286-159320286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529278382 Chr.2: 159320387-159320387 on Build GRCh38
rs200199378 Chr.2: 159320529-159320529 on Build GRCh38
rs751940695 Chr.2: 159320434-159320434 on Build GRCh38
rs745812306 Chr.2: 159320449-159320449 on Build GRCh38
rs1000852504 Chr.2: 159320340-159320340 on Build GRCh38
rs781444864 Chr.2: 159320439-159320439 on Build GRCh38
rs570192302 Chr.2: 159320555-159320555 on Build GRCh38
rs758932030 Chr.2: 159320410-159320410 on Build GRCh38
rs185581125 Chr.2: 159320456-159320456 on Build GRCh38
rs904287556 Chr.2: 159320134-159320135 on Build GRCh38
rs1025674972 Chr.2: 159320347-159320347 on Build GRCh38
rs781189615 Chr.2: 159320240-159320240 on Build GRCh38
rs573401351 Chr.2: 159320186-159320186 on Build GRCh38
rs1017174728 Chr.2: 159320465-159320465 on Build GRCh38
rs750989169 Chr.2: 159320407-159320407 on Build GRCh38
rs551990263 Chr.2: 159320554-159320554 on Build GRCh38
rs570129428 Chr.2: 159320151-159320151 on Build GRCh38
rs943890948 Chr.2: 159320549-159320549 on Build GRCh38
rs763786685 Chr.2: 159320343-159320343 on Build GRCh38
rs765892621 Chr.2: 159320393-159320393 on Build GRCh38
rs189135439 Chr.2: 159320546-159320546 on Build GRCh38
rs777795050 Chr.2: 159320448-159320448 on Build GRCh38
rs1056073335 Chr.2: 159320359-159320359 on Build GRCh38
rs920924239 Chr.2: 159320174-159320174 on Build GRCh38
rs945145655 Chr.2: 159320260-159320260 on Build GRCh38
rs748226163 Chr.2: 159320444-159320444 on Build GRCh38
rs375763059 Chr.2: 159320294-159320294 on Build GRCh38
rs983304589 Chr.2: 159320481-159320481 on Build GRCh38
rs955305860 Chr.2: 159320559-159320559 on Build GRCh38
rs770456868 Chr.2: 159320239-159320239 on Build GRCh38
rs933679913 Chr.2: 159320185-159320185 on Build GRCh38
rs192683214 Chr.2: 159320278-159320278 on Build GRCh38
rs975719513 Chr.2: 159320537-159320537 on Build GRCh38
rs998970679 Chr.2: 159320135-159320135 on Build GRCh38
rs11551107 Chr.2: 159320144-159320144 on Build GRCh38
rs745761107 Chr.2: 159320299-159320299 on Build GRCh38
rs866243610 Chr.2: 159320413-159320413 on Build GRCh38
rs980782935 Chr.2: 159320436-159320436 on Build GRCh38
rs760254838 Chr.2: 159320342-159320342 on Build GRCh38
rs774253537 Chr.2: 159320297-159320297 on Build GRCh38
rs951401711 Chr.2: 159320446-159320446 on Build GRCh38
rs374903873 Chr.2: 159320453-159320453 on Build GRCh38
rs540750870 Chr.2: 159320217-159320217 on Build GRCh38
rs546466546 Chr.2: 159320102-159320102 on Build GRCh38
rs776067541 Chr.2: 159320348-159320348 on Build GRCh38
rs755224224 Chr.2: 159320437-159320437 on Build GRCh38
rs174229 Chr.2: 159320432-159320432 on Build GRCh38
rs904723339 Chr.2: 159320296-159320296 on Build GRCh38
rs1031419354 Chr.2: 159320148-159320148 on Build GRCh38
rs775203155 Chr.2: 159320336-159320336 on Build GRCh38
rs779352538 Chr.2: 159320292-159320293 on Build GRCh38
rs752280419 Chr.2: 159320237-159320237 on Build GRCh38
rs78844759 Chr.2: 159320128-159320128 on Build GRCh38
rs755172123 Chr.2: 159320221-159320221 on Build GRCh38
rs748255710 Chr.2: 159320263-159320263 on Build GRCh38
rs11551106 Chr.2: 159320254-159320254 on Build GRCh38
rs756211305 Chr.2: 159320447-159320447 on Build GRCh38
rs530612704 Chr.2: 159320552-159320552 on Build GRCh38
rs913624881 Chr.2: 159320191-159320191 on Build GRCh38
rs771941856 Chr.2: 159320461-159320461 on Build GRCh38
rs771888475 Chr.2: 159320310-159320310 on Build GRCh38

View Full Product Details