Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159324442-159324953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554471 c.6229G>A p.E2077K Chr.2: 159324935-159324935 on Build GRCh38
4085878 c.6267A>G p.V2089V Chr.2: 159324897-159324897 on Build GRCh38
1400333 c.6319T>G p.F2107V Chr.2: 159324845-159324845 on Build GRCh38
4085877 c.6348T>C p.S2116S Chr.2: 159324816-159324816 on Build GRCh38
1008141 c.6210-16_6232del39 p.? Chr.2: 159324932-159324970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038862889 Chr.2: 159324655-159324655 on Build GRCh38
rs768228502 Chr.2: 159324779-159324779 on Build GRCh38
rs906265631 Chr.2: 159324564-159324564 on Build GRCh38
rs1026374155 Chr.2: 159324751-159324751 on Build GRCh38
rs752998931 Chr.2: 159324900-159324900 on Build GRCh38
rs1011311164 Chr.2: 159324783-159324783 on Build GRCh38
rs769459015 Chr.2: 159324814-159324814 on Build GRCh38
rs761323045 Chr.2: 159324786-159324786 on Build GRCh38
rs772779849 Chr.2: 159324815-159324815 on Build GRCh38
rs759922704 Chr.2: 159324880-159324880 on Build GRCh38
rs70994292 Chr.2: 159324682-159324689 on Build GRCh38
rs67030583 Chr.2: 159324626-159324651 on Build GRCh38
rs1049637619 Chr.2: 159324647-159324647 on Build GRCh38
rs901729069 Chr.2: 159324688-159324688 on Build GRCh38
rs939600144 Chr.2: 159324488-159324488 on Build GRCh38
rs530025864 Chr.2: 159324693-159324693 on Build GRCh38
rs72416884 Chr.2: 159324626-159324649 on Build GRCh38
rs955922034 Chr.2: 159324781-159324781 on Build GRCh38
rs1056436203 Chr.2: 159324521-159324521 on Build GRCh38
rs867314147 Chr.2: 159324651-159324651 on Build GRCh38
rs749368294 Chr.2: 159324642-159324665 on Build GRCh38
rs60009917 Chr.2: 159324664-159324689 on Build GRCh38
rs866491417 Chr.2: 159324664-159324664 on Build GRCh38
rs776324386 Chr.2: 159324784-159324784 on Build GRCh38
rs993887166 Chr.2: 159324750-159324751 on Build GRCh38
rs919286228 Chr.2: 159324538-159324538 on Build GRCh38
rs987272965 Chr.2: 159324856-159324856 on Build GRCh38
rs756264266 Chr.2: 159324906-159324906 on Build GRCh38
rs766990267 Chr.2: 159324866-159324866 on Build GRCh38
rs1003062318 Chr.2: 159324579-159324579 on Build GRCh38
rs532569192 Chr.2: 159324486-159324486 on Build GRCh38
rs371273841 Chr.2: 159324918-159324918 on Build GRCh38
rs945030768 Chr.2: 159324553-159324553 on Build GRCh38
rs963438003 Chr.2: 159324720-159324720 on Build GRCh38
rs73967816 Chr.2: 159324614-159324614 on Build GRCh38
rs559836187 Chr.2: 159324629-159324629 on Build GRCh38
rs887598895 Chr.2: 159324649-159324649 on Build GRCh38
rs1042169160 Chr.2: 159324560-159324560 on Build GRCh38
rs969811311 Chr.2: 159324820-159324820 on Build GRCh38
rs1013205107 Chr.2: 159324653-159324653 on Build GRCh38
rs763613970 Chr.2: 159324840-159324840 on Build GRCh38
rs775819575 Chr.2: 159324666-159324691 on Build GRCh38
rs751910757 Chr.2: 159324870-159324870 on Build GRCh38
rs1057295111 Chr.2: 159324608-159324608 on Build GRCh38
rs746812816 Chr.2: 159324778-159324778 on Build GRCh38
rs1031331316 Chr.2: 159324733-159324733 on Build GRCh38
rs779768532 Chr.2: 159324777-159324777 on Build GRCh38
rs538106370 Chr.2: 159324926-159324926 on Build GRCh38
rs188090321 Chr.2: 159324887-159324887 on Build GRCh38

View Full Product Details