Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159332530-159333065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511053.2 XM_011511054.2 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246002 c.5813G>A p.R1938Q Chr.2: 159332670-159332670 on Build GRCh38
1400334 c.5842C>A p.L1948I Chr.2: 159332641-159332641 on Build GRCh38
5015956 c.5937_5938insT p.A1980fs*2 Chr.2: 159332545-159332546 on Build GRCh38
6305252 c.5869C>G p.H1957D Chr.2: 159332614-159332614 on Build GRCh38
6299832 c.5830G>A p.E1944K Chr.2: 159332653-159332653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902208610 Chr.2: 159333026-159333026 on Build GRCh38
rs767013275 Chr.2: 159332713-159332713 on Build GRCh38
rs954590666 Chr.2: 159332741-159332741 on Build GRCh38
rs770088268 Chr.2: 159332598-159332598 on Build GRCh38
rs992972847 Chr.2: 159332734-159332734 on Build GRCh38
rs375203329 Chr.2: 159332718-159332718 on Build GRCh38
rs749294629 Chr.2: 159333038-159333041 on Build GRCh38
rs370209163 Chr.2: 159332857-159332857 on Build GRCh38
rs775458669 Chr.2: 159332771-159332771 on Build GRCh38
rs139171947 Chr.2: 159332719-159332719 on Build GRCh38
rs147228605 Chr.2: 159333032-159333032 on Build GRCh38
rs757609995 Chr.2: 159332793-159332793 on Build GRCh38
rs779259228 Chr.2: 159332725-159332725 on Build GRCh38
rs1030156198 Chr.2: 159333003-159333003 on Build GRCh38
rs766015568 Chr.2: 159332698-159332698 on Build GRCh38
rs748640091 Chr.2: 159332595-159332595 on Build GRCh38
rs937121660 Chr.2: 159332792-159332792 on Build GRCh38
rs201931006 Chr.2: 159332702-159332702 on Build GRCh38
rs779551972 Chr.2: 159332571-159332571 on Build GRCh38
rs113493764 Chr.2: 159332974-159332974 on Build GRCh38
rs988755762 Chr.2: 159332761-159332761 on Build GRCh38
rs746034674 Chr.2: 159332730-159332730 on Build GRCh38
rs973624344 Chr.2: 159332754-159332754 on Build GRCh38
rs751919333 Chr.2: 159332775-159332775 on Build GRCh38
rs757998702 Chr.2: 159332563-159332563 on Build GRCh38
rs781649546 Chr.2: 159332584-159332584 on Build GRCh38
rs566798826 Chr.2: 159332739-159332739 on Build GRCh38
rs774643086 Chr.2: 159332630-159332630 on Build GRCh38
rs748046322 Chr.2: 159332737-159332737 on Build GRCh38
rs772305162 Chr.2: 159332731-159332731 on Build GRCh38
rs1055229388 Chr.2: 159332586-159332586 on Build GRCh38
rs1016709898 Chr.2: 159332950-159332950 on Build GRCh38
rs750173148 Chr.2: 159332696-159332696 on Build GRCh38
rs771266885 Chr.2: 159332612-159332612 on Build GRCh38
rs778134470 Chr.2: 159332609-159332609 on Build GRCh38
rs751054456 Chr.2: 159332574-159332574 on Build GRCh38
rs750927997 Chr.2: 159332845-159332845 on Build GRCh38
rs762788790 Chr.2: 159332840-159332840 on Build GRCh38
rs775570448 Chr.2: 159332736-159332736 on Build GRCh38
rs543720581 Chr.2: 159332758-159332758 on Build GRCh38
rs1006649754 Chr.2: 159332714-159332714 on Build GRCh38
rs111450767 Chr.2: 159332604-159332604 on Build GRCh38
rs900329158 Chr.2: 159332885-159332885 on Build GRCh38
rs35846087 Chr.2: 159332791-159332792 on Build GRCh38
rs534390539 Chr.2: 159332842-159332842 on Build GRCh38
rs757597081 Chr.2: 159332724-159332724 on Build GRCh38
rs541511325 Chr.2: 159332864-159332864 on Build GRCh38
rs765028566 Chr.2: 159332555-159332555 on Build GRCh38
rs1010865316 Chr.2: 159332906-159332906 on Build GRCh38
rs913165345 Chr.2: 159332762-159332762 on Build GRCh38
rs751109595 Chr.2: 159332701-159332701 on Build GRCh38
rs755544449 Chr.2: 159332575-159332575 on Build GRCh38
rs761768005 Chr.2: 159332665-159332665 on Build GRCh38
rs900580052 Chr.2: 159332963-159332963 on Build GRCh38
rs764943316 Chr.2: 159332670-159332670 on Build GRCh38
rs192910254 Chr.2: 159332645-159332645 on Build GRCh38
rs759660853 Chr.2: 159332631-159332631 on Build GRCh38
rs559631260 Chr.2: 159333031-159333031 on Build GRCh38
rs978837544 Chr.2: 159332785-159332785 on Build GRCh38
rs559065547 Chr.2: 159332830-159332830 on Build GRCh38
rs776451242 Chr.2: 159332648-159332648 on Build GRCh38
rs762650109 Chr.2: 159332697-159332697 on Build GRCh38
rs927028809 Chr.2: 159332786-159332786 on Build GRCh38
rs929311780 Chr.2: 159332759-159332759 on Build GRCh38
rs750071542 Chr.2: 159332556-159332556 on Build GRCh38
rs896683883 Chr.2: 159332794-159332794 on Build GRCh38
rs996284594 Chr.2: 159333001-159333001 on Build GRCh38

View Full Product Details