Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159769131-159769678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD302
Entrez Gene ID
9936
Gene Name
CD302 molecule
Gene Aliases
BIMLEC, CLEC13A, DCL-1, DCL1
UniGene
Hs.130014
Gene Chromosome Location
Chr.2: 159768628-159798255 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

Gene Symbol
LY75-CD302
Entrez Gene ID
100526664
Gene Name
LY75-CD302 readthrough
Gene Aliases
-
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159768628-159904756 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

SNP Detail

SNP ID Chromosome Location
rs535380873 Chr.2: 159769381-159769381 on Build GRCh38
rs763894886 Chr.2: 159769333-159769334 on Build GRCh38
rs563611959 Chr.2: 159769612-159769612 on Build GRCh38
rs979352828 Chr.2: 159769489-159769489 on Build GRCh38
rs929971789 Chr.2: 159769578-159769578 on Build GRCh38
rs13939 Chr.2: 159769641-159769641 on Build GRCh38
rs899561989 Chr.2: 159769640-159769640 on Build GRCh38
rs58332704 Chr.2: 159769644-159769645 on Build GRCh38
rs912178052 Chr.2: 159769282-159769282 on Build GRCh38
rs10206114 Chr.2: 159769330-159769330 on Build GRCh38
rs909816479 Chr.2: 159769596-159769596 on Build GRCh38
rs892096731 Chr.2: 159769648-159769648 on Build GRCh38
rs142841729 Chr.2: 159769653-159769654 on Build GRCh38
rs138629857 Chr.2: 159769581-159769581 on Build GRCh38
rs992841846 Chr.2: 159769573-159769573 on Build GRCh38
rs550908482 Chr.2: 159769424-159769424 on Build GRCh38
rs1046924989 Chr.2: 159769284-159769284 on Build GRCh38
rs974219081 Chr.2: 159769221-159769221 on Build GRCh38
rs1050775798 Chr.2: 159769644-159769644 on Build GRCh38
rs756943936 Chr.2: 159769642-159769643 on Build GRCh38
rs1139546 Chr.2: 159769557-159769557 on Build GRCh38
rs546641272 Chr.2: 159769219-159769219 on Build GRCh38
rs997847368 Chr.2: 159769357-159769357 on Build GRCh38
rs1056471609 Chr.2: 159769414-159769414 on Build GRCh38
rs1057449019 Chr.2: 159769226-159769226 on Build GRCh38
rs946834248 Chr.2: 159769347-159769347 on Build GRCh38
rs978829942 Chr.2: 159769197-159769197 on Build GRCh38
rs922088054 Chr.2: 159769341-159769341 on Build GRCh38
rs534047941 Chr.2: 159769472-159769472 on Build GRCh38
rs1046973978 Chr.2: 159769583-159769583 on Build GRCh38
rs143135371 Chr.2: 159769468-159769468 on Build GRCh38
rs1020500877 Chr.2: 159769471-159769471 on Build GRCh38
rs775124033 Chr.2: 159769533-159769533 on Build GRCh38
rs555788276 Chr.2: 159769544-159769544 on Build GRCh38
rs1126851 Chr.2: 159769588-159769588 on Build GRCh38
rs929919611 Chr.2: 159769278-159769278 on Build GRCh38
rs13432 Chr.2: 159769216-159769216 on Build GRCh38
rs747189530 Chr.2: 159769280-159769280 on Build GRCh38
rs1039430998 Chr.2: 159769315-159769315 on Build GRCh38
rs888249404 Chr.2: 159769292-159769292 on Build GRCh38
rs962835858 Chr.2: 159769536-159769536 on Build GRCh38
rs765656996 Chr.2: 159769475-159769475 on Build GRCh38
rs983724977 Chr.2: 159769279-159769279 on Build GRCh38
rs1025778095 Chr.2: 159769552-159769552 on Build GRCh38
rs902513125 Chr.2: 159769378-159769378 on Build GRCh38
rs1044176597 Chr.2: 159769349-159769349 on Build GRCh38
rs927389822 Chr.2: 159769201-159769201 on Build GRCh38
rs1005390371 Chr.2: 159769295-159769295 on Build GRCh38
rs1043824137 Chr.2: 159769647-159769648 on Build GRCh38
rs968765204 Chr.2: 159769172-159769172 on Build GRCh38
rs34620033 Chr.2: 159769643-159769644 on Build GRCh38
rs917292078 Chr.2: 159769574-159769574 on Build GRCh38
rs868321257 Chr.2: 159769367-159769367 on Build GRCh38
rs776957462 Chr.2: 159769345-159769345 on Build GRCh38
rs186975609 Chr.2: 159769163-159769163 on Build GRCh38
rs201615629 Chr.2: 159769642-159769642 on Build GRCh38
rs539679199 Chr.2: 159769463-159769463 on Build GRCh38
rs974935374 Chr.2: 159769639-159769640 on Build GRCh38
rs1039971098 Chr.2: 159769639-159769639 on Build GRCh38
rs912210543 Chr.2: 159769600-159769600 on Build GRCh38
rs199559120 Chr.2: 159769643-159769643 on Build GRCh38
rs917157977 Chr.2: 159769273-159769273 on Build GRCh38
rs772128768 Chr.2: 159769190-159769190 on Build GRCh38
rs929609859 Chr.2: 159769275-159769275 on Build GRCh38
rs75658831 Chr.2: 159769469-159769469 on Build GRCh38
rs569198171 Chr.2: 159769452-159769452 on Build GRCh38

View Full Product Details