Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159797915-159798407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD302
Entrez Gene ID
9936
Gene Name
CD302 molecule
Gene Aliases
BIMLEC, CLEC13A, DCL-1, DCL1
UniGene
Hs.130014
Gene Chromosome Location
Chr.2: 159768628-159798255 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

Gene Symbol
LY75-CD302
Entrez Gene ID
100526664
Gene Name
LY75-CD302 readthrough
Gene Aliases
-
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159768628-159904756 on Build GRCh38
Transcripts
NM_001198763.1 NM_001198764.1 NM_014880.4 NM_001198759.1 NM_001198760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217782 c.48C>T p.L16L Chr.2: 159798151-159798151 on Build GRCh38
6366124 c.67G>T p.D23Y Chr.2: 159798132-159798132 on Build GRCh38
5649367 c.20C>T p.P7L Chr.2: 159798179-159798179 on Build GRCh38
5822151 c.19C>T p.P7S Chr.2: 159798180-159798180 on Build GRCh38
6422342 c.10G>A p.A4T Chr.2: 159798189-159798189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559349775 Chr.2: 159798324-159798324 on Build GRCh38
rs568058533 Chr.2: 159798021-159798021 on Build GRCh38
rs1016752896 Chr.2: 159798050-159798050 on Build GRCh38
rs995661589 Chr.2: 159798236-159798236 on Build GRCh38
rs1003919519 Chr.2: 159798009-159798009 on Build GRCh38
rs948707919 Chr.2: 159798202-159798202 on Build GRCh38
rs914552369 Chr.2: 159798177-159798177 on Build GRCh38
rs529650649 Chr.2: 159798358-159798358 on Build GRCh38
rs536095045 Chr.2: 159798296-159798297 on Build GRCh38
rs542251109 Chr.2: 159798312-159798312 on Build GRCh38
rs2556106 Chr.2: 159798013-159798013 on Build GRCh38
rs1015448915 Chr.2: 159797994-159797994 on Build GRCh38
rs989955091 Chr.2: 159798214-159798214 on Build GRCh38
rs1048718822 Chr.2: 159798361-159798361 on Build GRCh38
rs961177543 Chr.2: 159798051-159798051 on Build GRCh38
rs374135673 Chr.2: 159798186-159798186 on Build GRCh38
rs890849734 Chr.2: 159798243-159798243 on Build GRCh38
rs368356224 Chr.2: 159798302-159798302 on Build GRCh38
rs111456791 Chr.2: 159798279-159798279 on Build GRCh38
rs1018022690 Chr.2: 159798300-159798301 on Build GRCh38
rs369799589 Chr.2: 159798298-159798298 on Build GRCh38
rs537100673 Chr.2: 159798071-159798071 on Build GRCh38
rs747230826 Chr.2: 159798192-159798192 on Build GRCh38
rs138846154 Chr.2: 159798196-159798196 on Build GRCh38
rs985533517 Chr.2: 159798053-159798053 on Build GRCh38
rs1026697021 Chr.2: 159798246-159798246 on Build GRCh38
rs924807620 Chr.2: 159798094-159798094 on Build GRCh38
rs114953340 Chr.2: 159798257-159798257 on Build GRCh38
rs755751360 Chr.2: 159798112-159798112 on Build GRCh38
rs1038251555 Chr.2: 159798298-159798299 on Build GRCh38
rs945518173 Chr.2: 159798165-159798165 on Build GRCh38
rs993918681 Chr.2: 159798054-159798054 on Build GRCh38
rs530428043 Chr.2: 159797986-159797986 on Build GRCh38
rs929926505 Chr.2: 159798375-159798375 on Build GRCh38
rs757802841 Chr.2: 159798146-159798146 on Build GRCh38
rs556674325 Chr.2: 159798293-159798294 on Build GRCh38
rs948827395 Chr.2: 159798299-159798299 on Build GRCh38
rs548897979 Chr.2: 159798365-159798365 on Build GRCh38
rs760644038 Chr.2: 159798075-159798075 on Build GRCh38
rs368187159 Chr.2: 159798108-159798108 on Build GRCh38
rs1002672525 Chr.2: 159797989-159797989 on Build GRCh38
rs1029371382 Chr.2: 159798241-159798242 on Build GRCh38
rs374871936 Chr.2: 159798345-159798345 on Build GRCh38
rs777168646 Chr.2: 159798113-159798113 on Build GRCh38
rs991349977 Chr.2: 159798325-159798325 on Build GRCh38
rs897014816 Chr.2: 159798233-159798233 on Build GRCh38
rs976199315 Chr.2: 159798083-159798083 on Build GRCh38
rs961394177 Chr.2: 159798006-159798006 on Build GRCh38
rs183102695 Chr.2: 159798300-159798300 on Build GRCh38
rs927995526 Chr.2: 159798193-159798193 on Build GRCh38
rs993928709 Chr.2: 159798237-159798237 on Build GRCh38
rs758418643 Chr.2: 159798079-159798079 on Build GRCh38
rs924772981 Chr.2: 159798295-159798295 on Build GRCh38
rs953937082 Chr.2: 159798252-159798252 on Build GRCh38
rs776626573 Chr.2: 159798209-159798209 on Build GRCh38
rs957349751 Chr.2: 159798208-159798208 on Build GRCh38
rs966869545 Chr.2: 159798295-159798296 on Build GRCh38
rs536617882 Chr.2: 159798255-159798260 on Build GRCh38
rs1044422889 Chr.2: 159798212-159798212 on Build GRCh38
rs746290027 Chr.2: 159798176-159798176 on Build GRCh38
rs907089638 Chr.2: 159798221-159798221 on Build GRCh38
rs1019259589 Chr.2: 159798259-159798259 on Build GRCh38
rs377678382 Chr.2: 159797961-159797961 on Build GRCh38
rs1002808735 Chr.2: 159798225-159798225 on Build GRCh38
rs1013240178 Chr.2: 159797975-159797975 on Build GRCh38
rs867228181 Chr.2: 159798149-159798149 on Build GRCh38
rs953777728 Chr.2: 159798046-159798046 on Build GRCh38
rs552282127 Chr.2: 159798179-159798179 on Build GRCh38
rs1036990041 Chr.2: 159798328-159798328 on Build GRCh38
rs569958676 Chr.2: 159798016-159798016 on Build GRCh38
rs761977627 Chr.2: 159798222-159798222 on Build GRCh38
rs779390887 Chr.2: 159798164-159798164 on Build GRCh38
rs960896212 Chr.2: 159798314-159798314 on Build GRCh38
rs995318539 Chr.2: 159798354-159798354 on Build GRCh38
rs766478080 Chr.2: 159798227-159798227 on Build GRCh38
rs146923392 Chr.2: 159798294-159798295 on Build GRCh38
rs906565740 Chr.2: 159798003-159798003 on Build GRCh38
rs552622200 Chr.2: 159798200-159798201 on Build GRCh38
rs938567065 Chr.2: 159798302-159798302 on Build GRCh38
rs536588390 Chr.2: 159798296-159798296 on Build GRCh38
rs574736662 Chr.2: 159798294-159798294 on Build GRCh38
rs941298944 Chr.2: 159798219-159798219 on Build GRCh38
rs554888596 Chr.2: 159798297-159798297 on Build GRCh38
rs1055506501 Chr.2: 159798203-159798203 on Build GRCh38
rs373836907 Chr.2: 159798329-159798329 on Build GRCh38
rs910771153 Chr.2: 159798070-159798070 on Build GRCh38
rs528076445 Chr.2: 159797981-159797981 on Build GRCh38
rs773758915 Chr.2: 159798137-159798161 on Build GRCh38
rs576882730 Chr.2: 159798301-159798301 on Build GRCh38
rs1036847340 Chr.2: 159798231-159798231 on Build GRCh38

View Full Product Details