Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 163608143-163608641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGN
Entrez Gene ID
55137
Gene Name
fidgetin, microtubule severing factor
Gene Aliases
-
UniGene
Hs.593650
Gene Chromosome Location
Chr.2: 163602611-163736155 on Build GRCh38
Transcripts
NM_001321825.1 NM_018086.3 XM_011511389.2 XM_017004419.1

SNP Detail

SNP ID Chromosome Location
rs912970368 Chr.2: 163608251-163608251 on Build GRCh38
rs1056568291 Chr.2: 163608436-163608436 on Build GRCh38
rs756793897 Chr.2: 163608266-163608267 on Build GRCh38
rs570104543 Chr.2: 163608455-163608455 on Build GRCh38
rs886435039 Chr.2: 163608201-163608201 on Build GRCh38
rs192578170 Chr.2: 163608460-163608460 on Build GRCh38
rs535436207 Chr.2: 163608190-163608190 on Build GRCh38
rs912131672 Chr.2: 163608499-163608499 on Build GRCh38
rs914091278 Chr.2: 163608523-163608523 on Build GRCh38
rs764946745 Chr.2: 163608354-163608354 on Build GRCh38
rs1015181969 Chr.2: 163608413-163608413 on Build GRCh38
rs1000169855 Chr.2: 163608309-163608309 on Build GRCh38
rs948106779 Chr.2: 163608461-163608461 on Build GRCh38
rs999816182 Chr.2: 163608575-163608575 on Build GRCh38
rs558768584 Chr.2: 163608474-163608474 on Build GRCh38
rs202186132 Chr.2: 163608256-163608257 on Build GRCh38
rs556872756 Chr.2: 163608314-163608314 on Build GRCh38
rs974575966 Chr.2: 163608372-163608372 on Build GRCh38
rs532128095 Chr.2: 163608207-163608207 on Build GRCh38
rs1042681762 Chr.2: 163608265-163608265 on Build GRCh38
rs953572206 Chr.2: 163608195-163608195 on Build GRCh38
rs10209354 Chr.2: 163608267-163608267 on Build GRCh38
rs962096489 Chr.2: 163608418-163608418 on Build GRCh38
rs966724544 Chr.2: 163608500-163608500 on Build GRCh38
rs955891299 Chr.2: 163608353-163608353 on Build GRCh38
rs994924718 Chr.2: 163608422-163608422 on Build GRCh38
rs148690140 Chr.2: 163608257-163608257 on Build GRCh38
rs867305094 Chr.2: 163608396-163608396 on Build GRCh38
rs964527473 Chr.2: 163608370-163608370 on Build GRCh38
rs1008655900 Chr.2: 163608359-163608359 on Build GRCh38
rs907876540 Chr.2: 163608404-163608404 on Build GRCh38
rs550264542 Chr.2: 163608381-163608381 on Build GRCh38
rs187289827 Chr.2: 163608366-163608366 on Build GRCh38
rs988298416 Chr.2: 163608501-163608501 on Build GRCh38
rs878856274 Chr.2: 163608213-163608213 on Build GRCh38
rs552226490 Chr.2: 163608218-163608218 on Build GRCh38
rs939307613 Chr.2: 163608406-163608406 on Build GRCh38
rs76144574 Chr.2: 163608224-163608224 on Build GRCh38
rs566031126 Chr.2: 163608480-163608480 on Build GRCh38
rs567689988 Chr.2: 163608364-163608364 on Build GRCh38
rs565621269 Chr.2: 163608219-163608219 on Build GRCh38
rs562512848 Chr.2: 163608169-163608169 on Build GRCh38
rs34917512 Chr.2: 163608281-163608282 on Build GRCh38
rs983763679 Chr.2: 163608397-163608397 on Build GRCh38

View Full Product Details