Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 163611665-163612164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGN
Entrez Gene ID
55137
Gene Name
fidgetin, microtubule severing factor
Gene Aliases
-
UniGene
Hs.593650
Gene Chromosome Location
Chr.2: 163602611-163736155 on Build GRCh38
Transcripts
NM_001321825.1 NM_018086.3 XM_011511389.2 XM_017004419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1400542 c.44C>T p.T15M Chr.2: 163611788-163611788 on Build GRCh38
6307399 c.86C>T p.T29I Chr.2: 163611746-163611746 on Build GRCh38
5015960 c.82delA p.I28fs*35 Chr.2: 163611750-163611750 on Build GRCh38
1482099 c.55G>T p.A19S Chr.2: 163611777-163611777 on Build GRCh38
6199288 c.153G>A p.Q51Q Chr.2: 163611679-163611679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199883388 Chr.2: 163611788-163611788 on Build GRCh38
rs752019959 Chr.2: 163611708-163611708 on Build GRCh38
rs147035155 Chr.2: 163611685-163611685 on Build GRCh38
rs2231900 Chr.2: 163611824-163611824 on Build GRCh38
rs780889086 Chr.2: 163612140-163612140 on Build GRCh38
rs541255073 Chr.2: 163612012-163612012 on Build GRCh38
rs918591884 Chr.2: 163611742-163611742 on Build GRCh38
rs184060478 Chr.2: 163611784-163611784 on Build GRCh38
rs372868068 Chr.2: 163611990-163611990 on Build GRCh38
rs774914442 Chr.2: 163611818-163611818 on Build GRCh38
rs745962085 Chr.2: 163611701-163611701 on Build GRCh38
rs922766020 Chr.2: 163612048-163612049 on Build GRCh38
rs368261519 Chr.2: 163611951-163611951 on Build GRCh38
rs985412025 Chr.2: 163612055-163612055 on Build GRCh38
rs2231899 Chr.2: 163611850-163611850 on Build GRCh38
rs766019629 Chr.2: 163611776-163611776 on Build GRCh38
rs201681537 Chr.2: 163611890-163611890 on Build GRCh38
rs74777091 Chr.2: 163611933-163611933 on Build GRCh38
rs865952685 Chr.2: 163611948-163611948 on Build GRCh38
rs981489151 Chr.2: 163612125-163612125 on Build GRCh38
rs745921022 Chr.2: 163611822-163611822 on Build GRCh38
rs902196651 Chr.2: 163611871-163611871 on Build GRCh38
rs1010660231 Chr.2: 163611758-163611758 on Build GRCh38
rs1053630725 Chr.2: 163611884-163611884 on Build GRCh38
rs372052490 Chr.2: 163612068-163612068 on Build GRCh38
rs572918511 Chr.2: 163611821-163611821 on Build GRCh38
rs1055079483 Chr.2: 163611967-163611967 on Build GRCh38
rs957909505 Chr.2: 163611707-163611707 on Build GRCh38
rs773838725 Chr.2: 163611851-163611851 on Build GRCh38
rs765780931 Chr.2: 163611734-163611734 on Build GRCh38
rs1020605317 Chr.2: 163612010-163612010 on Build GRCh38
rs1021102104 Chr.2: 163611765-163611765 on Build GRCh38
rs368129070 Chr.2: 163612036-163612036 on Build GRCh38
rs2231898 Chr.2: 163611904-163611904 on Build GRCh38
rs772541947 Chr.2: 163611753-163611753 on Build GRCh38
rs775910769 Chr.2: 163611704-163611704 on Build GRCh38
rs985308219 Chr.2: 163611849-163611849 on Build GRCh38
rs984145230 Chr.2: 163612027-163612027 on Build GRCh38
rs762187812 Chr.2: 163611727-163611727 on Build GRCh38
rs748560584 Chr.2: 163611847-163611847 on Build GRCh38
rs747180137 Chr.2: 163611836-163611836 on Build GRCh38
rs769193296 Chr.2: 163611721-163611721 on Build GRCh38
rs2231897 Chr.2: 163611927-163611927 on Build GRCh38
rs184485541 Chr.2: 163612046-163612046 on Build GRCh38
rs201486174 Chr.2: 163611828-163611829 on Build GRCh38
rs746213027 Chr.2: 163611846-163611847 on Build GRCh38
rs868581244 Chr.2: 163612061-163612061 on Build GRCh38
rs778814837 Chr.2: 163611699-163611699 on Build GRCh38
rs777077620 Chr.2: 163611726-163611726 on Build GRCh38
rs953840111 Chr.2: 163612050-163612050 on Build GRCh38
rs933090295 Chr.2: 163611845-163611845 on Build GRCh38
rs371250164 Chr.2: 163611919-163611919 on Build GRCh38
rs997888994 Chr.2: 163612032-163612032 on Build GRCh38
rs747372432 Chr.2: 163611715-163611715 on Build GRCh38
rs910121211 Chr.2: 163612033-163612033 on Build GRCh38
rs777777783 Chr.2: 163611820-163611820 on Build GRCh38
rs962644917 Chr.2: 163612069-163612069 on Build GRCh38
rs754740615 Chr.2: 163611787-163611787 on Build GRCh38
rs771540538 Chr.2: 163611817-163611818 on Build GRCh38
rs752565413 Chr.2: 163611806-163611806 on Build GRCh38
rs376725518 Chr.2: 163611703-163611703 on Build GRCh38
rs951572652 Chr.2: 163611814-163611814 on Build GRCh38
rs371299603 Chr.2: 163612111-163612111 on Build GRCh38
rs758616478 Chr.2: 163611823-163611823 on Build GRCh38
rs1053893218 Chr.2: 163612119-163612119 on Build GRCh38
rs375799242 Chr.2: 163611995-163611995 on Build GRCh38
rs762510306 Chr.2: 163611755-163611755 on Build GRCh38
rs776796045 Chr.2: 163611841-163611841 on Build GRCh38
rs780120401 Chr.2: 163612113-163612113 on Build GRCh38
rs2231901 Chr.2: 163611818-163611818 on Build GRCh38
rs2231896 Chr.2: 163612096-163612096 on Build GRCh38
rs756095635 Chr.2: 163611690-163611690 on Build GRCh38
rs964657906 Chr.2: 163612042-163612042 on Build GRCh38
rs555584457 Chr.2: 163611839-163611839 on Build GRCh38
rs552127215 Chr.2: 163612114-163612114 on Build GRCh38
rs1037131719 Chr.2: 163611918-163611918 on Build GRCh38
rs999184510 Chr.2: 163611876-163611876 on Build GRCh38
rs748081011 Chr.2: 163611689-163611689 on Build GRCh38

View Full Product Details