Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169520798-169521328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL41
Entrez Gene ID
10324
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10, Krp1, SARCOSIN
UniGene
Hs.50550
Gene Chromosome Location
Chr.2: 169509702-169526262 on Build GRCh38
Transcripts
NM_006063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5058247 c.1654A>G p.M552V Chr.2: 169520952-169520952 on Build GRCh38
5651682 c.1578C>T p.T526T Chr.2: 169520876-169520876 on Build GRCh38
3315203 c.1674A>G p.K558K Chr.2: 169520972-169520972 on Build GRCh38
5626776 c.1595G>C p.R532T Chr.2: 169520893-169520893 on Build GRCh38
1631437 c.1569T>C p.D523D Chr.2: 169520867-169520867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900388205 Chr.2: 169520849-169520849 on Build GRCh38
rs777120116 Chr.2: 169520972-169520972 on Build GRCh38
rs375214560 Chr.2: 169520997-169520997 on Build GRCh38
rs6738934 Chr.2: 169521227-169521227 on Build GRCh38
rs1042006125 Chr.2: 169521272-169521272 on Build GRCh38
rs74832281 Chr.2: 169520949-169520949 on Build GRCh38
rs181844196 Chr.2: 169521181-169521181 on Build GRCh38
rs766304026 Chr.2: 169520871-169520871 on Build GRCh38
rs951864108 Chr.2: 169520973-169520973 on Build GRCh38
rs919279802 Chr.2: 169521166-169521166 on Build GRCh38
rs773033779 Chr.2: 169521008-169521008 on Build GRCh38
rs113704776 Chr.2: 169520913-169520913 on Build GRCh38
rs762480893 Chr.2: 169521027-169521027 on Build GRCh38
rs900853962 Chr.2: 169521291-169521291 on Build GRCh38
rs983593653 Chr.2: 169520976-169520976 on Build GRCh38
rs111615063 Chr.2: 169520861-169520861 on Build GRCh38
rs762284386 Chr.2: 169520979-169520979 on Build GRCh38
rs570441759 Chr.2: 169520964-169520964 on Build GRCh38
rs748005003 Chr.2: 169520900-169520900 on Build GRCh38
rs113071956 Chr.2: 169520860-169520860 on Build GRCh38
rs915380216 Chr.2: 169521213-169521213 on Build GRCh38
rs990980662 Chr.2: 169521209-169521209 on Build GRCh38
rs970606845 Chr.2: 169521168-169521168 on Build GRCh38
rs566344019 Chr.2: 169521226-169521226 on Build GRCh38
rs74839160 Chr.2: 169521034-169521034 on Build GRCh38
rs757410091 Chr.2: 169520911-169520911 on Build GRCh38
rs370877619 Chr.2: 169520922-169520922 on Build GRCh38
rs914973685 Chr.2: 169521260-169521260 on Build GRCh38
rs981039731 Chr.2: 169521041-169521041 on Build GRCh38
rs766465411 Chr.2: 169521067-169521067 on Build GRCh38
rs373954289 Chr.2: 169520867-169520867 on Build GRCh38
rs1027488195 Chr.2: 169520877-169520877 on Build GRCh38
rs530632249 Chr.2: 169520885-169520885 on Build GRCh38
rs864309595 Chr.2: 169520937-169520937 on Build GRCh38
rs376724200 Chr.2: 169520888-169520888 on Build GRCh38
rs562170123 Chr.2: 169520876-169520876 on Build GRCh38
rs745908034 Chr.2: 169520925-169520925 on Build GRCh38
rs113285249 Chr.2: 169521170-169521170 on Build GRCh38
rs963483226 Chr.2: 169521029-169521029 on Build GRCh38
rs550333896 Chr.2: 169520939-169520939 on Build GRCh38
rs998188694 Chr.2: 169520919-169520919 on Build GRCh38
rs749898629 Chr.2: 169520862-169520862 on Build GRCh38
rs769519206 Chr.2: 169521019-169521023 on Build GRCh38
rs747958063 Chr.2: 169520832-169520833 on Build GRCh38
rs747439201 Chr.2: 169520960-169520960 on Build GRCh38
rs944699022 Chr.2: 169521278-169521278 on Build GRCh38
rs926439738 Chr.2: 169521192-169521192 on Build GRCh38
rs1007763828 Chr.2: 169520975-169520975 on Build GRCh38
rs777586212 Chr.2: 169521267-169521267 on Build GRCh38

View Full Product Details