Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169674525-169675036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC173
Entrez Gene ID
129881
Gene Name
coiled-coil domain containing 173
Gene Aliases
C2orf77
UniGene
Hs.370111
Gene Chromosome Location
Chr.2: 169645425-169694421 on Build GRCh38
Transcripts
NM_001085447.1 XM_011510589.1 XM_011510590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6206971 c.503G>A p.R168H Chr.2: 169674735-169674735 on Build GRCh38
7447359 c.448G>C p.E150Q Chr.2: 169674887-169674887 on Build GRCh38
1009345 c.331C>T p.R111C Chr.2: 169675004-169675004 on Build GRCh38
4667650 c.618delA p.K206fs*14 Chr.2: 169674620-169674620 on Build GRCh38
1401114 c.428A>G p.Q143R Chr.2: 169674907-169674907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763715447 Chr.2: 169674704-169674704 on Build GRCh38
rs370520932 Chr.2: 169674761-169674761 on Build GRCh38
rs754114168 Chr.2: 169674747-169674747 on Build GRCh38
rs375356931 Chr.2: 169674575-169674575 on Build GRCh38
rs781565724 Chr.2: 169674960-169674960 on Build GRCh38
rs370900443 Chr.2: 169674950-169674950 on Build GRCh38
rs928638821 Chr.2: 169674716-169674716 on Build GRCh38
rs750531318 Chr.2: 169674685-169674686 on Build GRCh38
rs757588572 Chr.2: 169674906-169674906 on Build GRCh38
rs200589066 Chr.2: 169674652-169674652 on Build GRCh38
rs376838379 Chr.2: 169674927-169674927 on Build GRCh38
rs959827780 Chr.2: 169674991-169674991 on Build GRCh38
rs749362148 Chr.2: 169674657-169674657 on Build GRCh38
rs771538661 Chr.2: 169674559-169674559 on Build GRCh38
rs1053578874 Chr.2: 169674685-169674685 on Build GRCh38
rs181014642 Chr.2: 169674808-169674808 on Build GRCh38
rs767431511 Chr.2: 169674884-169674884 on Build GRCh38
rs951452516 Chr.2: 169674690-169674690 on Build GRCh38
rs767780209 Chr.2: 169674598-169674598 on Build GRCh38
rs772432034 Chr.2: 169674771-169674771 on Build GRCh38
rs997263698 Chr.2: 169674892-169674892 on Build GRCh38
rs763558909 Chr.2: 169674606-169674606 on Build GRCh38
rs769077494 Chr.2: 169674781-169674781 on Build GRCh38
rs985701209 Chr.2: 169674572-169674572 on Build GRCh38
rs746348957 Chr.2: 169674767-169674767 on Build GRCh38
rs993178991 Chr.2: 169674889-169674889 on Build GRCh38
rs759335649 Chr.2: 169674874-169674874 on Build GRCh38
rs765147831 Chr.2: 169674618-169674618 on Build GRCh38
rs747322778 Chr.2: 169674775-169674775 on Build GRCh38
rs375540019 Chr.2: 169674998-169674998 on Build GRCh38
rs545621974 Chr.2: 169674869-169674869 on Build GRCh38
rs781120595 Chr.2: 169674882-169674885 on Build GRCh38
rs780422550 Chr.2: 169674932-169674932 on Build GRCh38
rs1051545954 Chr.2: 169674938-169674938 on Build GRCh38
rs947310916 Chr.2: 169674768-169674768 on Build GRCh38
rs775752655 Chr.2: 169674684-169674684 on Build GRCh38
rs377686934 Chr.2: 169675003-169675003 on Build GRCh38
rs762573033 Chr.2: 169674837-169674837 on Build GRCh38
rs762365403 Chr.2: 169675000-169675000 on Build GRCh38
rs753314829 Chr.2: 169674607-169674607 on Build GRCh38
rs756809578 Chr.2: 169674613-169674613 on Build GRCh38
rs925942525 Chr.2: 169674626-169674626 on Build GRCh38
rs914457324 Chr.2: 169674753-169674753 on Build GRCh38
rs771199136 Chr.2: 169674660-169674660 on Build GRCh38
rs772564753 Chr.2: 169674823-169674823 on Build GRCh38
rs750324427 Chr.2: 169674714-169674714 on Build GRCh38
rs775154607 Chr.2: 169674604-169674604 on Build GRCh38
rs565351165 Chr.2: 169674945-169674945 on Build GRCh38
rs758422921 Chr.2: 169674755-169674757 on Build GRCh38
rs199751298 Chr.2: 169674729-169674729 on Build GRCh38
rs369756121 Chr.2: 169674619-169674619 on Build GRCh38
rs779239363 Chr.2: 169674914-169674914 on Build GRCh38
rs765271237 Chr.2: 169674635-169674637 on Build GRCh38
rs761475032 Chr.2: 169674802-169674802 on Build GRCh38
rs750451704 Chr.2: 169674922-169674922 on Build GRCh38
rs1016360334 Chr.2: 169674965-169674965 on Build GRCh38
rs769253135 Chr.2: 169674813-169674813 on Build GRCh38
rs766379493 Chr.2: 169674728-169674728 on Build GRCh38
rs267599001 Chr.2: 169674715-169674715 on Build GRCh38
rs368546077 Chr.2: 169674735-169674735 on Build GRCh38
rs780660297 Chr.2: 169674774-169674774 on Build GRCh38
rs764737812 Chr.2: 169674713-169674713 on Build GRCh38
rs749415366 Chr.2: 169674557-169674557 on Build GRCh38
rs761309902 Chr.2: 169674709-169674709 on Build GRCh38
rs754874295 Chr.2: 169674736-169674736 on Build GRCh38
rs898800118 Chr.2: 169674880-169674880 on Build GRCh38
rs774929204 Chr.2: 169674664-169674664 on Build GRCh38
rs748548666 Chr.2: 169674971-169674971 on Build GRCh38
rs748361850 Chr.2: 169674650-169674650 on Build GRCh38
rs376884574 Chr.2: 169674673-169674673 on Build GRCh38
rs768957180 Chr.2: 169674956-169674956 on Build GRCh38
rs769305324 Chr.2: 169674987-169674987 on Build GRCh38
rs758152092 Chr.2: 169674620-169674620 on Build GRCh38
rs757357171 Chr.2: 169674752-169674752 on Build GRCh38
rs754066987 Chr.2: 169674887-169674887 on Build GRCh38
rs766208972 Chr.2: 169674855-169674855 on Build GRCh38
rs776324974 Chr.2: 169674705-169674705 on Build GRCh38
rs373350614 Chr.2: 169674786-169674786 on Build GRCh38
rs779734131 Chr.2: 169674633-169674633 on Build GRCh38
rs761146574 Chr.2: 169674686-169674686 on Build GRCh38
rs772435536 Chr.2: 169674674-169674674 on Build GRCh38
rs760271101 Chr.2: 169674605-169674605 on Build GRCh38
rs778133148 Chr.2: 169674740-169674740 on Build GRCh38
rs951590184 Chr.2: 169675005-169675005 on Build GRCh38
rs759938720 Chr.2: 169674578-169674578 on Build GRCh38
rs774095741 Chr.2: 169675004-169675004 on Build GRCh38

View Full Product Details