Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171522600-171523092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBRD1
Entrez Gene ID
79901
Gene Name
cytochrome b reductase 1
Gene Aliases
CYB561A2, DCYTB, FRRS3
UniGene
Hs.221941
Gene Chromosome Location
Chr.2: 171522247-171558133 on Build GRCh38
Transcripts
NM_001127383.1 NM_001256909.1 NM_024843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3569989 c.180C>T p.F60F Chr.2: 171522725-171522725 on Build GRCh38
6412591 c.169G>A p.G57S Chr.2: 171522714-171522714 on Build GRCh38
3714038 c.89G>T p.W30L Chr.2: 171522634-171522634 on Build GRCh38
5690485 c.136G>C p.E46Q Chr.2: 171522681-171522681 on Build GRCh38
4470917 c.168C>G p.T56T Chr.2: 171522713-171522713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775711731 Chr.2: 171522649-171522649 on Build GRCh38
rs576298473 Chr.2: 171522889-171522889 on Build GRCh38
rs1017584530 Chr.2: 171522678-171522678 on Build GRCh38
rs1018073892 Chr.2: 171523057-171523057 on Build GRCh38
rs1035842150 Chr.2: 171522968-171522968 on Build GRCh38
rs757599014 Chr.2: 171522759-171522759 on Build GRCh38
rs746335457 Chr.2: 171522640-171522640 on Build GRCh38
rs779171072 Chr.2: 171522630-171522630 on Build GRCh38
rs1041370527 Chr.2: 171522754-171522755 on Build GRCh38
rs887770495 Chr.2: 171522848-171522848 on Build GRCh38
rs962529305 Chr.2: 171522901-171522901 on Build GRCh38
rs776216750 Chr.2: 171522674-171522674 on Build GRCh38
rs1014970369 Chr.2: 171522899-171522899 on Build GRCh38
rs769020511 Chr.2: 171522667-171522667 on Build GRCh38
rs914281678 Chr.2: 171523015-171523015 on Build GRCh38
rs934841865 Chr.2: 171523008-171523008 on Build GRCh38
rs533268769 Chr.2: 171523016-171523016 on Build GRCh38
rs368999568 Chr.2: 171522762-171522762 on Build GRCh38
rs761485444 Chr.2: 171522677-171522677 on Build GRCh38
rs868480546 Chr.2: 171522726-171522726 on Build GRCh38
rs1003020888 Chr.2: 171522906-171522906 on Build GRCh38
rs941072134 Chr.2: 171522814-171522814 on Build GRCh38
rs762797801 Chr.2: 171523028-171523028 on Build GRCh38
rs761032280 Chr.2: 171522656-171522656 on Build GRCh38
rs764696712 Chr.2: 171522692-171522692 on Build GRCh38
rs780551986 Chr.2: 171522778-171522778 on Build GRCh38
rs958973609 Chr.2: 171522993-171522993 on Build GRCh38
rs924358424 Chr.2: 171523025-171523025 on Build GRCh38
rs991602555 Chr.2: 171522852-171522852 on Build GRCh38
rs771090771 Chr.2: 171522629-171522629 on Build GRCh38
rs754748261 Chr.2: 171522720-171522720 on Build GRCh38
rs376668865 Chr.2: 171523020-171523020 on Build GRCh38
rs767447864 Chr.2: 171522723-171522723 on Build GRCh38
rs758411421 Chr.2: 171522763-171522763 on Build GRCh38
rs553321272 Chr.2: 171522753-171522754 on Build GRCh38
rs537360619 Chr.2: 171522898-171522898 on Build GRCh38
rs553503102 Chr.2: 171522707-171522707 on Build GRCh38
rs909662688 Chr.2: 171522790-171522790 on Build GRCh38
rs951214472 Chr.2: 171522738-171522738 on Build GRCh38
rs201989257 Chr.2: 171522648-171522648 on Build GRCh38
rs1040905706 Chr.2: 171523044-171523044 on Build GRCh38
rs760626194 Chr.2: 171523009-171523009 on Build GRCh38
rs200632435 Chr.2: 171522714-171522714 on Build GRCh38
rs915572547 Chr.2: 171522878-171522878 on Build GRCh38
rs558005396 Chr.2: 171522745-171522745 on Build GRCh38
rs555499880 Chr.2: 171522979-171522979 on Build GRCh38
rs747492380 Chr.2: 171522786-171522786 on Build GRCh38
rs766408302 Chr.2: 171522711-171522711 on Build GRCh38
rs748192522 Chr.2: 171522762-171522773 on Build GRCh38
rs996699517 Chr.2: 171522828-171522828 on Build GRCh38
rs1045983389 Chr.2: 171522964-171522964 on Build GRCh38
rs1028977732 Chr.2: 171522833-171522833 on Build GRCh38
rs369799517 Chr.2: 171523023-171523023 on Build GRCh38
rs957759728 Chr.2: 171522800-171522800 on Build GRCh38
rs943886078 Chr.2: 171523034-171523034 on Build GRCh38
rs779120242 Chr.2: 171522760-171522760 on Build GRCh38
rs749538088 Chr.2: 171522626-171522626 on Build GRCh38
rs991177451 Chr.2: 171522689-171522689 on Build GRCh38
rs749999527 Chr.2: 171522694-171522694 on Build GRCh38
rs372089033 Chr.2: 171522737-171522737 on Build GRCh38
rs762788179 Chr.2: 171522710-171522710 on Build GRCh38

View Full Product Details